Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 109017625-109018124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM245
Entrez Gene ID
23731
Gene Name
transmembrane protein 245
Gene Aliases
C9orf5, CG-2, CG2
UniGene
Hs.308074
Gene Chromosome Location
Chr.9: 109015135-109119963 on Build GRCh38
Transcripts
NM_032012.3 XM_011518446.2 XM_011518447.1 XM_011518449.1 XM_011518451.1 XM_011518452.1 XM_017014571.1 XM_017014572.1

SNP Detail

SNP ID Chromosome Location
rs1012037466 Chr.9: 109017985-109017985 on Build GRCh38
rs143449947 Chr.9: 109017980-109017980 on Build GRCh38
rs915655965 Chr.9: 109017752-109017752 on Build GRCh38
rs575775390 Chr.9: 109018014-109018014 on Build GRCh38
rs755865638 Chr.9: 109017988-109017989 on Build GRCh38
rs991760074 Chr.9: 109017735-109017735 on Build GRCh38
rs933493709 Chr.9: 109017933-109017933 on Build GRCh38
rs1022086071 Chr.9: 109017688-109017688 on Build GRCh38
rs374552554 Chr.9: 109017655-109017655 on Build GRCh38
rs148840932 Chr.9: 109017879-109017879 on Build GRCh38
rs1011570818 Chr.9: 109017670-109017670 on Build GRCh38
rs79184925 Chr.9: 109018065-109018065 on Build GRCh38
rs578237985 Chr.9: 109017949-109017949 on Build GRCh38
rs773893389 Chr.9: 109017932-109017932 on Build GRCh38
rs959842770 Chr.9: 109017707-109017707 on Build GRCh38
rs796959036 Chr.9: 109017884-109017884 on Build GRCh38
rs983888540 Chr.9: 109017843-109017843 on Build GRCh38
rs938110279 Chr.9: 109017956-109017956 on Build GRCh38
rs926594911 Chr.9: 109017951-109017951 on Build GRCh38
rs908836955 Chr.9: 109017913-109017913 on Build GRCh38
rs142518974 Chr.9: 109017656-109017656 on Build GRCh38
rs1042242991 Chr.9: 109018084-109018084 on Build GRCh38
rs138479287 Chr.9: 109017698-109017698 on Build GRCh38
rs868682895 Chr.9: 109017934-109017934 on Build GRCh38
rs537115742 Chr.9: 109017657-109017657 on Build GRCh38
rs181001714 Chr.9: 109017692-109017692 on Build GRCh38
rs908173368 Chr.9: 109017861-109017861 on Build GRCh38
rs889230768 Chr.9: 109017665-109017665 on Build GRCh38
rs952607323 Chr.9: 109017781-109017781 on Build GRCh38
rs952876421 Chr.9: 109017870-109017870 on Build GRCh38
rs1008608066 Chr.9: 109017739-109017739 on Build GRCh38
rs973148412 Chr.9: 109017819-109017819 on Build GRCh38
rs150120875 Chr.9: 109017693-109017693 on Build GRCh38
rs761411321 Chr.9: 109018054-109018054 on Build GRCh38
rs890371809 Chr.9: 109017724-109017724 on Build GRCh38
rs545637841 Chr.9: 109017961-109017961 on Build GRCh38
rs769834993 Chr.9: 109017671-109017671 on Build GRCh38

View Full Product Details