Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110387187-110387721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SVEP1
Entrez Gene ID
79987
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Gene Aliases
C9orf13, CCP22, POLYDOM, SEL-OB, SELOB
UniGene
Hs.522334
Gene Chromosome Location
Chr.9: 110365248-110579880 on Build GRCh38
Transcripts
NM_153366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3847314 c.9902C>A p.T3301N Chr.9: 110387443-110387443 on Build GRCh38
3652811 c.10048C>T p.P3350S Chr.9: 110387297-110387297 on Build GRCh38
5027120 c.9910G>C p.E3304Q Chr.9: 110387435-110387435 on Build GRCh38
86710 c.10037G>A p.S3346N Chr.9: 110387308-110387308 on Build GRCh38
3952192 c.9927A>C p.K3309N Chr.9: 110387418-110387418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777699610 Chr.9: 110387387-110387387 on Build GRCh38
rs142463374 Chr.9: 110387328-110387328 on Build GRCh38
rs934784122 Chr.9: 110387596-110387596 on Build GRCh38
rs539407540 Chr.9: 110387698-110387698 on Build GRCh38
rs746170979 Chr.9: 110387334-110387334 on Build GRCh38
rs373456114 Chr.9: 110387506-110387506 on Build GRCh38
rs921472934 Chr.9: 110387227-110387227 on Build GRCh38
rs570620715 Chr.9: 110387418-110387418 on Build GRCh38
rs367929442 Chr.9: 110387386-110387386 on Build GRCh38
rs767077311 Chr.9: 110387256-110387256 on Build GRCh38
rs746181779 Chr.9: 110387394-110387394 on Build GRCh38
rs775715823 Chr.9: 110387497-110387497 on Build GRCh38
rs780851750 Chr.9: 110387693-110387695 on Build GRCh38
rs1032671949 Chr.9: 110387676-110387676 on Build GRCh38
rs777716373 Chr.9: 110387269-110387269 on Build GRCh38
rs371503132 Chr.9: 110387393-110387393 on Build GRCh38
rs769876413 Chr.9: 110387459-110387460 on Build GRCh38
rs750188941 Chr.9: 110387382-110387382 on Build GRCh38
rs987069562 Chr.9: 110387516-110387516 on Build GRCh38
rs1053379165 Chr.9: 110387621-110387621 on Build GRCh38
rs978933232 Chr.9: 110387550-110387550 on Build GRCh38
rs752136641 Chr.9: 110387265-110387265 on Build GRCh38
rs943557513 Chr.9: 110387275-110387275 on Build GRCh38
rs137911727 Chr.9: 110387215-110387215 on Build GRCh38
rs775162445 Chr.9: 110387397-110387397 on Build GRCh38
rs781522248 Chr.9: 110387404-110387405 on Build GRCh38
rs933105750 Chr.9: 110387250-110387250 on Build GRCh38
rs905977778 Chr.9: 110387655-110387655 on Build GRCh38
rs771382502 Chr.9: 110387333-110387333 on Build GRCh38
rs898540188 Chr.9: 110387580-110387580 on Build GRCh38
rs768933750 Chr.9: 110387296-110387296 on Build GRCh38
rs753665110 Chr.9: 110387347-110387347 on Build GRCh38
rs553304502 Chr.9: 110387421-110387422 on Build GRCh38
rs1016093411 Chr.9: 110387411-110387411 on Build GRCh38
rs201866666 Chr.9: 110387398-110387398 on Build GRCh38
rs938087573 Chr.9: 110387613-110387613 on Build GRCh38
rs766258144 Chr.9: 110387452-110387452 on Build GRCh38
rs756018074 Chr.9: 110387268-110387268 on Build GRCh38
rs368624561 Chr.9: 110387239-110387239 on Build GRCh38
rs559268210 Chr.9: 110387240-110387240 on Build GRCh38
rs1002063921 Chr.9: 110387410-110387410 on Build GRCh38
rs926139061 Chr.9: 110387590-110387590 on Build GRCh38
rs781527818 Chr.9: 110387385-110387385 on Build GRCh38
rs1044400764 Chr.9: 110387645-110387645 on Build GRCh38
rs772484487 Chr.9: 110387491-110387491 on Build GRCh38
rs114581956 Chr.9: 110387332-110387332 on Build GRCh38
rs577484502 Chr.9: 110387273-110387273 on Build GRCh38
rs201533942 Chr.9: 110387346-110387346 on Build GRCh38
rs756283926 Chr.9: 110387471-110387471 on Build GRCh38
rs2255007 Chr.9: 110387672-110387672 on Build GRCh38
rs903086686 Chr.9: 110387660-110387660 on Build GRCh38
rs774690333 Chr.9: 110387335-110387335 on Build GRCh38
rs1016551469 Chr.9: 110387679-110387679 on Build GRCh38
rs200311711 Chr.9: 110387281-110387281 on Build GRCh38
rs779350335 Chr.9: 110387482-110387482 on Build GRCh38
rs759190053 Chr.9: 110387340-110387340 on Build GRCh38
rs552175675 Chr.9: 110387413-110387413 on Build GRCh38
rs963660922 Chr.9: 110387416-110387416 on Build GRCh38
rs749868711 Chr.9: 110387389-110387389 on Build GRCh38
rs943109153 Chr.9: 110387542-110387542 on Build GRCh38
rs899000888 Chr.9: 110387686-110387686 on Build GRCh38
rs777909503 Chr.9: 110387476-110387476 on Build GRCh38
rs763898532 Chr.9: 110387285-110387285 on Build GRCh38
rs868116757 Chr.9: 110387374-110387374 on Build GRCh38
rs368850323 Chr.9: 110387425-110387425 on Build GRCh38
rs1051462695 Chr.9: 110387263-110387263 on Build GRCh38
rs780367145 Chr.9: 110387627-110387627 on Build GRCh38
rs770237687 Chr.9: 110387325-110387325 on Build GRCh38
rs753017562 Chr.9: 110387453-110387453 on Build GRCh38
rs757782477 Chr.9: 110387481-110387481 on Build GRCh38
rs756410094 Chr.9: 110387466-110387466 on Build GRCh38
rs910325692 Chr.9: 110387539-110387539 on Build GRCh38
rs368437025 Chr.9: 110387528-110387528 on Build GRCh38
rs771271924 Chr.9: 110387391-110387391 on Build GRCh38
rs749264743 Chr.9: 110387479-110387479 on Build GRCh38
rs553858774 Chr.9: 110387624-110387624 on Build GRCh38
rs189994659 Chr.9: 110387513-110387513 on Build GRCh38
rs759171842 Chr.9: 110387254-110387254 on Build GRCh38
rs868612511 Chr.9: 110387423-110387423 on Build GRCh38
rs909150481 Chr.9: 110387214-110387214 on Build GRCh38
rs372409135 Chr.9: 110387357-110387357 on Build GRCh38
rs980573618 Chr.9: 110387212-110387212 on Build GRCh38
rs775936857 Chr.9: 110387402-110387402 on Build GRCh38
rs138268746 Chr.9: 110387507-110387507 on Build GRCh38
rs957034253 Chr.9: 110387678-110387678 on Build GRCh38
rs993704412 Chr.9: 110387691-110387691 on Build GRCh38
rs757138069 Chr.9: 110387376-110387376 on Build GRCh38
rs781439987 Chr.9: 110387317-110387317 on Build GRCh38
rs765128964 Chr.9: 110387379-110387379 on Build GRCh38
rs746341302 Chr.9: 110387490-110387490 on Build GRCh38
rs746678073 Chr.9: 110387498-110387498 on Build GRCh38
rs778561103 Chr.9: 110387284-110387284 on Build GRCh38
rs544465911 Chr.9: 110387324-110387324 on Build GRCh38
rs753872713 Chr.9: 110387358-110387358 on Build GRCh38
rs144712744 Chr.9: 110387443-110387443 on Build GRCh38
rs768136355 Chr.9: 110387501-110387501 on Build GRCh38
rs767097945 Chr.9: 110387343-110387343 on Build GRCh38
rs79932775 Chr.9: 110387228-110387228 on Build GRCh38
rs375396556 Chr.9: 110387384-110387384 on Build GRCh38

View Full Product Details