Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110942076-110942583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAR1
Entrez Gene ID
1902
Gene Name
lysophosphatidic acid receptor 1
Gene Aliases
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
UniGene
Hs.126667
Gene Chromosome Location
Chr.9: 110873252-111039259 on Build GRCh38
Transcripts
NM_001401.3 NM_057159.2 XM_005251781.3 XM_005251782.3 XM_017014383.1 XM_017014384.1 XM_017014385.1 XM_017014386.1 XM_017014387.1 XM_017014388.1 XM_017014389.1 XM_017014390.1 XM_017014391.1 XM_017014392.1 XM_017014393.1 XM_017014394.1 XM_017014395.1 XM_017014396.1 XM_017014397.1 XM_017014398.1 XM_017014399.1 XM_017014400.1 XM_017014401.1 XM_017014402.1 XM_017014403.1 XM_017014404.1 XM_017014405.1 XM_017014406.1 XM_017014407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1472181 c.106C>T p.R36* Chr.9: 110942108-110942108 on Build GRCh38
3169751 c.90T>C p.I30I Chr.9: 110942124-110942124 on Build GRCh38
7375264 c.137A>G p.N46S Chr.9: 110942077-110942077 on Build GRCh38
1104018 c.107G>A p.R36Q Chr.9: 110942107-110942107 on Build GRCh38
3903044 c.46-2A>G p.? Chr.9: 110942170-110942170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549498612 Chr.9: 110942292-110942292 on Build GRCh38
rs34483952 Chr.9: 110942133-110942133 on Build GRCh38
rs186001742 Chr.9: 110942544-110942544 on Build GRCh38
rs935949106 Chr.9: 110942132-110942132 on Build GRCh38
rs751594313 Chr.9: 110942176-110942176 on Build GRCh38
rs749766946 Chr.9: 110942431-110942431 on Build GRCh38
rs545376901 Chr.9: 110942343-110942343 on Build GRCh38
rs184961053 Chr.9: 110942410-110942410 on Build GRCh38
rs75907283 Chr.9: 110942165-110942165 on Build GRCh38
rs757912248 Chr.9: 110942155-110942155 on Build GRCh38
rs771217048 Chr.9: 110942229-110942229 on Build GRCh38
rs1014796808 Chr.9: 110942316-110942316 on Build GRCh38
rs1041908082 Chr.9: 110942126-110942126 on Build GRCh38
rs921837669 Chr.9: 110942122-110942122 on Build GRCh38
rs562722471 Chr.9: 110942471-110942471 on Build GRCh38
rs781232372 Chr.9: 110942194-110942194 on Build GRCh38
rs527714967 Chr.9: 110942492-110942492 on Build GRCh38
rs754985796 Chr.9: 110942189-110942189 on Build GRCh38
rs560214328 Chr.9: 110942285-110942285 on Build GRCh38
rs755876769 Chr.9: 110942200-110942200 on Build GRCh38
rs916911983 Chr.9: 110942491-110942491 on Build GRCh38
rs13286391 Chr.9: 110942456-110942456 on Build GRCh38
rs754224955 Chr.9: 110942108-110942108 on Build GRCh38
rs13287550 Chr.9: 110942415-110942415 on Build GRCh38
rs906627369 Chr.9: 110942251-110942251 on Build GRCh38
rs1039795542 Chr.9: 110942549-110942549 on Build GRCh38
rs779066122 Chr.9: 110942203-110942203 on Build GRCh38
rs774707115 Chr.9: 110942503-110942503 on Build GRCh38
rs990052108 Chr.9: 110942519-110942519 on Build GRCh38
rs539927641 Chr.9: 110942244-110942244 on Build GRCh38
rs779679530 Chr.9: 110942171-110942171 on Build GRCh38
rs1004570715 Chr.9: 110942344-110942344 on Build GRCh38
rs926141387 Chr.9: 110942530-110942530 on Build GRCh38
rs755872645 Chr.9: 110942171-110942171 on Build GRCh38
rs531631706 Chr.9: 110942475-110942475 on Build GRCh38
rs548503236 Chr.9: 110942476-110942476 on Build GRCh38
rs147950032 Chr.9: 110942204-110942204 on Build GRCh38
rs950334723 Chr.9: 110942307-110942307 on Build GRCh38
rs1055737051 Chr.9: 110942276-110942276 on Build GRCh38
rs1034485809 Chr.9: 110942374-110942374 on Build GRCh38
rs764494647 Chr.9: 110942106-110942106 on Build GRCh38
rs1003682995 Chr.9: 110942260-110942260 on Build GRCh38
rs549966264 Chr.9: 110942537-110942537 on Build GRCh38
rs181074250 Chr.9: 110942512-110942512 on Build GRCh38
rs199936061 Chr.9: 110942197-110942197 on Build GRCh38
rs936123191 Chr.9: 110942187-110942187 on Build GRCh38
rs141838965 Chr.9: 110942238-110942238 on Build GRCh38
rs570630950 Chr.9: 110942507-110942507 on Build GRCh38
rs762199012 Chr.9: 110942128-110942128 on Build GRCh38
rs371567118 Chr.9: 110942149-110942149 on Build GRCh38
rs181457269 Chr.9: 110942297-110942297 on Build GRCh38
rs1027310837 Chr.9: 110942306-110942306 on Build GRCh38
rs561921536 Chr.9: 110942488-110942488 on Build GRCh38
rs761142271 Chr.9: 110942103-110942103 on Build GRCh38
rs554265524 Chr.9: 110942231-110942231 on Build GRCh38
rs769052215 Chr.9: 110942443-110942443 on Build GRCh38
rs966193913 Chr.9: 110942396-110942396 on Build GRCh38
rs189437232 Chr.9: 110942504-110942504 on Build GRCh38
rs1018666981 Chr.9: 110942354-110942354 on Build GRCh38

View Full Product Details