Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111360891-111361403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0368
Entrez Gene ID
23392
Gene Name
KIAA0368
Gene Aliases
ECM29
UniGene
Hs.368255
Gene Chromosome Location
Chr.9: 111360693-111484745 on Build GRCh38
Transcripts
NM_001080398.1 XM_005251850.3

SNP Detail

SNP ID Chromosome Location
rs925549624 Chr.9: 111361041-111361041 on Build GRCh38
rs758449563 Chr.9: 111360921-111360921 on Build GRCh38
rs913225755 Chr.9: 111361000-111361000 on Build GRCh38
rs1041167077 Chr.9: 111361339-111361339 on Build GRCh38
rs531303043 Chr.9: 111361155-111361155 on Build GRCh38
rs958081087 Chr.9: 111361336-111361336 on Build GRCh38
rs969255259 Chr.9: 111361199-111361199 on Build GRCh38
rs564329882 Chr.9: 111360942-111360942 on Build GRCh38
rs866769316 Chr.9: 111361263-111361263 on Build GRCh38
rs547654694 Chr.9: 111361290-111361290 on Build GRCh38
rs749732172 Chr.9: 111361149-111361149 on Build GRCh38
rs986855527 Chr.9: 111360962-111360962 on Build GRCh38
rs1025293009 Chr.9: 111361176-111361176 on Build GRCh38
rs191368250 Chr.9: 111361231-111361231 on Build GRCh38
rs936883857 Chr.9: 111361072-111361072 on Build GRCh38
rs964158904 Chr.9: 111361296-111361296 on Build GRCh38
rs541656447 Chr.9: 111360932-111360932 on Build GRCh38
rs189991935 Chr.9: 111361276-111361276 on Build GRCh38
rs949567022 Chr.9: 111361119-111361119 on Build GRCh38
rs1037741811 Chr.9: 111361208-111361208 on Build GRCh38
rs993805985 Chr.9: 111361163-111361163 on Build GRCh38
rs926395875 Chr.9: 111361175-111361175 on Build GRCh38
rs1043533195 Chr.9: 111361042-111361042 on Build GRCh38
rs1032717002 Chr.9: 111361324-111361324 on Build GRCh38
rs561353948 Chr.9: 111361340-111361340 on Build GRCh38
rs764099019 Chr.9: 111361293-111361293 on Build GRCh38
rs988785192 Chr.9: 111360996-111360996 on Build GRCh38
rs930252020 Chr.9: 111361361-111361361 on Build GRCh38
rs547990112 Chr.9: 111361230-111361230 on Build GRCh38
rs942829066 Chr.9: 111361108-111361108 on Build GRCh38
rs911258995 Chr.9: 111361311-111361312 on Build GRCh38
rs889995812 Chr.9: 111361089-111361089 on Build GRCh38
rs964022366 Chr.9: 111361030-111361030 on Build GRCh38
rs890281726 Chr.9: 111361269-111361269 on Build GRCh38
rs914047233 Chr.9: 111361101-111361101 on Build GRCh38
rs978877358 Chr.9: 111361311-111361311 on Build GRCh38
rs969622811 Chr.9: 111360921-111360921 on Build GRCh38
rs914182142 Chr.9: 111361376-111361376 on Build GRCh38
rs533477962 Chr.9: 111360957-111360957 on Build GRCh38
rs200637126 Chr.9: 111361205-111361205 on Build GRCh38
rs187550351 Chr.9: 111360977-111360977 on Build GRCh38
rs982083581 Chr.9: 111360973-111360973 on Build GRCh38
rs993800435 Chr.9: 111361213-111361213 on Build GRCh38
rs947520283 Chr.9: 111361016-111361016 on Build GRCh38
rs562168239 Chr.9: 111360990-111360990 on Build GRCh38
rs937687615 Chr.9: 111361186-111361186 on Build GRCh38
rs988412413 Chr.9: 111361074-111361074 on Build GRCh38
rs183516071 Chr.9: 111361235-111361235 on Build GRCh38
rs751331457 Chr.9: 111361295-111361295 on Build GRCh38
rs1023795072 Chr.9: 111361187-111361187 on Build GRCh38
rs1047298434 Chr.9: 111361373-111361373 on Build GRCh38
rs975858619 Chr.9: 111361034-111361034 on Build GRCh38
rs780139911 Chr.9: 111361341-111361341 on Build GRCh38
rs996949202 Chr.9: 111361315-111361315 on Build GRCh38
rs947367193 Chr.9: 111361302-111361305 on Build GRCh38
rs556958300 Chr.9: 111361210-111361210 on Build GRCh38
rs757034650 Chr.9: 111361327-111361327 on Build GRCh38

View Full Product Details