Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 68483767-68484098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGM5
Entrez Gene ID
5239
Gene Name
phosphoglucomutase 5
Gene Aliases
PGMRP
UniGene
Hs.307835
Gene Chromosome Location
Chr.9: 68356438-68531061 on Build GRCh38
Transcripts
NM_021965.3 XM_017014808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6183554 c.1444G>T p.V482L Chr.9: 68484013-68484013 on Build GRCh38
1625047 c.1421C>A p.A474E Chr.9: 68483990-68483990 on Build GRCh38
7119267 c.1366G>C p.D456H Chr.9: 68483935-68483935 on Build GRCh38
1462724 c.1427C>T p.T476M Chr.9: 68483996-68483996 on Build GRCh38
1462723 c.1421C>T p.A474V Chr.9: 68483990-68483990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781811890 Chr.9: 68484076-68484076 on Build GRCh38
rs782335493 Chr.9: 68484013-68484013 on Build GRCh38
rs141481382 Chr.9: 68484021-68484021 on Build GRCh38
rs547897747 Chr.9: 68484012-68484012 on Build GRCh38
rs531516830 Chr.9: 68483821-68483821 on Build GRCh38
rs782088543 Chr.9: 68483843-68483843 on Build GRCh38
rs377702519 Chr.9: 68484070-68484072 on Build GRCh38
rs373525562 Chr.9: 68483991-68483991 on Build GRCh38
rs782514355 Chr.9: 68483989-68483989 on Build GRCh38
rs781843179 Chr.9: 68483990-68483990 on Build GRCh38
rs199548660 Chr.9: 68483965-68483965 on Build GRCh38
rs782667641 Chr.9: 68484070-68484070 on Build GRCh38
rs782703326 Chr.9: 68484074-68484074 on Build GRCh38
rs372446112 Chr.9: 68483841-68483841 on Build GRCh38
rs782741383 Chr.9: 68483979-68483979 on Build GRCh38
rs1024875794 Chr.9: 68483938-68483938 on Build GRCh38
rs565179747 Chr.9: 68483852-68483852 on Build GRCh38
rs7037864 Chr.9: 68483915-68483915 on Build GRCh38
rs782323267 Chr.9: 68484052-68484052 on Build GRCh38
rs888846546 Chr.9: 68483815-68483815 on Build GRCh38
rs377710211 Chr.9: 68484006-68484006 on Build GRCh38
rs10868913 Chr.9: 68484071-68484071 on Build GRCh38
rs201467646 Chr.9: 68483884-68483884 on Build GRCh38
rs782384644 Chr.9: 68483967-68483967 on Build GRCh38
rs782680556 Chr.9: 68483937-68483937 on Build GRCh38
rs782099613 Chr.9: 68484073-68484073 on Build GRCh38
rs201672908 Chr.9: 68483894-68483894 on Build GRCh38
rs139989462 Chr.9: 68483997-68483997 on Build GRCh38
rs782414200 Chr.9: 68483823-68483823 on Build GRCh38
rs782298580 Chr.9: 68483962-68483962 on Build GRCh38
rs781925256 Chr.9: 68483861-68483861 on Build GRCh38
rs150283095 Chr.9: 68483984-68483984 on Build GRCh38
rs782070590 Chr.9: 68483879-68483879 on Build GRCh38
rs549835772 Chr.9: 68483827-68483827 on Build GRCh38
rs782546219 Chr.9: 68483929-68483929 on Build GRCh38
rs782632445 Chr.9: 68484007-68484007 on Build GRCh38
rs200484784 Chr.9: 68483956-68483956 on Build GRCh38
rs782345591 Chr.9: 68483945-68483945 on Build GRCh38
rs782063471 Chr.9: 68483985-68483985 on Build GRCh38
rs368463883 Chr.9: 68483817-68483817 on Build GRCh38
rs200812025 Chr.9: 68483996-68483996 on Build GRCh38
rs782279448 Chr.9: 68483820-68483820 on Build GRCh38
rs782023048 Chr.9: 68483960-68483960 on Build GRCh38
rs781820403 Chr.9: 68483895-68483895 on Build GRCh38
rs782014216 Chr.9: 68484057-68484057 on Build GRCh38
rs141086525 Chr.9: 68483953-68483953 on Build GRCh38
rs782474795 Chr.9: 68483899-68483899 on Build GRCh38
rs781901814 Chr.9: 68483986-68483986 on Build GRCh38
rs781993879 Chr.9: 68483970-68483970 on Build GRCh38
rs200775373 Chr.9: 68483948-68483948 on Build GRCh38
rs376761551 Chr.9: 68483922-68483922 on Build GRCh38
rs782266902 Chr.9: 68483944-68483944 on Build GRCh38
rs369200475 Chr.9: 68483936-68483936 on Build GRCh38
rs782077177 Chr.9: 68483975-68483975 on Build GRCh38
rs781802650 Chr.9: 68483924-68483924 on Build GRCh38
rs373143413 Chr.9: 68484061-68484061 on Build GRCh38
rs10868912 Chr.9: 68484070-68484070 on Build GRCh38
rs1006421617 Chr.9: 68483865-68483865 on Build GRCh38

View Full Product Details