Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72230748-72231269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
UniGene
Hs.494163
Gene Chromosome Location
Chr.9: 72114595-72256045 on Build GRCh38
Transcripts
NM_001242505.2 NM_001242506.2 NM_001242507.2 NM_004293.4 XM_005252317.1 XM_011519213.1 XM_011519214.1 XM_011519215.1 XM_011519216.1 XM_011519217.1 XM_011519218.2 XM_011519219.1 XM_011519220.1 XM_011519221.1 XM_011519222.1 XM_011519223.2 XM_017015333.1 XM_017015334.1 XM_017015335.1 XM_017015336.1 XM_017015337.1 XM_017015338.1 XM_017015339.1 XM_017015340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3982916 c.988+7A>G p.? Chr.9: 72231188-72231188 on Build GRCh38
3982917 c.988+7A>G p.? Chr.9: 72231188-72231188 on Build GRCh38
6354647 c.980T>A p.L327Q Chr.9: 72231173-72231173 on Build GRCh38
6354646 c.980T>A p.L327Q Chr.9: 72231173-72231173 on Build GRCh38
4686687 c.531A>G p.G177G Chr.9: 72231126-72231126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556517763 Chr.9: 72230857-72230857 on Build GRCh38
rs769439352 Chr.9: 72231196-72231196 on Build GRCh38
rs775978984 Chr.9: 72231172-72231172 on Build GRCh38
rs759577515 Chr.9: 72231232-72231232 on Build GRCh38
rs561764912 Chr.9: 72230985-72230985 on Build GRCh38
rs1038118183 Chr.9: 72230856-72230856 on Build GRCh38
rs954927098 Chr.9: 72231068-72231068 on Build GRCh38
rs77290181 Chr.9: 72231227-72231227 on Build GRCh38
rs751241001 Chr.9: 72231045-72231045 on Build GRCh38
rs370557253 Chr.9: 72231164-72231164 on Build GRCh38
rs140452216 Chr.9: 72231127-72231127 on Build GRCh38
rs762471573 Chr.9: 72231217-72231217 on Build GRCh38
rs138155126 Chr.9: 72231003-72231003 on Build GRCh38
rs10869150 Chr.9: 72231073-72231073 on Build GRCh38
rs1047940406 Chr.9: 72231147-72231147 on Build GRCh38
rs545684750 Chr.9: 72230932-72230932 on Build GRCh38
rs1047933847 Chr.9: 72230931-72230931 on Build GRCh38
rs181367478 Chr.9: 72231119-72231119 on Build GRCh38
rs765913650 Chr.9: 72231220-72231220 on Build GRCh38
rs756428524 Chr.9: 72231130-72231130 on Build GRCh38
rs749999963 Chr.9: 72231066-72231066 on Build GRCh38
rs772089034 Chr.9: 72231167-72231167 on Build GRCh38
rs1033735051 Chr.9: 72230976-72230976 on Build GRCh38
rs1009124731 Chr.9: 72230965-72230965 on Build GRCh38
rs890731061 Chr.9: 72230958-72230958 on Build GRCh38
rs75167293 Chr.9: 72231019-72231019 on Build GRCh38
rs201812111 Chr.9: 72231088-72231088 on Build GRCh38
rs543188331 Chr.9: 72230788-72230788 on Build GRCh38
rs971728104 Chr.9: 72231040-72231040 on Build GRCh38
rs375873368 Chr.9: 72231081-72231081 on Build GRCh38
rs559234646 Chr.9: 72230936-72230936 on Build GRCh38
rs973846119 Chr.9: 72231076-72231076 on Build GRCh38
rs977853369 Chr.9: 72231053-72231053 on Build GRCh38
rs1048032451 Chr.9: 72230942-72230942 on Build GRCh38
rs749236424 Chr.9: 72231108-72231108 on Build GRCh38
rs900460740 Chr.9: 72230982-72230982 on Build GRCh38
rs771090625 Chr.9: 72231162-72231162 on Build GRCh38
rs899328041 Chr.9: 72230879-72230879 on Build GRCh38
rs542021356 Chr.9: 72230949-72230949 on Build GRCh38
rs772758487 Chr.9: 72231101-72231101 on Build GRCh38
rs750940716 Chr.9: 72230848-72230851 on Build GRCh38
rs17057536 Chr.9: 72231188-72231188 on Build GRCh38
rs1000189168 Chr.9: 72231206-72231206 on Build GRCh38
rs1018032880 Chr.9: 72230979-72230979 on Build GRCh38
rs576479628 Chr.9: 72230922-72230922 on Build GRCh38
rs145667878 Chr.9: 72231151-72231151 on Build GRCh38
rs754620309 Chr.9: 72230987-72230987 on Build GRCh38
rs928168201 Chr.9: 72231241-72231241 on Build GRCh38
rs929690406 Chr.9: 72231096-72231096 on Build GRCh38
rs1024063252 Chr.9: 72231038-72231038 on Build GRCh38
rs778077706 Chr.9: 72231145-72231145 on Build GRCh38
rs751012127 Chr.9: 72231226-72231226 on Build GRCh38
rs796997197 Chr.9: 72230804-72230804 on Build GRCh38
rs562016146 Chr.9: 72230888-72230888 on Build GRCh38
rs910524534 Chr.9: 72231080-72231080 on Build GRCh38
rs772432458 Chr.9: 72231171-72231171 on Build GRCh38
rs188772449 Chr.9: 72230940-72230940 on Build GRCh38
rs920812685 Chr.9: 72231148-72231148 on Build GRCh38
rs112925720 Chr.9: 72231028-72231028 on Build GRCh38
rs772793036 Chr.9: 72231209-72231209 on Build GRCh38
rs188446998 Chr.9: 72231221-72231221 on Build GRCh38
rs754840971 Chr.9: 72231118-72231118 on Build GRCh38
rs890597553 Chr.9: 72231149-72231149 on Build GRCh38
rs34354742 Chr.9: 72231033-72231033 on Build GRCh38

View Full Product Details