Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76613251-76613783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
UniGene
Hs.262857
Gene Chromosome Location
Chr.9: 76611376-76906220 on Build GRCh38
Transcripts
NM_001308047.1 NM_001308048.1 NM_001308049.1 NM_001308050.1 NM_001308051.1 NM_015225.2 NR_131751.1 XM_005251745.1 XM_005251746.1 XM_005251748.1 XM_005251750.1 XM_005251751.1 XM_005251754.1 XM_006716982.1 XM_006716983.1 XM_006716985.1 XM_006716986.1 XM_011518323.1 XM_011518327.1 XM_017014345.1 XM_017014346.1 XM_017014347.1 XM_017014348.1 XM_017014349.1 XM_017014350.1 XM_017014351.1 XM_017014352.1 XM_017014354.1 XM_017014355.1 XM_017014356.1 XM_017014357.1 XM_017014358.1 XM_017014359.1

SNP Detail

SNP ID Chromosome Location
rs769945523 Chr.9: 76613374-76613374 on Build GRCh38
rs1054809991 Chr.9: 76613656-76613656 on Build GRCh38
rs922903344 Chr.9: 76613355-76613355 on Build GRCh38
rs956914442 Chr.9: 76613724-76613724 on Build GRCh38
rs76157996 Chr.9: 76613379-76613379 on Build GRCh38
rs186656717 Chr.9: 76613318-76613318 on Build GRCh38
rs905510086 Chr.9: 76613523-76613523 on Build GRCh38
rs894986092 Chr.9: 76613419-76613419 on Build GRCh38
rs1024861586 Chr.9: 76613435-76613435 on Build GRCh38
rs908912404 Chr.9: 76613634-76613634 on Build GRCh38
rs751464971 Chr.9: 76613486-76613486 on Build GRCh38
rs867337057 Chr.9: 76613424-76613424 on Build GRCh38
rs555904599 Chr.9: 76613759-76613759 on Build GRCh38
rs985612613 Chr.9: 76613279-76613279 on Build GRCh38
rs1047876138 Chr.9: 76613360-76613360 on Build GRCh38
rs935492184 Chr.9: 76613455-76613455 on Build GRCh38
rs572569462 Chr.9: 76613426-76613429 on Build GRCh38
rs1044466311 Chr.9: 76613511-76613511 on Build GRCh38
rs1041329636 Chr.9: 76613335-76613335 on Build GRCh38
rs905782160 Chr.9: 76613277-76613277 on Build GRCh38
rs766692867 Chr.9: 76613475-76613475 on Build GRCh38
rs574241292 Chr.9: 76613450-76613450 on Build GRCh38
rs113526558 Chr.9: 76613557-76613557 on Build GRCh38
rs894953298 Chr.9: 76613666-76613666 on Build GRCh38
rs958139708 Chr.9: 76613484-76613484 on Build GRCh38
rs534435803 Chr.9: 76613381-76613381 on Build GRCh38
rs549713585 Chr.9: 76613333-76613333 on Build GRCh38
rs3087612 Chr.9: 76613436-76613436 on Build GRCh38
rs182367330 Chr.9: 76613304-76613304 on Build GRCh38
rs3174373 Chr.9: 76613688-76613688 on Build GRCh38
rs79973033 Chr.9: 76613364-76613364 on Build GRCh38
rs200687761 Chr.9: 76613485-76613485 on Build GRCh38
rs118012415 Chr.9: 76613535-76613535 on Build GRCh38
rs1013422141 Chr.9: 76613425-76613425 on Build GRCh38
rs755966589 Chr.9: 76613641-76613641 on Build GRCh38
rs911352902 Chr.9: 76613515-76613515 on Build GRCh38
rs145005783 Chr.9: 76613615-76613615 on Build GRCh38
rs115888754 Chr.9: 76613687-76613687 on Build GRCh38
rs147841749 Chr.9: 76613309-76613309 on Build GRCh38
rs965551335 Chr.9: 76613534-76613534 on Build GRCh38
rs999514217 Chr.9: 76613446-76613446 on Build GRCh38
rs531896686 Chr.9: 76613358-76613358 on Build GRCh38
rs149918939 Chr.9: 76613558-76613558 on Build GRCh38
rs947102249 Chr.9: 76613506-76613506 on Build GRCh38
rs141462498 Chr.9: 76613549-76613549 on Build GRCh38
rs3174372 Chr.9: 76613638-76613638 on Build GRCh38
rs924962402 Chr.9: 76613760-76613760 on Build GRCh38
rs750993261 Chr.9: 76613460-76613460 on Build GRCh38
rs888547217 Chr.9: 76613620-76613620 on Build GRCh38
rs1031249381 Chr.9: 76613706-76613706 on Build GRCh38
rs989614110 Chr.9: 76613757-76613757 on Build GRCh38

View Full Product Details