Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 79574771-79575287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLE4
Entrez Gene ID
7091
Gene Name
transducin like enhancer of split 4
Gene Aliases
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
UniGene
Hs.444213
Gene Chromosome Location
Chr.9: 79571773-79726882 on Build GRCh38
Transcripts
NM_001282748.1 NM_001282749.1 NM_001282753.1 NM_001282760.1 NM_007005.4 NR_104239.1 XM_005252177.1 XM_006717264.1 XM_006717268.1 XM_011518952.1 XM_011518953.1 XM_011518954.1 XM_011518955.1 XM_011518956.1 XM_011518957.1 XM_011518958.1 XM_011518959.1 XM_011518960.1 XM_011518961.1 XM_011518962.1 XM_011518963.1 XM_011518964.1 XM_011518965.1 XM_011518966.1 XM_011518967.1 XM_011518968.1 XM_011518969.1 XM_011518970.2 XM_017015067.1 XM_017015068.1 XM_017015069.1 XM_017015070.1 XM_017015071.1 XM_017015072.1 XM_017015073.1 XM_017015074.1 XM_017015075.1 XM_017015076.1 XM_017015077.1 XM_017015078.1 XM_017015079.1 XM_017015080.1 XM_017015081.1 XM_017015082.1 XM_017015083.1 XM_017015084.1 XM_017015085.1 XM_017015086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6196434 c.159delT p.C53fs*25 Chr.9: 79574888-79574888 on Build GRCh38
1756232 c.193C>T p.R65W Chr.9: 79574922-79574922 on Build GRCh38
4798628 c.195G>A p.R65R Chr.9: 79574924-79574924 on Build GRCh38
285741 c.193C>T p.R65W Chr.9: 79574922-79574922 on Build GRCh38
1110366 c.194G>A p.R65Q Chr.9: 79574923-79574923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535578698 Chr.9: 79575074-79575074 on Build GRCh38
rs1029414827 Chr.9: 79575016-79575016 on Build GRCh38
rs781666963 Chr.9: 79574957-79574957 on Build GRCh38
rs773801480 Chr.9: 79574853-79574853 on Build GRCh38
rs1057511882 Chr.9: 79575203-79575203 on Build GRCh38
rs906210334 Chr.9: 79575192-79575192 on Build GRCh38
rs753612094 Chr.9: 79574867-79574867 on Build GRCh38
rs1035923363 Chr.9: 79574810-79574810 on Build GRCh38
rs940564005 Chr.9: 79575082-79575082 on Build GRCh38
rs985126908 Chr.9: 79575055-79575055 on Build GRCh38
rs906101369 Chr.9: 79575193-79575193 on Build GRCh38
rs374088818 Chr.9: 79574961-79574961 on Build GRCh38
rs553821489 Chr.9: 79575101-79575101 on Build GRCh38
rs970321505 Chr.9: 79574830-79574830 on Build GRCh38
rs771694863 Chr.9: 79575162-79575162 on Build GRCh38
rs757913564 Chr.9: 79574909-79574909 on Build GRCh38
rs974784268 Chr.9: 79575014-79575014 on Build GRCh38
rs779775864 Chr.9: 79574912-79574912 on Build GRCh38
rs779794270 Chr.9: 79574832-79574832 on Build GRCh38
rs199667147 Chr.9: 79574860-79574860 on Build GRCh38
rs367712800 Chr.9: 79574970-79574970 on Build GRCh38
rs746648577 Chr.9: 79574948-79574948 on Build GRCh38
rs920390775 Chr.9: 79575156-79575156 on Build GRCh38
rs570337167 Chr.9: 79574864-79574864 on Build GRCh38
rs770245511 Chr.9: 79574847-79574847 on Build GRCh38
rs571995671 Chr.9: 79575160-79575160 on Build GRCh38
rs1045910026 Chr.9: 79575192-79575193 on Build GRCh38
rs774440168 Chr.9: 79575013-79575013 on Build GRCh38
rs79606881 Chr.9: 79575234-79575234 on Build GRCh38
rs1010447844 Chr.9: 79575216-79575216 on Build GRCh38
rs1021802893 Chr.9: 79575258-79575258 on Build GRCh38
rs368693340 Chr.9: 79575262-79575262 on Build GRCh38
rs770219864 Chr.9: 79574962-79574962 on Build GRCh38
rs892038921 Chr.9: 79575206-79575206 on Build GRCh38
rs771634166 Chr.9: 79574825-79574825 on Build GRCh38
rs774378920 Chr.9: 79574934-79574934 on Build GRCh38
rs954701928 Chr.9: 79575061-79575061 on Build GRCh38
rs191638143 Chr.9: 79574807-79574807 on Build GRCh38
rs989319452 Chr.9: 79574898-79574898 on Build GRCh38
rs112466188 Chr.9: 79574802-79574802 on Build GRCh38
rs907774703 Chr.9: 79575075-79575075 on Build GRCh38
rs762360597 Chr.9: 79574844-79574844 on Build GRCh38
rs539424469 Chr.9: 79575191-79575191 on Build GRCh38
rs537705134 Chr.9: 79574897-79574897 on Build GRCh38
rs770997282 Chr.9: 79574971-79574971 on Build GRCh38
rs766463681 Chr.9: 79574856-79574856 on Build GRCh38
rs773537753 Chr.9: 79574963-79574963 on Build GRCh38
rs530869053 Chr.9: 79575202-79575202 on Build GRCh38
rs1021751874 Chr.9: 79574992-79574992 on Build GRCh38
rs765035827 Chr.9: 79574885-79574885 on Build GRCh38
rs565978720 Chr.9: 79574841-79574841 on Build GRCh38
rs769136330 Chr.9: 79574838-79574838 on Build GRCh38
rs893138859 Chr.9: 79575246-79575246 on Build GRCh38
rs752161919 Chr.9: 79574853-79574854 on Build GRCh38
rs533531677 Chr.9: 79574903-79574903 on Build GRCh38
rs2791566 Chr.9: 79574999-79574999 on Build GRCh38
rs745326552 Chr.9: 79574822-79574822 on Build GRCh38
rs757102907 Chr.9: 79574880-79574880 on Build GRCh38
rs754729059 Chr.9: 79574953-79574953 on Build GRCh38
rs909559989 Chr.9: 79575069-79575069 on Build GRCh38
rs146585022 Chr.9: 79574857-79574857 on Build GRCh38

View Full Product Details