Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85586587-85587119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTPBP1
Entrez Gene ID
23287
Gene Name
ATP/GTP binding protein 1
Gene Aliases
CCP1, NNA1
UniGene
Hs.719980
Gene Chromosome Location
Chr.9: 85543799-85742029 on Build GRCh38
Transcripts
NM_001286715.1 NM_001286717.1 NM_015239.2 XM_005251847.3 XM_005251848.2 XM_005251849.3 XM_011518418.2 XM_011518420.2 XM_017014544.1 XM_017014545.1 XM_017014546.1 XM_017014547.1 XM_017014548.1 XM_017014549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996665 c.2883A>T p.Q961H Chr.9: 85586861-85586861 on Build GRCh38
6485876 c.2804G>A p.G935E Chr.9: 85586940-85586940 on Build GRCh38
6485878 c.2924G>A p.G975E Chr.9: 85586940-85586940 on Build GRCh38
110248 c.2862C>T p.Y954Y Chr.9: 85586882-85586882 on Build GRCh38
3220663 c.2793T>C p.C931C Chr.9: 85586951-85586951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935215986 Chr.9: 85586662-85586662 on Build GRCh38
rs146610043 Chr.9: 85586891-85586891 on Build GRCh38
rs1043859169 Chr.9: 85586623-85586623 on Build GRCh38
rs183814474 Chr.9: 85586912-85586912 on Build GRCh38
rs771595414 Chr.9: 85586989-85586989 on Build GRCh38
rs765716154 Chr.9: 85587009-85587009 on Build GRCh38
rs576838938 Chr.9: 85586850-85586850 on Build GRCh38
rs777123357 Chr.9: 85586863-85586863 on Build GRCh38
rs776927146 Chr.9: 85587004-85587004 on Build GRCh38
rs756553805 Chr.9: 85586983-85586992 on Build GRCh38
rs750427009 Chr.9: 85586881-85586881 on Build GRCh38
rs562920839 Chr.9: 85586646-85586658 on Build GRCh38
rs754032234 Chr.9: 85586663-85586663 on Build GRCh38
rs1038858305 Chr.9: 85586822-85586822 on Build GRCh38
rs531985222 Chr.9: 85586841-85586841 on Build GRCh38
rs745416854 Chr.9: 85586986-85586986 on Build GRCh38
rs202122009 Chr.9: 85587008-85587008 on Build GRCh38
rs755106121 Chr.9: 85586682-85586682 on Build GRCh38
rs540324312 Chr.9: 85587060-85587060 on Build GRCh38
rs1056784546 Chr.9: 85586676-85586676 on Build GRCh38
rs940153940 Chr.9: 85586802-85586802 on Build GRCh38
rs767066823 Chr.9: 85586640-85586640 on Build GRCh38
rs112536234 Chr.9: 85586785-85586785 on Build GRCh38
rs746596648 Chr.9: 85586998-85586998 on Build GRCh38
rs775183679 Chr.9: 85586842-85586842 on Build GRCh38
rs566398817 Chr.9: 85587071-85587071 on Build GRCh38
rs777927517 Chr.9: 85586784-85586784 on Build GRCh38
rs77347683 Chr.9: 85586948-85586948 on Build GRCh38
rs762246308 Chr.9: 85586870-85586870 on Build GRCh38
rs767434021 Chr.9: 85586635-85586635 on Build GRCh38
rs143027240 Chr.9: 85586882-85586882 on Build GRCh38
rs891959028 Chr.9: 85587016-85587016 on Build GRCh38
rs143236977 Chr.9: 85586772-85586772 on Build GRCh38
rs764437523 Chr.9: 85586862-85586862 on Build GRCh38
rs1022002313 Chr.9: 85586700-85586700 on Build GRCh38
rs755608937 Chr.9: 85586965-85586965 on Build GRCh38
rs188577703 Chr.9: 85587053-85587053 on Build GRCh38
rs769018280 Chr.9: 85587000-85587000 on Build GRCh38
rs765378045 Chr.9: 85586874-85586874 on Build GRCh38
rs751831272 Chr.9: 85586903-85586903 on Build GRCh38
rs767483870 Chr.9: 85586809-85586809 on Build GRCh38
rs564056609 Chr.9: 85586657-85586657 on Build GRCh38
rs553838748 Chr.9: 85586752-85586752 on Build GRCh38
rs147500747 Chr.9: 85586831-85586831 on Build GRCh38
rs988290411 Chr.9: 85586691-85586694 on Build GRCh38
rs968101471 Chr.9: 85586708-85586708 on Build GRCh38
rs775189316 Chr.9: 85586996-85586996 on Build GRCh38
rs1009344802 Chr.9: 85587051-85587051 on Build GRCh38
rs770579988 Chr.9: 85586786-85586786 on Build GRCh38
rs148730956 Chr.9: 85586629-85586629 on Build GRCh38
rs902333605 Chr.9: 85586653-85586653 on Build GRCh38
rs750586809 Chr.9: 85586981-85586984 on Build GRCh38
rs755981511 Chr.9: 85586904-85586904 on Build GRCh38
rs778880433 Chr.9: 85586960-85586960 on Build GRCh38
rs758485580 Chr.9: 85586884-85586884 on Build GRCh38
rs375881738 Chr.9: 85586958-85586958 on Build GRCh38
rs947953412 Chr.9: 85587085-85587085 on Build GRCh38
rs773758793 Chr.9: 85587010-85587010 on Build GRCh38
rs1044686597 Chr.9: 85586763-85586763 on Build GRCh38
rs1056269463 Chr.9: 85587040-85587040 on Build GRCh38
rs766570911 Chr.9: 85586887-85586887 on Build GRCh38
rs896366637 Chr.9: 85586679-85586679 on Build GRCh38
rs996443071 Chr.9: 85587079-85587079 on Build GRCh38
rs773686523 Chr.9: 85586814-85586814 on Build GRCh38
rs540104058 Chr.9: 85586782-85586782 on Build GRCh38

View Full Product Details