Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134033215-134033723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD3
Entrez Gene ID
8019
Gene Name
bromodomain containing 3
Gene Aliases
ORFX, RING3L
UniGene
Hs.522472
Gene Chromosome Location
Chr.9: 134030305-134068533 on Build GRCh38
Transcripts
NM_007371.3 XM_006717291.2 XM_011519052.2 XM_017015165.1 XM_017015166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5763954 c.2161G>A p.D721N Chr.9: 134033610-134033610 on Build GRCh38
7414782 c.2103G>T p.R701S Chr.9: 134033668-134033668 on Build GRCh38
4476764 c.2095C>T p.P699S Chr.9: 134033676-134033676 on Build GRCh38
5983732 c.2176G>C p.E726Q Chr.9: 134033595-134033595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs117016781 Chr.9: 134033425-134033425 on Build GRCh38
rs201661238 Chr.9: 134033455-134033455 on Build GRCh38
rs749179090 Chr.9: 134033581-134033581 on Build GRCh38
rs564145386 Chr.9: 134033566-134033566 on Build GRCh38
rs750137578 Chr.9: 134033534-134033534 on Build GRCh38
rs147541047 Chr.9: 134033697-134033697 on Build GRCh38
rs201715107 Chr.9: 134033646-134033646 on Build GRCh38
rs893073088 Chr.9: 134033587-134033587 on Build GRCh38
rs200598840 Chr.9: 134033674-134033674 on Build GRCh38
rs201500184 Chr.9: 134033675-134033675 on Build GRCh38
rs191946819 Chr.9: 134033413-134033413 on Build GRCh38
rs1031613438 Chr.9: 134033688-134033688 on Build GRCh38
rs112507804 Chr.9: 134033549-134033549 on Build GRCh38
rs775107837 Chr.9: 134033653-134033655 on Build GRCh38
rs753316014 Chr.9: 134033647-134033647 on Build GRCh38
rs200215272 Chr.9: 134033460-134033460 on Build GRCh38
rs201856792 Chr.9: 134033245-134033245 on Build GRCh38
rs774314861 Chr.9: 134033585-134033585 on Build GRCh38
rs201884964 Chr.9: 134033439-134033439 on Build GRCh38
rs200262807 Chr.9: 134033377-134033377 on Build GRCh38
rs770744699 Chr.9: 134033318-134033318 on Build GRCh38
rs1002625691 Chr.9: 134033435-134033435 on Build GRCh38
rs201137123 Chr.9: 134033351-134033351 on Build GRCh38
rs199540864 Chr.9: 134033431-134033431 on Build GRCh38
rs772859912 Chr.9: 134033620-134033620 on Build GRCh38
rs964052833 Chr.9: 134033571-134033571 on Build GRCh38
rs143290331 Chr.9: 134033623-134033623 on Build GRCh38
rs919777143 Chr.9: 134033317-134033317 on Build GRCh38
rs1057224336 Chr.9: 134033447-134033447 on Build GRCh38
rs758733130 Chr.9: 134033653-134033653 on Build GRCh38
rs917516837 Chr.9: 134033242-134033242 on Build GRCh38
rs113698627 Chr.9: 134033416-134033416 on Build GRCh38
rs200585854 Chr.9: 134033453-134033453 on Build GRCh38
rs56166940 Chr.9: 134033698-134033698 on Build GRCh38
rs201986524 Chr.9: 134033291-134033291 on Build GRCh38
rs199973695 Chr.9: 134033250-134033250 on Build GRCh38
rs17151353 Chr.9: 134033463-134033463 on Build GRCh38
rs778044579 Chr.9: 134033556-134033556 on Build GRCh38
rs754330448 Chr.9: 134033548-134033548 on Build GRCh38
rs767271095 Chr.9: 134033593-134033593 on Build GRCh38
rs918673787 Chr.9: 134033465-134033465 on Build GRCh38
rs945898275 Chr.9: 134033622-134033622 on Build GRCh38
rs1046808145 Chr.9: 134033375-134033375 on Build GRCh38
rs547796842 Chr.9: 134033546-134033546 on Build GRCh38
rs200488233 Chr.9: 134033418-134033418 on Build GRCh38
rs1001099094 Chr.9: 134033687-134033687 on Build GRCh38
rs34389248 Chr.9: 134033303-134033303 on Build GRCh38
rs201808057 Chr.9: 134033358-134033358 on Build GRCh38
rs906926648 Chr.9: 134033627-134033627 on Build GRCh38
rs748046104 Chr.9: 134033701-134033701 on Build GRCh38
rs747210334 Chr.9: 134033563-134033563 on Build GRCh38
rs566397754 Chr.9: 134033302-134033302 on Build GRCh38
rs769309748 Chr.9: 134033570-134033574 on Build GRCh38
rs758758415 Chr.9: 134033555-134033555 on Build GRCh38
rs201025845 Chr.9: 134033396-134033396 on Build GRCh38
rs1042736436 Chr.9: 134033662-134033662 on Build GRCh38
rs761589766 Chr.9: 134033588-134033588 on Build GRCh38
rs754582064 Chr.9: 134033645-134033645 on Build GRCh38
rs931224455 Chr.9: 134033584-134033584 on Build GRCh38
rs745664416 Chr.9: 134033423-134033423 on Build GRCh38
rs201205178 Chr.9: 134033260-134033260 on Build GRCh38
rs776064740 Chr.9: 134033316-134033316 on Build GRCh38
rs372807731 Chr.9: 134033554-134033554 on Build GRCh38
rs776610067 Chr.9: 134033580-134033580 on Build GRCh38
rs956804555 Chr.9: 134033252-134033252 on Build GRCh38
rs770977813 Chr.9: 134033567-134033567 on Build GRCh38
rs768562282 Chr.9: 134033583-134033583 on Build GRCh38
rs113301770 Chr.9: 134033638-134033638 on Build GRCh38
rs959574543 Chr.9: 134033429-134033429 on Build GRCh38
rs889701192 Chr.9: 134033254-134033254 on Build GRCh38
rs1054657817 Chr.9: 134033319-134033319 on Build GRCh38
rs433402 Chr.9: 134033505-134033505 on Build GRCh38
rs1001066942 Chr.9: 134033499-134033499 on Build GRCh38
rs201871642 Chr.9: 134033550-134033550 on Build GRCh38
rs200987204 Chr.9: 134033438-134033438 on Build GRCh38
rs987245097 Chr.9: 134033263-134033263 on Build GRCh38
rs199674805 Chr.9: 134033449-134033449 on Build GRCh38
rs778004765 Chr.9: 134033659-134033659 on Build GRCh38
rs199922240 Chr.9: 134033535-134033535 on Build GRCh38
rs748745510 Chr.9: 134033693-134033704 on Build GRCh38

View Full Product Details