Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135521318-135521816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCN1
Entrez Gene ID
3933
Gene Name
lipocalin 1
Gene Aliases
PMFA, TLC, TP, VEGP
UniGene
Hs.530311
Gene Chromosome Location
Chr.9: 135521296-135526540 on Build GRCh38
Transcripts
NM_001252617.1 NM_001252618.1 NM_001252619.1 NM_002297.3 XM_011518675.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
204739 c.30T>C p.L10L Chr.9: 135521527-135521527 on Build GRCh38
3952475 c.1A>T p.M1L Chr.9: 135521498-135521498 on Build GRCh38
6841055 c.22G>A p.V8I Chr.9: 135521519-135521519 on Build GRCh38
5658021 c.75C>A p.D25E Chr.9: 135521572-135521572 on Build GRCh38
3905514 c.21C>T p.A7A Chr.9: 135521518-135521518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866568295 Chr.9: 135521463-135521463 on Build GRCh38
rs755106174 Chr.9: 135521598-135521598 on Build GRCh38
rs200530565 Chr.9: 135521519-135521519 on Build GRCh38
rs1052006703 Chr.9: 135521498-135521498 on Build GRCh38
rs1036109908 Chr.9: 135521380-135521380 on Build GRCh38
rs201058632 Chr.9: 135521493-135521493 on Build GRCh38
rs61735422 Chr.9: 135521555-135521555 on Build GRCh38
rs576047351 Chr.9: 135521573-135521573 on Build GRCh38
rs770691280 Chr.9: 135521478-135521478 on Build GRCh38
rs764556368 Chr.9: 135521497-135521497 on Build GRCh38
rs866382803 Chr.9: 135521606-135521606 on Build GRCh38
rs769628368 Chr.9: 135521633-135521633 on Build GRCh38
rs759401763 Chr.9: 135521559-135521559 on Build GRCh38
rs749340456 Chr.9: 135521526-135521526 on Build GRCh38
rs541956450 Chr.9: 135521596-135521596 on Build GRCh38
rs898916500 Chr.9: 135521393-135521393 on Build GRCh38
rs748274038 Chr.9: 135521607-135521607 on Build GRCh38
rs566136825 Chr.9: 135521704-135521704 on Build GRCh38
rs940935174 Chr.9: 135521770-135521770 on Build GRCh38
rs368953450 Chr.9: 135521479-135521479 on Build GRCh38
rs1029110788 Chr.9: 135521717-135521717 on Build GRCh38
rs979238615 Chr.9: 135521708-135521708 on Build GRCh38
rs890111184 Chr.9: 135521365-135521365 on Build GRCh38
rs543076568 Chr.9: 135521557-135521557 on Build GRCh38
rs199966911 Chr.9: 135521499-135521499 on Build GRCh38
rs139335104 Chr.9: 135521549-135521549 on Build GRCh38
rs562674837 Chr.9: 135521656-135521656 on Build GRCh38
rs555089513 Chr.9: 135521560-135521560 on Build GRCh38
rs759044260 Chr.9: 135521482-135521482 on Build GRCh38
rs772084504 Chr.9: 135521470-135521470 on Build GRCh38
rs994644882 Chr.9: 135521407-135521407 on Build GRCh38
rs551872805 Chr.9: 135521797-135521797 on Build GRCh38
rs912112958 Chr.9: 135521761-135521761 on Build GRCh38
rs143259160 Chr.9: 135521502-135521502 on Build GRCh38
rs561421478 Chr.9: 135521725-135521725 on Build GRCh38
rs779071186 Chr.9: 135521602-135521602 on Build GRCh38
rs1003852504 Chr.9: 135521432-135521432 on Build GRCh38
rs573421494 Chr.9: 135521562-135521562 on Build GRCh38
rs747010685 Chr.9: 135521547-135521547 on Build GRCh38
rs762156818 Chr.9: 135521578-135521578 on Build GRCh38
rs372970561 Chr.9: 135521597-135521597 on Build GRCh38
rs547830553 Chr.9: 135521703-135521703 on Build GRCh38
rs527347336 Chr.9: 135521737-135521737 on Build GRCh38
rs780444789 Chr.9: 135521523-135521523 on Build GRCh38
rs62577203 Chr.9: 135521716-135521716 on Build GRCh38
rs768895070 Chr.9: 135521527-135521527 on Build GRCh38
rs1034898560 Chr.9: 135521619-135521619 on Build GRCh38
rs768078622 Chr.9: 135521579-135521579 on Build GRCh38
rs773618500 Chr.9: 135521378-135521378 on Build GRCh38
rs375223502 Chr.9: 135521630-135521630 on Build GRCh38
rs760513644 Chr.9: 135521475-135521475 on Build GRCh38
rs529632731 Chr.9: 135521679-135521679 on Build GRCh38
rs757094899 Chr.9: 135521622-135521622 on Build GRCh38
rs749004316 Chr.9: 135521634-135521634 on Build GRCh38
rs143345996 Chr.9: 135521569-135521569 on Build GRCh38
rs777829454 Chr.9: 135521543-135521543 on Build GRCh38
rs764956367 Chr.9: 135521490-135521490 on Build GRCh38
rs1126820 Chr.9: 135521566-135521566 on Build GRCh38
rs369805485 Chr.9: 135521631-135521631 on Build GRCh38
rs781224936 Chr.9: 135521625-135521625 on Build GRCh38
rs762485484 Chr.9: 135521494-135521494 on Build GRCh38
rs781474202 Chr.9: 135521510-135521510 on Build GRCh38
rs773312165 Chr.9: 135521473-135521473 on Build GRCh38
rs534886215 Chr.9: 135521408-135521408 on Build GRCh38
rs969309115 Chr.9: 135521425-135521425 on Build GRCh38
rs750703568 Chr.9: 135521595-135521595 on Build GRCh38
rs988812347 Chr.9: 135521476-135521476 on Build GRCh38
rs553106680 Chr.9: 135521420-135521420 on Build GRCh38
rs978926372 Chr.9: 135521486-135521486 on Build GRCh38
rs368871873 Chr.9: 135521590-135521590 on Build GRCh38
rs147081757 Chr.9: 135521572-135521572 on Build GRCh38
rs922114952 Chr.9: 135521491-135521491 on Build GRCh38
rs371365986 Chr.9: 135521462-135521462 on Build GRCh38

View Full Product Details