Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135565324-135565816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAEP
Entrez Gene ID
5047
Gene Name
progestagen associated endometrial protein
Gene Aliases
GD, GdA, GdF, GdS, PAEG, PEP, PP14
UniGene
Hs.532325
Gene Chromosome Location
Chr.9: 135561624-135566776 on Build GRCh38
Transcripts
NM_001018048.1 NM_001018049.2 NM_002571.3 XM_011518745.2 XM_011518746.2 XM_011518747.2 XM_011518748.1 XM_011518749.2 XM_011518751.1 XM_011518752.2 XM_017014782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3952477 c.220C>T p.P74S Chr.9: 135565357-135565357 on Build GRCh38
5626169 c.529C>G p.P177A Chr.9: 135565787-135565787 on Build GRCh38
3763742 c.536G>A p.R179H Chr.9: 135565794-135565794 on Build GRCh38
1106656 c.519G>A p.Q173Q Chr.9: 135565507-135565507 on Build GRCh38
3952476 c.215G>T p.G72V Chr.9: 135565352-135565352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780524826 Chr.9: 135565343-135565343 on Build GRCh38
rs143616209 Chr.9: 135565794-135565794 on Build GRCh38
rs987676682 Chr.9: 135565683-135565683 on Build GRCh38
rs771917399 Chr.9: 135565654-135565654 on Build GRCh38
rs780761801 Chr.9: 135565357-135565357 on Build GRCh38
rs903044936 Chr.9: 135565542-135565542 on Build GRCh38
rs933546102 Chr.9: 135565669-135565669 on Build GRCh38
rs773164732 Chr.9: 135565581-135565581 on Build GRCh38
rs760364342 Chr.9: 135565585-135565585 on Build GRCh38
rs3831489 Chr.9: 135565630-135565630 on Build GRCh38
rs991612260 Chr.9: 135565584-135565584 on Build GRCh38
rs561964518 Chr.9: 135565643-135565643 on Build GRCh38
rs753319217 Chr.9: 135565645-135565645 on Build GRCh38
rs759775076 Chr.9: 135565429-135565429 on Build GRCh38
rs868377271 Chr.9: 135565741-135565741 on Build GRCh38
rs783774 Chr.9: 135565636-135565636 on Build GRCh38
rs111290016 Chr.9: 135565638-135565638 on Build GRCh38
rs371331642 Chr.9: 135565531-135565531 on Build GRCh38
rs777682171 Chr.9: 135565737-135565737 on Build GRCh38
rs763483993 Chr.9: 135565769-135565769 on Build GRCh38
rs754340819 Chr.9: 135565439-135565439 on Build GRCh38
rs756566132 Chr.9: 135565641-135565641 on Build GRCh38
rs747730596 Chr.9: 135565653-135565653 on Build GRCh38
rs747182758 Chr.9: 135565549-135565549 on Build GRCh38
rs1047730549 Chr.9: 135565397-135565397 on Build GRCh38
rs200107494 Chr.9: 135565504-135565504 on Build GRCh38
rs943476049 Chr.9: 135565726-135565726 on Build GRCh38
rs562372164 Chr.9: 135565493-135565493 on Build GRCh38
rs150080982 Chr.9: 135565440-135565440 on Build GRCh38
rs370101071 Chr.9: 135565751-135565751 on Build GRCh38
rs767248188 Chr.9: 135565418-135565418 on Build GRCh38
rs779351497 Chr.9: 135565378-135565378 on Build GRCh38
rs769614075 Chr.9: 135565602-135565606 on Build GRCh38
rs770696500 Chr.9: 135565750-135565750 on Build GRCh38
rs753184926 Chr.9: 135565626-135565626 on Build GRCh38
rs761339596 Chr.9: 135565417-135565417 on Build GRCh38
rs769077012 Chr.9: 135565368-135565368 on Build GRCh38
rs1011535253 Chr.9: 135565513-135565513 on Build GRCh38
rs149314008 Chr.9: 135565483-135565483 on Build GRCh38
rs776711925 Chr.9: 135565473-135565473 on Build GRCh38
rs377698141 Chr.9: 135565412-135565412 on Build GRCh38
rs758531872 Chr.9: 135565461-135565461 on Build GRCh38
rs781213479 Chr.9: 135565573-135565573 on Build GRCh38
rs751907235 Chr.9: 135565788-135565788 on Build GRCh38
rs773135393 Chr.9: 135565623-135565623 on Build GRCh38
rs756667245 Chr.9: 135565356-135565356 on Build GRCh38
rs531806388 Chr.9: 135565577-135565577 on Build GRCh38
rs543583253 Chr.9: 135565466-135565466 on Build GRCh38
rs958482723 Chr.9: 135565578-135565578 on Build GRCh38
rs766176374 Chr.9: 135565597-135565597 on Build GRCh38
rs762888016 Chr.9: 135565410-135565410 on Build GRCh38
rs778548305 Chr.9: 135565650-135565650 on Build GRCh38
rs758848176 Chr.9: 135565628-135565628 on Build GRCh38
rs200082958 Chr.9: 135565630-135565630 on Build GRCh38
rs371322222 Chr.9: 135565427-135565427 on Build GRCh38
rs759119544 Chr.9: 135565757-135565757 on Build GRCh38
rs764259096 Chr.9: 135565633-135565633 on Build GRCh38
rs779664158 Chr.9: 135565644-135565644 on Build GRCh38
rs1002718377 Chr.9: 135565569-135565569 on Build GRCh38
rs748589237 Chr.9: 135565450-135565450 on Build GRCh38
rs373160904 Chr.9: 135565383-135565383 on Build GRCh38
rs763280519 Chr.9: 135565632-135565632 on Build GRCh38
rs779716594 Chr.9: 135565430-135565430 on Build GRCh38
rs749989070 Chr.9: 135565354-135565354 on Build GRCh38
rs368093964 Chr.9: 135565544-135565544 on Build GRCh38
rs775407150 Chr.9: 135565631-135565631 on Build GRCh38
rs753262643 Chr.9: 135565382-135565382 on Build GRCh38
rs186327730 Chr.9: 135565576-135565576 on Build GRCh38
rs751039762 Chr.9: 135565640-135565640 on Build GRCh38
rs767928614 Chr.9: 135565639-135565639 on Build GRCh38
rs771796316 Chr.9: 135565535-135565535 on Build GRCh38
rs775240380 Chr.9: 135565614-135565614 on Build GRCh38
rs371952789 Chr.9: 135565448-135565448 on Build GRCh38
rs746851084 Chr.9: 135565739-135565739 on Build GRCh38
rs749179379 Chr.9: 135565477-135565477 on Build GRCh38
rs765412011 Chr.9: 135565629-135565629 on Build GRCh38
rs779108772 Chr.9: 135565443-135565443 on Build GRCh38
rs757497696 Chr.9: 135565553-135565553 on Build GRCh38
rs751398314 Chr.9: 135565416-135565416 on Build GRCh38
rs778010278 Chr.9: 135565540-135565540 on Build GRCh38
rs1030643569 Chr.9: 135565708-135565708 on Build GRCh38
rs373972549 Chr.9: 135565534-135565534 on Build GRCh38
rs144616486 Chr.9: 135565508-135565508 on Build GRCh38
rs764631815 Chr.9: 135565775-135565775 on Build GRCh38
rs745697635 Chr.9: 135565580-135565580 on Build GRCh38
rs773138552 Chr.9: 135565387-135565387 on Build GRCh38
rs373981009 Chr.9: 135565467-135565467 on Build GRCh38
rs770937897 Chr.9: 135565468-135565468 on Build GRCh38
rs911030127 Chr.9: 135565554-135565554 on Build GRCh38
rs138236506 Chr.9: 135565789-135565789 on Build GRCh38
rs759145812 Chr.9: 135565612-135565612 on Build GRCh38
rs750586274 Chr.9: 135565793-135565793 on Build GRCh38
rs377654834 Chr.9: 135565517-135565517 on Build GRCh38
rs773993548 Chr.9: 135565497-135565497 on Build GRCh38
rs374250987 Chr.9: 135565568-135565568 on Build GRCh38
rs769294960 Chr.9: 135565760-135565760 on Build GRCh38
rs774889770 Chr.9: 135565763-135565763 on Build GRCh38
rs764521684 Chr.9: 135565724-135565724 on Build GRCh38
rs748373190 Chr.9: 135565624-135565633 on Build GRCh38
rs572439418 Chr.9: 135565509-135565509 on Build GRCh38
rs2070909 Chr.9: 135565363-135565363 on Build GRCh38
rs765941303 Chr.9: 135565524-135565524 on Build GRCh38
rs537242473 Chr.9: 135565785-135565799 on Build GRCh38
rs745995335 Chr.9: 135565574-135565574 on Build GRCh38
rs769709895 Chr.9: 135565407-135565407 on Build GRCh38
rs141511524 Chr.9: 135565463-135565463 on Build GRCh38
rs754462273 Chr.9: 135565646-135565646 on Build GRCh38
rs375530489 Chr.9: 135565580-135565580 on Build GRCh38
rs775336705 Chr.9: 135565409-135565409 on Build GRCh38
rs776262704 Chr.9: 135565603-135565603 on Build GRCh38

View Full Product Details