Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136218557-136219055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QSOX2
Entrez Gene ID
169714
Gene Name
quiescin sulfhydryl oxidase 2
Gene Aliases
QSCN6L1, SOXN
UniGene
Hs.144073
Gene Chromosome Location
Chr.9: 136206333-136245841 on Build GRCh38
Transcripts
NM_181701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1106744 c.1024C>T p.H342Y Chr.9: 136218741-136218741 on Build GRCh38
1461038 c.1055C>T p.T352M Chr.9: 136218710-136218710 on Build GRCh38
241334 c.1039G>A p.G347R Chr.9: 136218726-136218726 on Build GRCh38
1461039 c.1022C>T p.A341V Chr.9: 136218743-136218743 on Build GRCh38
291176 c.972G>A p.T324T Chr.9: 136218793-136218793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760195182 Chr.9: 136219002-136219002 on Build GRCh38
rs749995120 Chr.9: 136218734-136218734 on Build GRCh38
rs377581112 Chr.9: 136218821-136218821 on Build GRCh38
rs761486801 Chr.9: 136218814-136218814 on Build GRCh38
rs778977056 Chr.9: 136218990-136218990 on Build GRCh38
rs200912016 Chr.9: 136218600-136218600 on Build GRCh38
rs780402478 Chr.9: 136218670-136218670 on Build GRCh38
rs765704218 Chr.9: 136218838-136218838 on Build GRCh38
rs756684580 Chr.9: 136218662-136218662 on Build GRCh38
rs375020015 Chr.9: 136218653-136218653 on Build GRCh38
rs779108665 Chr.9: 136218679-136218679 on Build GRCh38
rs965095471 Chr.9: 136218807-136218807 on Build GRCh38
rs775276630 Chr.9: 136218709-136218709 on Build GRCh38
rs1011433810 Chr.9: 136218669-136218669 on Build GRCh38
rs772734481 Chr.9: 136218816-136218816 on Build GRCh38
rs199709197 Chr.9: 136219028-136219028 on Build GRCh38
rs1044442743 Chr.9: 136218611-136218612 on Build GRCh38
rs534667675 Chr.9: 136218842-136218842 on Build GRCh38
rs373702986 Chr.9: 136219024-136219024 on Build GRCh38
rs771384898 Chr.9: 136218993-136218994 on Build GRCh38
rs377419025 Chr.9: 136218682-136218682 on Build GRCh38
rs146448254 Chr.9: 136218733-136218733 on Build GRCh38
rs368839456 Chr.9: 136218654-136218654 on Build GRCh38
rs755566167 Chr.9: 136218735-136218735 on Build GRCh38
rs754363653 Chr.9: 136218829-136218829 on Build GRCh38
rs61744120 Chr.9: 136218710-136218710 on Build GRCh38
rs931181535 Chr.9: 136218581-136218581 on Build GRCh38
rs755348356 Chr.9: 136218746-136218746 on Build GRCh38
rs533080907 Chr.9: 136218730-136218730 on Build GRCh38
rs916320402 Chr.9: 136218926-136218926 on Build GRCh38
rs767002715 Chr.9: 136218646-136218646 on Build GRCh38
rs768600766 Chr.9: 136218811-136218811 on Build GRCh38
rs372770375 Chr.9: 136218631-136218631 on Build GRCh38
rs36061641 Chr.9: 136218619-136218619 on Build GRCh38
rs562718746 Chr.9: 136218972-136218972 on Build GRCh38
rs200647931 Chr.9: 136218835-136218835 on Build GRCh38
rs745530657 Chr.9: 136218922-136218922 on Build GRCh38
rs940188156 Chr.9: 136218599-136218599 on Build GRCh38
rs747032551 Chr.9: 136218700-136218700 on Build GRCh38
rs552700851 Chr.9: 136218889-136218889 on Build GRCh38
rs900338784 Chr.9: 136218920-136218920 on Build GRCh38
rs201549036 Chr.9: 136218828-136218828 on Build GRCh38
rs774324178 Chr.9: 136218812-136218812 on Build GRCh38
rs929888494 Chr.9: 136218965-136218965 on Build GRCh38
rs1004579050 Chr.9: 136218956-136218956 on Build GRCh38
rs983206727 Chr.9: 136218595-136218595 on Build GRCh38
rs747175246 Chr.9: 136218794-136218794 on Build GRCh38
rs945575933 Chr.9: 136218605-136218605 on Build GRCh38
rs191663534 Chr.9: 136219004-136219004 on Build GRCh38
rs573806099 Chr.9: 136219001-136219001 on Build GRCh38
rs748309442 Chr.9: 136218681-136218681 on Build GRCh38
rs769823166 Chr.9: 136218703-136218703 on Build GRCh38
rs77984401 Chr.9: 136218673-136218673 on Build GRCh38
rs759240058 Chr.9: 136219012-136219012 on Build GRCh38
rs754275770 Chr.9: 136218743-136218743 on Build GRCh38
rs867980366 Chr.9: 136218786-136218786 on Build GRCh38
rs770914309 Chr.9: 136218805-136218805 on Build GRCh38
rs756770808 Chr.9: 136218815-136218815 on Build GRCh38
rs576274721 Chr.9: 136218611-136218611 on Build GRCh38
rs879242240 Chr.9: 136218867-136218867 on Build GRCh38
rs891609026 Chr.9: 136218652-136218652 on Build GRCh38
rs927774650 Chr.9: 136218618-136218618 on Build GRCh38
rs369920025 Chr.9: 136218726-136218726 on Build GRCh38
rs377023676 Chr.9: 136218701-136218701 on Build GRCh38
rs1022536307 Chr.9: 136218809-136218809 on Build GRCh38
rs199618366 Chr.9: 136218598-136218598 on Build GRCh38
rs777836530 Chr.9: 136218793-136218793 on Build GRCh38
rs183495350 Chr.9: 136218634-136218634 on Build GRCh38
rs559591326 Chr.9: 136219007-136219007 on Build GRCh38
rs915474047 Chr.9: 136218589-136218589 on Build GRCh38
rs564525785 Chr.9: 136218607-136218607 on Build GRCh38
rs1038040926 Chr.9: 136218957-136218957 on Build GRCh38
rs778128288 Chr.9: 136218843-136218843 on Build GRCh38
rs865807908 Chr.9: 136218971-136218971 on Build GRCh38
rs758676802 Chr.9: 136218841-136218841 on Build GRCh38
rs187369560 Chr.9: 136218739-136218739 on Build GRCh38
rs1006708210 Chr.9: 136218976-136218976 on Build GRCh38
rs753109890 Chr.9: 136218840-136218840 on Build GRCh38
rs561912050 Chr.9: 136219000-136219000 on Build GRCh38
rs747220596 Chr.9: 136218886-136218886 on Build GRCh38
rs954373660 Chr.9: 136218706-136218706 on Build GRCh38
rs1053147506 Chr.9: 136218625-136218625 on Build GRCh38
rs377647134 Chr.9: 136218757-136218757 on Build GRCh38
rs113368496 Chr.9: 136218619-136218619 on Build GRCh38
rs890106509 Chr.9: 136218952-136218952 on Build GRCh38
rs753201873 Chr.9: 136219019-136219019 on Build GRCh38
rs913236898 Chr.9: 136219011-136219011 on Build GRCh38
rs751669325 Chr.9: 136218980-136218980 on Build GRCh38
rs202194021 Chr.9: 136218806-136218806 on Build GRCh38
rs372069538 Chr.9: 136218930-136218930 on Build GRCh38
rs765792193 Chr.9: 136219014-136219014 on Build GRCh38
rs551924895 Chr.9: 136218958-136218958 on Build GRCh38
rs374042823 Chr.9: 136218732-136218732 on Build GRCh38
rs759026519 Chr.9: 136219012-136219012 on Build GRCh38
rs538251617 Chr.9: 136218967-136218967 on Build GRCh38
rs543636245 Chr.9: 136218989-136218989 on Build GRCh38
rs140545425 Chr.9: 136218691-136218691 on Build GRCh38
rs756834822 Chr.9: 136218645-136218645 on Build GRCh38
rs778077729 Chr.9: 136218629-136218630 on Build GRCh38
rs758688345 Chr.9: 136218770-136218770 on Build GRCh38
rs748041440 Chr.9: 136218999-136218999 on Build GRCh38
rs751254331 Chr.9: 136218628-136218628 on Build GRCh38
rs908681151 Chr.9: 136218596-136218596 on Build GRCh38
rs576312795 Chr.9: 136218981-136218981 on Build GRCh38
rs928915273 Chr.9: 136218992-136218992 on Build GRCh38
rs111419070 Chr.9: 136218747-136218747 on Build GRCh38
rs928130944 Chr.9: 136218888-136218888 on Build GRCh38
rs760235373 Chr.9: 136218827-136218827 on Build GRCh38
rs12684650 Chr.9: 136218808-136218808 on Build GRCh38
rs559659543 Chr.9: 136218727-136218727 on Build GRCh38
rs991001221 Chr.9: 136218899-136218899 on Build GRCh38
rs749706199 Chr.9: 136218671-136218671 on Build GRCh38
rs371056879 Chr.9: 136218783-136218783 on Build GRCh38
rs575247172 Chr.9: 136218608-136218608 on Build GRCh38
rs533000915 Chr.9: 136218612-136218612 on Build GRCh38
rs776856025 Chr.9: 136219000-136219001 on Build GRCh38

View Full Product Details