Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136370690-136371183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
UniGene
Hs.694071
Gene Chromosome Location
Chr.9: 136363956-136373681 on Build GRCh38
Transcripts
NM_052813.4 NM_052814.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4551742 c.538G>A p.E180K Chr.9: 136370930-136370930 on Build GRCh38
6371709 c.514A>G p.S172G Chr.9: 136370954-136370954 on Build GRCh38
4554563 c.624G>A p.L208L Chr.9: 136370844-136370844 on Build GRCh38
4876136 c.408G>A p.L136L Chr.9: 136371060-136371060 on Build GRCh38
5472705 c.504C>T p.C168C Chr.9: 136370964-136370964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775704708 Chr.9: 136370756-136370756 on Build GRCh38
rs768842654 Chr.9: 136370823-136370823 on Build GRCh38
rs539919395 Chr.9: 136371013-136371013 on Build GRCh38
rs201183869 Chr.9: 136371058-136371058 on Build GRCh38
rs768281299 Chr.9: 136370882-136370882 on Build GRCh38
rs772862677 Chr.9: 136371000-136371000 on Build GRCh38
rs769711471 Chr.9: 136370823-136370824 on Build GRCh38
rs753025581 Chr.9: 136370915-136370915 on Build GRCh38
rs758246421 Chr.9: 136371081-136371081 on Build GRCh38
rs367638485 Chr.9: 136371074-136371074 on Build GRCh38
rs372553152 Chr.9: 136370828-136370828 on Build GRCh38
rs528585158 Chr.9: 136370919-136370919 on Build GRCh38
rs773083092 Chr.9: 136370731-136370731 on Build GRCh38
rs758297418 Chr.9: 136371152-136371152 on Build GRCh38
rs557252130 Chr.9: 136370744-136370744 on Build GRCh38
rs139295959 Chr.9: 136370876-136370876 on Build GRCh38
rs931489423 Chr.9: 136370752-136370752 on Build GRCh38
rs567447776 Chr.9: 136370778-136370778 on Build GRCh38
rs765697772 Chr.9: 136371018-136371018 on Build GRCh38
rs181987989 Chr.9: 136370837-136370837 on Build GRCh38
rs558145829 Chr.9: 136370958-136370958 on Build GRCh38
rs776001547 Chr.9: 136371020-136371020 on Build GRCh38
rs977007616 Chr.9: 136371038-136371038 on Build GRCh38
rs766127176 Chr.9: 136370733-136370733 on Build GRCh38
rs948124005 Chr.9: 136370894-136370894 on Build GRCh38
rs560748276 Chr.9: 136370770-136370770 on Build GRCh38
rs760181332 Chr.9: 136370952-136370952 on Build GRCh38
rs751375400 Chr.9: 136371160-136371160 on Build GRCh38
rs760681802 Chr.9: 136371142-136371142 on Build GRCh38
rs759954061 Chr.9: 136370956-136370956 on Build GRCh38
rs769183409 Chr.9: 136371137-136371137 on Build GRCh38
rs773353025 Chr.9: 136370826-136370826 on Build GRCh38
rs760577746 Chr.9: 136370773-136370773 on Build GRCh38
rs759264924 Chr.9: 136370951-136370951 on Build GRCh38
rs749290461 Chr.9: 136370979-136370979 on Build GRCh38
rs1031537800 Chr.9: 136370817-136370817 on Build GRCh38
rs774113192 Chr.9: 136370885-136370885 on Build GRCh38
rs764104112 Chr.9: 136370727-136370727 on Build GRCh38
rs547043978 Chr.9: 136370940-136370940 on Build GRCh38
rs140508365 Chr.9: 136370957-136370957 on Build GRCh38
rs760135155 Chr.9: 136371017-136371017 on Build GRCh38
rs139395199 Chr.9: 136371141-136371141 on Build GRCh38
rs779130876 Chr.9: 136370912-136370912 on Build GRCh38
rs760658143 Chr.9: 136370732-136370732 on Build GRCh38
rs772249417 Chr.9: 136370873-136370873 on Build GRCh38
rs752451970 Chr.9: 136370781-136370781 on Build GRCh38
rs755380617 Chr.9: 136370906-136370906 on Build GRCh38
rs757342734 Chr.9: 136370939-136370939 on Build GRCh38
rs370234838 Chr.9: 136370740-136370740 on Build GRCh38
rs761400518 Chr.9: 136370843-136370843 on Build GRCh38
rs777535858 Chr.9: 136371084-136371084 on Build GRCh38
rs375341696 Chr.9: 136371047-136371047 on Build GRCh38
rs757627986 Chr.9: 136371025-136371025 on Build GRCh38
rs368657832 Chr.9: 136370950-136370950 on Build GRCh38
rs753644019 Chr.9: 136370736-136370736 on Build GRCh38
rs112244317 Chr.9: 136370960-136370960 on Build GRCh38
rs756885929 Chr.9: 136370744-136370745 on Build GRCh38
rs776391777 Chr.9: 136370777-136370777 on Build GRCh38
rs34971035 Chr.9: 136371144-136371144 on Build GRCh38
rs190628465 Chr.9: 136370824-136370824 on Build GRCh38
rs753492242 Chr.9: 136370948-136370948 on Build GRCh38
rs754223003 Chr.9: 136370864-136370864 on Build GRCh38
rs373386414 Chr.9: 136370747-136370747 on Build GRCh38
rs774424748 Chr.9: 136370877-136370877 on Build GRCh38
rs763564545 Chr.9: 136370793-136370793 on Build GRCh38
rs370943995 Chr.9: 136370909-136370909 on Build GRCh38
rs1043128646 Chr.9: 136370938-136370938 on Build GRCh38
rs375434244 Chr.9: 136371138-136371138 on Build GRCh38
rs769292662 Chr.9: 136371116-136371116 on Build GRCh38
rs766925666 Chr.9: 136370853-136370853 on Build GRCh38
rs768564945 Chr.9: 136370943-136370943 on Build GRCh38
rs746700619 Chr.9: 136371087-136371087 on Build GRCh38
rs144115318 Chr.9: 136370890-136370890 on Build GRCh38
rs757991472 Chr.9: 136370741-136370741 on Build GRCh38
rs35984637 Chr.9: 136370988-136370988 on Build GRCh38
rs78538457 Chr.9: 136370955-136370955 on Build GRCh38
rs895657922 Chr.9: 136370954-136370954 on Build GRCh38
rs982980603 Chr.9: 136370899-136370899 on Build GRCh38
rs755256889 Chr.9: 136370865-136370865 on Build GRCh38
rs572881831 Chr.9: 136370872-136370872 on Build GRCh38
rs778718632 Chr.9: 136371065-136371065 on Build GRCh38
rs765063795 Chr.9: 136370779-136370779 on Build GRCh38
rs149206311 Chr.9: 136371026-136371026 on Build GRCh38
rs775922619 Chr.9: 136371117-136371117 on Build GRCh38
rs745801532 Chr.9: 136370764-136370765 on Build GRCh38
rs752622733 Chr.9: 136371076-136371076 on Build GRCh38
rs143393026 Chr.9: 136371062-136371062 on Build GRCh38
rs369155916 Chr.9: 136370878-136370878 on Build GRCh38
rs780444469 Chr.9: 136370859-136370859 on Build GRCh38
rs567812719 Chr.9: 136370840-136370840 on Build GRCh38
rs765062166 Chr.9: 136370925-136370927 on Build GRCh38
rs779733829 Chr.9: 136370890-136370899 on Build GRCh38
rs781419489 Chr.9: 136370975-136370975 on Build GRCh38
rs371695061 Chr.9: 136371136-136371136 on Build GRCh38
rs550282141 Chr.9: 136370821-136370821 on Build GRCh38
rs771788289 Chr.9: 136370995-136370995 on Build GRCh38
rs374298700 Chr.9: 136370918-136370918 on Build GRCh38
rs199631159 Chr.9: 136370816-136370816 on Build GRCh38
rs375692399 Chr.9: 136370716-136370716 on Build GRCh38
rs780845153 Chr.9: 136370752-136370753 on Build GRCh38
rs746279044 Chr.9: 136370754-136370754 on Build GRCh38
rs745377757 Chr.9: 136371105-136371105 on Build GRCh38
rs1015554677 Chr.9: 136370863-136370863 on Build GRCh38
rs145198648 Chr.9: 136371004-136371004 on Build GRCh38
rs756177404 Chr.9: 136371064-136371064 on Build GRCh38
rs372648269 Chr.9: 136370804-136370804 on Build GRCh38
rs775407569 Chr.9: 136370834-136370834 on Build GRCh38
rs747120908 Chr.9: 136370920-136370920 on Build GRCh38
rs770079443 Chr.9: 136370755-136370755 on Build GRCh38
rs750996208 Chr.9: 136370857-136370857 on Build GRCh38
rs780846137 Chr.9: 136371101-136371101 on Build GRCh38
rs779198505 Chr.9: 136370868-136370868 on Build GRCh38
rs757473624 Chr.9: 136370964-136370964 on Build GRCh38
rs932662849 Chr.9: 136370936-136370936 on Build GRCh38
rs199947855 Chr.9: 136371005-136371005 on Build GRCh38
rs1027396974 Chr.9: 136370970-136370970 on Build GRCh38
rs146808845 Chr.9: 136370949-136370949 on Build GRCh38
rs758753704 Chr.9: 136370959-136370959 on Build GRCh38
rs763026951 Chr.9: 136370725-136370725 on Build GRCh38
rs749543068 Chr.9: 136370737-136370737 on Build GRCh38
rs374999342 Chr.9: 136370805-136370805 on Build GRCh38
rs766360861 Chr.9: 136370774-136370774 on Build GRCh38
rs921246629 Chr.9: 136370935-136370935 on Build GRCh38
rs756583189 Chr.9: 136370858-136370858 on Build GRCh38
rs376998598 Chr.9: 136370748-136370748 on Build GRCh38
rs761369989 Chr.9: 136370886-136370886 on Build GRCh38
rs973150104 Chr.9: 136370867-136370867 on Build GRCh38
rs749914021 Chr.9: 136370893-136370893 on Build GRCh38
rs749499469 Chr.9: 136370760-136370760 on Build GRCh38
rs746304716 Chr.9: 136370811-136370811 on Build GRCh38
rs746649890 Chr.9: 136371003-136371003 on Build GRCh38
rs765010468 Chr.9: 136370738-136370738 on Build GRCh38
rs376932110 Chr.9: 136370836-136370836 on Build GRCh38
rs766903486 Chr.9: 136370905-136370905 on Build GRCh38
rs758115953 Chr.9: 136370792-136370792 on Build GRCh38
rs757521454 Chr.9: 136371042-136371042 on Build GRCh38
rs571629670 Chr.9: 136370941-136370941 on Build GRCh38
rs769775448 Chr.9: 136370833-136370833 on Build GRCh38
rs201802044 Chr.9: 136370953-136370953 on Build GRCh38
rs751776187 Chr.9: 136370963-136370963 on Build GRCh38
rs542455027 Chr.9: 136370769-136370769 on Build GRCh38
rs766469254 Chr.9: 136371143-136371143 on Build GRCh38
rs760128484 Chr.9: 136370898-136370898 on Build GRCh38
rs767094927 Chr.9: 136370888-136370888 on Build GRCh38
rs770545753 Chr.9: 136371089-136371089 on Build GRCh38
rs777590946 Chr.9: 136371158-136371158 on Build GRCh38
rs926038111 Chr.9: 136371096-136371096 on Build GRCh38
rs747925378 Chr.9: 136370989-136370989 on Build GRCh38
rs531220860 Chr.9: 136370832-136370832 on Build GRCh38
rs878978254 Chr.9: 136370815-136370815 on Build GRCh38

View Full Product Details