Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136659388-136659906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL7
Entrez Gene ID
51162
Gene Name
EGF like domain multiple 7
Gene Aliases
NEU1, VE-STATIN, ZNEU1
UniGene
Hs.91481
Gene Chromosome Location
Chr.9: 136654753-136672678 on Build GRCh38
Transcripts
NR_046367.1 XM_011518764.1 XM_011518765.1 XM_011518766.1 XM_011518767.1 XM_011518768.1 XM_017014795.1 XM_017014796.1 XM_017014797.1 XM_017014798.1 XM_017014799.1 NR_135132.1

Gene Symbol
HSPC324
Entrez Gene ID
101928612
Gene Name
uncharacterized LOC101928612
Gene Aliases
-
UniGene
Hs.191591
Gene Chromosome Location
Chr.9: 136648610-136660421 on Build GRCh38
Transcripts
NR_046367.1 XM_011518764.1 XM_011518765.1 XM_011518766.1 XM_011518767.1 XM_011518768.1 XM_017014795.1 XM_017014796.1 XM_017014797.1 XM_017014798.1 XM_017014799.1 NR_135132.1

SNP Detail

SNP ID Chromosome Location
rs928815032 Chr.9: 136659550-136659550 on Build GRCh38
rs992491814 Chr.9: 136659722-136659722 on Build GRCh38
rs13286733 Chr.9: 136659739-136659739 on Build GRCh38
rs528280166 Chr.9: 136659824-136659824 on Build GRCh38
rs958037692 Chr.9: 136659750-136659750 on Build GRCh38
rs957691142 Chr.9: 136659434-136659434 on Build GRCh38
rs954196885 Chr.9: 136659418-136659418 on Build GRCh38
rs1041090077 Chr.9: 136659881-136659881 on Build GRCh38
rs551296635 Chr.9: 136659869-136659869 on Build GRCh38
rs557081902 Chr.9: 136659432-136659432 on Build GRCh38
rs1034521639 Chr.9: 136659713-136659713 on Build GRCh38
rs1007810609 Chr.9: 136659701-136659701 on Build GRCh38
rs953235995 Chr.9: 136659425-136659425 on Build GRCh38
rs981587408 Chr.9: 136659526-136659526 on Build GRCh38
rs1006870298 Chr.9: 136659440-136659440 on Build GRCh38
rs992075238 Chr.9: 136659883-136659883 on Build GRCh38
rs952517624 Chr.9: 136659484-136659484 on Build GRCh38
rs567396470 Chr.9: 136659460-136659460 on Build GRCh38
rs772954738 Chr.9: 136659426-136659426 on Build GRCh38
rs563901551 Chr.9: 136659553-136659561 on Build GRCh38
rs1036339153 Chr.9: 136659588-136659588 on Build GRCh38
rs528930913 Chr.9: 136659759-136659759 on Build GRCh38
rs983930884 Chr.9: 136659711-136659711 on Build GRCh38
rs888995868 Chr.9: 136659679-136659679 on Build GRCh38
rs918422847 Chr.9: 136659476-136659476 on Build GRCh38
rs532534114 Chr.9: 136659620-136659620 on Build GRCh38
rs4880062 Chr.9: 136659413-136659413 on Build GRCh38
rs559536210 Chr.9: 136659794-136659794 on Build GRCh38
rs181860913 Chr.9: 136659647-136659647 on Build GRCh38
rs542720945 Chr.9: 136659789-136659789 on Build GRCh38
rs965257448 Chr.9: 136659462-136659462 on Build GRCh38
rs1054300913 Chr.9: 136659584-136659584 on Build GRCh38
rs1019548295 Chr.9: 136659441-136659441 on Build GRCh38
rs958474305 Chr.9: 136659718-136659718 on Build GRCh38
rs1032422581 Chr.9: 136659433-136659433 on Build GRCh38
rs545315640 Chr.9: 136659641-136659641 on Build GRCh38
rs1019620392 Chr.9: 136659608-136659608 on Build GRCh38
rs897729337 Chr.9: 136659599-136659599 on Build GRCh38
rs949634153 Chr.9: 136659561-136659561 on Build GRCh38
rs1025506184 Chr.9: 136659660-136659660 on Build GRCh38
rs937005578 Chr.9: 136659549-136659549 on Build GRCh38
rs889840512 Chr.9: 136659589-136659589 on Build GRCh38
rs937486417 Chr.9: 136659880-136659880 on Build GRCh38
rs560154668 Chr.9: 136659473-136659473 on Build GRCh38
rs1030114905 Chr.9: 136659675-136659675 on Build GRCh38
rs988218415 Chr.9: 136659451-136659451 on Build GRCh38
rs1053925936 Chr.9: 136659818-136659818 on Build GRCh38
rs1055922071 Chr.9: 136659555-136659555 on Build GRCh38
rs889904773 Chr.9: 136659867-136659867 on Build GRCh38
rs543375744 Chr.9: 136659678-136659678 on Build GRCh38
rs537411695 Chr.9: 136659420-136659420 on Build GRCh38
rs761681886 Chr.9: 136659438-136659438 on Build GRCh38
rs887861410 Chr.9: 136659568-136659568 on Build GRCh38
rs191748377 Chr.9: 136659574-136659574 on Build GRCh38
rs916893738 Chr.9: 136659737-136659737 on Build GRCh38
rs781234995 Chr.9: 136659702-136659702 on Build GRCh38
rs927038588 Chr.9: 136659497-136659497 on Build GRCh38
rs928791802 Chr.9: 136659873-136659873 on Build GRCh38
rs937413497 Chr.9: 136659554-136659554 on Build GRCh38
rs942012633 Chr.9: 136659577-136659577 on Build GRCh38
rs967609234 Chr.9: 136659868-136659868 on Build GRCh38
rs1007016249 Chr.9: 136659559-136659568 on Build GRCh38
rs149735775 Chr.9: 136659649-136659649 on Build GRCh38
rs936839449 Chr.9: 136659790-136659790 on Build GRCh38
rs1042555744 Chr.9: 136659573-136659573 on Build GRCh38
rs187382798 Chr.9: 136659496-136659496 on Build GRCh38
rs553086600 Chr.9: 136659572-136659572 on Build GRCh38

View Full Product Details