Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137206140-137206609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDOR1
Entrez Gene ID
27158
Gene Name
NADPH dependent diflavin oxidoreductase 1
Gene Aliases
NR1, bA350O14.9
UniGene
Hs.512564
Gene Chromosome Location
Chr.9: 137205667-137219361 on Build GRCh38
Transcripts
NM_001144026.2 NM_001144027.2 NM_001144028.2 NM_014434.3 XM_011518545.1 XM_011518547.1 XM_011518548.2 XM_017014626.1 XM_017014627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6723559 c.184G>T p.G62C Chr.9: 137206280-137206280 on Build GRCh38
3088877 c.195delC p.P67fs*4 Chr.9: 137206291-137206291 on Build GRCh38
5875935 c.213+1G>A p.? Chr.9: 137206310-137206310 on Build GRCh38
4791938 c.207C>T p.N69N Chr.9: 137206303-137206303 on Build GRCh38
6336237 c.175G>A p.A59T Chr.9: 137206271-137206271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560715423 Chr.9: 137206356-137206356 on Build GRCh38
rs748075115 Chr.9: 137206563-137206563 on Build GRCh38
rs199689564 Chr.9: 137206342-137206342 on Build GRCh38
rs754851736 Chr.9: 137206508-137206510 on Build GRCh38
rs749079765 Chr.9: 137206210-137206211 on Build GRCh38
rs1041565213 Chr.9: 137206289-137206289 on Build GRCh38
rs993685331 Chr.9: 137206546-137206546 on Build GRCh38
rs962598244 Chr.9: 137206353-137206353 on Build GRCh38
rs756703032 Chr.9: 137206310-137206310 on Build GRCh38
rs188093771 Chr.9: 137206182-137206182 on Build GRCh38
rs1041660019 Chr.9: 137206466-137206466 on Build GRCh38
rs1034454144 Chr.9: 137206340-137206340 on Build GRCh38
rs914799703 Chr.9: 137206484-137206484 on Build GRCh38
rs774085897 Chr.9: 137206218-137206218 on Build GRCh38
rs562606432 Chr.9: 137206383-137206383 on Build GRCh38
rs773822752 Chr.9: 137206199-137206199 on Build GRCh38
rs562922746 Chr.9: 137206477-137206477 on Build GRCh38
rs762362689 Chr.9: 137206266-137206266 on Build GRCh38
rs947605926 Chr.9: 137206491-137206491 on Build GRCh38
rs780019624 Chr.9: 137206207-137206208 on Build GRCh38
rs771484336 Chr.9: 137206344-137206344 on Build GRCh38
rs558189420 Chr.9: 137206262-137206262 on Build GRCh38
rs180946708 Chr.9: 137206380-137206380 on Build GRCh38
rs1055663030 Chr.9: 137206502-137206502 on Build GRCh38
rs761346581 Chr.9: 137206214-137206214 on Build GRCh38
rs760447962 Chr.9: 137206251-137206251 on Build GRCh38
rs904334254 Chr.9: 137206473-137206473 on Build GRCh38
rs141333798 Chr.9: 137206449-137206449 on Build GRCh38
rs922326608 Chr.9: 137206461-137206461 on Build GRCh38
rs111492193 Chr.9: 137206529-137206529 on Build GRCh38
rs780263477 Chr.9: 137206316-137206316 on Build GRCh38
rs972294854 Chr.9: 137206360-137206360 on Build GRCh38
rs897826654 Chr.9: 137206545-137206545 on Build GRCh38
rs760903195 Chr.9: 137206260-137206262 on Build GRCh38
rs1036921719 Chr.9: 137206580-137206580 on Build GRCh38
rs901067414 Chr.9: 137206216-137206216 on Build GRCh38
rs898443992 Chr.9: 137206584-137206584 on Build GRCh38
rs554478709 Chr.9: 137206163-137206163 on Build GRCh38
rs768349517 Chr.9: 137206198-137206198 on Build GRCh38
rs914750750 Chr.9: 137206222-137206222 on Build GRCh38
rs373217113 Chr.9: 137206304-137206304 on Build GRCh38
rs80080498 Chr.9: 137206455-137206455 on Build GRCh38
rs534216045 Chr.9: 137206161-137206161 on Build GRCh38
rs928607916 Chr.9: 137206213-137206213 on Build GRCh38
rs568346522 Chr.9: 137206543-137206543 on Build GRCh38
rs758671394 Chr.9: 137206256-137206256 on Build GRCh38
rs757428660 Chr.9: 137206297-137206297 on Build GRCh38
rs761915946 Chr.9: 137206291-137206291 on Build GRCh38
rs770059679 Chr.9: 137206183-137206183 on Build GRCh38
rs546027868 Chr.9: 137206465-137206465 on Build GRCh38
rs552232177 Chr.9: 137206372-137206372 on Build GRCh38
rs538921704 Chr.9: 137206173-137206173 on Build GRCh38
rs375967521 Chr.9: 137206320-137206320 on Build GRCh38
rs575000867 Chr.9: 137206329-137206329 on Build GRCh38
rs781676709 Chr.9: 137206301-137206301 on Build GRCh38
rs771139355 Chr.9: 137206358-137206358 on Build GRCh38
rs1024335633 Chr.9: 137206347-137206347 on Build GRCh38
rs754053267 Chr.9: 137206454-137206454 on Build GRCh38
rs904364988 Chr.9: 137206300-137206300 on Build GRCh38
rs531708462 Chr.9: 137206505-137206505 on Build GRCh38
rs762584780 Chr.9: 137206185-137206185 on Build GRCh38
rs540133047 Chr.9: 137206429-137206429 on Build GRCh38
rs995320515 Chr.9: 137206386-137206386 on Build GRCh38
rs768532089 Chr.9: 137206209-137206210 on Build GRCh38
rs932375634 Chr.9: 137206338-137206338 on Build GRCh38
rs750338916 Chr.9: 137206247-137206247 on Build GRCh38
rs974129933 Chr.9: 137206194-137206194 on Build GRCh38
rs749508564 Chr.9: 137206205-137206205 on Build GRCh38
rs372634634 Chr.9: 137206448-137206448 on Build GRCh38
rs983805706 Chr.9: 137206398-137206398 on Build GRCh38
rs767390351 Chr.9: 137206220-137206220 on Build GRCh38
rs962487282 Chr.9: 137206165-137206165 on Build GRCh38
rs1004095692 Chr.9: 137206362-137206362 on Build GRCh38
rs1015427978 Chr.9: 137206377-137206377 on Build GRCh38
rs539660960 Chr.9: 137206223-137206223 on Build GRCh38
rs772841503 Chr.9: 137206346-137206346 on Build GRCh38
rs974962504 Chr.9: 137206493-137206493 on Build GRCh38
rs766079829 Chr.9: 137206255-137206255 on Build GRCh38
rs187380177 Chr.9: 137206552-137206552 on Build GRCh38
rs995809731 Chr.9: 137206587-137206587 on Build GRCh38
rs1027873594 Chr.9: 137206556-137206556 on Build GRCh38
rs370578677 Chr.9: 137206321-137206321 on Build GRCh38
rs771134409 Chr.9: 137206262-137206265 on Build GRCh38
rs952230674 Chr.9: 137206384-137206384 on Build GRCh38

View Full Product Details