Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137255095-137255583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELFB
Entrez Gene ID
25920
Gene Name
negative elongation factor complex member B
Gene Aliases
COBRA1, NELF-B
UniGene
Hs.521809
Gene Chromosome Location
Chr.9: 137255300-137273548 on Build GRCh38
Transcripts
NM_015456.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359816 c.38G>A p.G13D Chr.9: 137255547-137255547 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs934277710 Chr.9: 137255121-137255121 on Build GRCh38
rs529762203 Chr.9: 137255217-137255217 on Build GRCh38
rs1024284520 Chr.9: 137255361-137255361 on Build GRCh38
rs565069477 Chr.9: 137255265-137255265 on Build GRCh38
rs117798615 Chr.9: 137255215-137255215 on Build GRCh38
rs998035968 Chr.9: 137255307-137255307 on Build GRCh38
rs772916977 Chr.9: 137255248-137255248 on Build GRCh38
rs373996201 Chr.9: 137255284-137255284 on Build GRCh38
rs1017356640 Chr.9: 137255256-137255256 on Build GRCh38
rs991781704 Chr.9: 137255259-137255259 on Build GRCh38
rs903359939 Chr.9: 137255128-137255128 on Build GRCh38
rs1006084403 Chr.9: 137255351-137255351 on Build GRCh38
rs893430733 Chr.9: 137255195-137255195 on Build GRCh38
rs1053504373 Chr.9: 137255281-137255281 on Build GRCh38
rs933918787 Chr.9: 137255458-137255458 on Build GRCh38
rs764511248 Chr.9: 137255492-137255492 on Build GRCh38
rs1046204876 Chr.9: 137255143-137255143 on Build GRCh38
rs902248880 Chr.9: 137255151-137255151 on Build GRCh38
rs11555841 Chr.9: 137255503-137255503 on Build GRCh38
rs573000140 Chr.9: 137255257-137255257 on Build GRCh38
rs547021018 Chr.9: 137255432-137255432 on Build GRCh38
rs868644302 Chr.9: 137255214-137255214 on Build GRCh38
rs1025754986 Chr.9: 137255238-137255239 on Build GRCh38
rs942397524 Chr.9: 137255416-137255416 on Build GRCh38
rs1012910636 Chr.9: 137255359-137255359 on Build GRCh38
rs951566258 Chr.9: 137255240-137255240 on Build GRCh38
rs1047342105 Chr.9: 137255319-137255319 on Build GRCh38
rs530778613 Chr.9: 137255280-137255280 on Build GRCh38
rs901084102 Chr.9: 137255302-137255302 on Build GRCh38
rs886912476 Chr.9: 137255332-137255332 on Build GRCh38
rs113294775 Chr.9: 137255182-137255182 on Build GRCh38
rs577690992 Chr.9: 137255174-137255174 on Build GRCh38
rs937593141 Chr.9: 137255119-137255119 on Build GRCh38
rs1017019301 Chr.9: 137255352-137255352 on Build GRCh38
rs758634827 Chr.9: 137255120-137255120 on Build GRCh38
rs138595858 Chr.9: 137255117-137255117 on Build GRCh38
rs567052045 Chr.9: 137255469-137255469 on Build GRCh38
rs984184389 Chr.9: 137255252-137255252 on Build GRCh38
rs28367295 Chr.9: 137255301-137255301 on Build GRCh38
rs989674073 Chr.9: 137255395-137255395 on Build GRCh38
rs999269326 Chr.9: 137255129-137255129 on Build GRCh38
rs971656454 Chr.9: 137255362-137255362 on Build GRCh38
rs767505314 Chr.9: 137255222-137255222 on Build GRCh38
rs1003098043 Chr.9: 137255221-137255221 on Build GRCh38
rs561854930 Chr.9: 137255130-137255131 on Build GRCh38
rs957550976 Chr.9: 137255394-137255394 on Build GRCh38
rs573881172 Chr.9: 137255211-137255211 on Build GRCh38
rs537079789 Chr.9: 137255175-137255175 on Build GRCh38
rs546268675 Chr.9: 137255486-137255486 on Build GRCh38
rs977629849 Chr.9: 137255385-137255385 on Build GRCh38
rs935064554 Chr.9: 137255163-137255163 on Build GRCh38
rs924778997 Chr.9: 137255387-137255387 on Build GRCh38
rs922232593 Chr.9: 137255445-137255445 on Build GRCh38
rs958720326 Chr.9: 137255356-137255356 on Build GRCh38
rs914979509 Chr.9: 137255397-137255397 on Build GRCh38
rs924778595 Chr.9: 137255272-137255272 on Build GRCh38
rs557932724 Chr.9: 137255127-137255127 on Build GRCh38
rs1030795912 Chr.9: 137255132-137255132 on Build GRCh38
rs557259359 Chr.9: 137255205-137255205 on Build GRCh38
rs942367871 Chr.9: 137255286-137255286 on Build GRCh38
rs751199805 Chr.9: 137255531-137255531 on Build GRCh38
rs935076525 Chr.9: 137255279-137255279 on Build GRCh38
rs199681335 Chr.9: 137255551-137255551 on Build GRCh38
rs545075027 Chr.9: 137255263-137255263 on Build GRCh38
rs559489844 Chr.9: 137255253-137255253 on Build GRCh38
rs186246889 Chr.9: 137255417-137255417 on Build GRCh38

View Full Product Details