Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127663116-127663643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STXBP1
Entrez Gene ID
6812
Gene Name
syntaxin binding protein 1
Gene Aliases
MUNC18-1, NSEC1, P67, RBSEC1, UNC18
UniGene
Hs.288229
Gene Chromosome Location
Chr.9: 127612207-127692716 on Build GRCh38
Transcripts
NM_001032221.3 NM_003165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6792683 c.578+1G>A p.? Chr.9: 127663354-127663354 on Build GRCh38
1105351 c.574C>T p.R192W Chr.9: 127663349-127663349 on Build GRCh38
7100522 c.547C>T p.L183L Chr.9: 127663322-127663322 on Build GRCh38
7079627 c.560C>A p.P187Q Chr.9: 127663335-127663335 on Build GRCh38
5977086 c.471C>T p.Y157Y Chr.9: 127663246-127663246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796053354 Chr.9: 127663204-127663204 on Build GRCh38
rs893036313 Chr.9: 127663595-127663595 on Build GRCh38
rs115329486 Chr.9: 127663544-127663544 on Build GRCh38
rs761390809 Chr.9: 127663279-127663279 on Build GRCh38
rs772316151 Chr.9: 127663460-127663460 on Build GRCh38
rs752943575 Chr.9: 127663351-127663351 on Build GRCh38
rs1013745084 Chr.9: 127663194-127663194 on Build GRCh38
rs769643462 Chr.9: 127663395-127663395 on Build GRCh38
rs201797980 Chr.9: 127663364-127663364 on Build GRCh38
rs759620661 Chr.9: 127663327-127663327 on Build GRCh38
rs376885363 Chr.9: 127663380-127663380 on Build GRCh38
rs950524734 Chr.9: 127663551-127663551 on Build GRCh38
rs771494216 Chr.9: 127663169-127663169 on Build GRCh38
rs542227827 Chr.9: 127663609-127663609 on Build GRCh38
rs930913126 Chr.9: 127663410-127663410 on Build GRCh38
rs796053356 Chr.9: 127663344-127663344 on Build GRCh38
rs142317249 Chr.9: 127663316-127663316 on Build GRCh38
rs796053355 Chr.9: 127663343-127663343 on Build GRCh38
rs587784455 Chr.9: 127663208-127663208 on Build GRCh38
rs1055180388 Chr.9: 127663143-127663143 on Build GRCh38
rs543685843 Chr.9: 127663393-127663393 on Build GRCh38
rs957598940 Chr.9: 127663430-127663430 on Build GRCh38
rs747481676 Chr.9: 127663168-127663168 on Build GRCh38
rs976532481 Chr.9: 127663519-127663519 on Build GRCh38
rs554886945 Chr.9: 127663201-127663201 on Build GRCh38
rs896193048 Chr.9: 127663144-127663144 on Build GRCh38
rs765256766 Chr.9: 127663347-127663347 on Build GRCh38
rs903312718 Chr.9: 127663221-127663221 on Build GRCh38
rs1033234416 Chr.9: 127663426-127663426 on Build GRCh38
rs201772971 Chr.9: 127663286-127663286 on Build GRCh38
rs972703931 Chr.9: 127663336-127663336 on Build GRCh38
rs557556080 Chr.9: 127663429-127663429 on Build GRCh38
rs981193334 Chr.9: 127663198-127663198 on Build GRCh38
rs7036307 Chr.9: 127663498-127663498 on Build GRCh38
rs368895797 Chr.9: 127663390-127663390 on Build GRCh38
rs773211322 Chr.9: 127663287-127663287 on Build GRCh38
rs758658621 Chr.9: 127663389-127663389 on Build GRCh38
rs753725349 Chr.9: 127663322-127663322 on Build GRCh38
rs779416499 Chr.9: 127663184-127663184 on Build GRCh38
rs748770943 Chr.9: 127663196-127663196 on Build GRCh38
rs760681174 Chr.9: 127663303-127663303 on Build GRCh38
rs187928936 Chr.9: 127663577-127663577 on Build GRCh38
rs928103989 Chr.9: 127663278-127663278 on Build GRCh38
rs794727859 Chr.9: 127663323-127663323 on Build GRCh38
rs17849178 Chr.9: 127663431-127663431 on Build GRCh38
rs796053382 Chr.9: 127663354-127663355 on Build GRCh38
rs796053357 Chr.9: 127663354-127663354 on Build GRCh38
rs1011947525 Chr.9: 127663604-127663604 on Build GRCh38
rs757724563 Chr.9: 127663583-127663583 on Build GRCh38
rs768052332 Chr.9: 127663237-127663237 on Build GRCh38
rs774013162 Chr.9: 127663252-127663252 on Build GRCh38
rs111983551 Chr.9: 127663474-127663474 on Build GRCh38
rs371232802 Chr.9: 127663167-127663167 on Build GRCh38
rs901652553 Chr.9: 127663510-127663510 on Build GRCh38
rs121918318 Chr.9: 127663314-127663314 on Build GRCh38

View Full Product Details