Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127742661-127743061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2D3C
Entrez Gene ID
10044
Gene Name
SH2 domain containing 3C
Gene Aliases
CHAT, NSP3, PRO34088, SHEP1
UniGene
Hs.306412
Gene Chromosome Location
Chr.9: 127738317-127778769 on Build GRCh38
Transcripts
NM_001142531.1 NM_001142532.1 NM_001142533.1 NM_001142534.1 NM_001252334.1 NM_005489.3 NM_170600.2 XM_005251639.1 XM_011518114.1 XM_011518115.1 XM_011518117.2 XM_017014174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6048021 c.1601C>A p.A534D Chr.9: 127742960-127742960 on Build GRCh38
6777017 c.1886G>A p.G629D Chr.9: 127742879-127742879 on Build GRCh38
6048020 c.1334C>A p.A445D Chr.9: 127742960-127742960 on Build GRCh38
4786013 c.1345G>T p.G449C Chr.9: 127742949-127742949 on Build GRCh38
5774530 c.1330-10G>A p.? Chr.9: 127742974-127742974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751806041 Chr.9: 127742883-127742883 on Build GRCh38
rs1047279080 Chr.9: 127742908-127742908 on Build GRCh38
rs758608953 Chr.9: 127742874-127742874 on Build GRCh38
rs762687185 Chr.9: 127742841-127742842 on Build GRCh38
rs780540521 Chr.9: 127743000-127743000 on Build GRCh38
rs756677326 Chr.9: 127742996-127742996 on Build GRCh38
rs778196058 Chr.9: 127742875-127742875 on Build GRCh38
rs568246379 Chr.9: 127742835-127742835 on Build GRCh38
rs771555665 Chr.9: 127742931-127742931 on Build GRCh38
rs374235995 Chr.9: 127742907-127742907 on Build GRCh38
rs773955599 Chr.9: 127742808-127742808 on Build GRCh38
rs746408761 Chr.9: 127742905-127742905 on Build GRCh38
rs867479645 Chr.9: 127742978-127742978 on Build GRCh38
rs375088470 Chr.9: 127743001-127743001 on Build GRCh38
rs764165148 Chr.9: 127742980-127742980 on Build GRCh38
rs750910877 Chr.9: 127742993-127742993 on Build GRCh38
rs766167327 Chr.9: 127742831-127742831 on Build GRCh38
rs372223305 Chr.9: 127742982-127742982 on Build GRCh38
rs751040543 Chr.9: 127742691-127742699 on Build GRCh38
rs900865439 Chr.9: 127742843-127742843 on Build GRCh38
rs573022434 Chr.9: 127742833-127742833 on Build GRCh38
rs759648478 Chr.9: 127742837-127742837 on Build GRCh38
rs528384536 Chr.9: 127742785-127742785 on Build GRCh38
rs781631366 Chr.9: 127742904-127742904 on Build GRCh38
rs936526282 Chr.9: 127742749-127742749 on Build GRCh38
rs772944188 Chr.9: 127742812-127742812 on Build GRCh38
rs960286239 Chr.9: 127742784-127742784 on Build GRCh38
rs772199643 Chr.9: 127743014-127743024 on Build GRCh38
rs781579976 Chr.9: 127742992-127742992 on Build GRCh38
rs1053708305 Chr.9: 127742789-127742789 on Build GRCh38
rs748742691 Chr.9: 127742799-127742799 on Build GRCh38
rs376803492 Chr.9: 127742984-127742984 on Build GRCh38
rs550688273 Chr.9: 127742952-127742952 on Build GRCh38
rs761415023 Chr.9: 127742810-127742810 on Build GRCh38
rs757131273 Chr.9: 127742760-127742760 on Build GRCh38
rs923786640 Chr.9: 127742743-127742743 on Build GRCh38
rs191866782 Chr.9: 127742953-127742953 on Build GRCh38
rs777250448 Chr.9: 127743013-127743013 on Build GRCh38
rs376987652 Chr.9: 127742974-127742974 on Build GRCh38
rs149793930 Chr.9: 127742801-127742801 on Build GRCh38
rs753849177 Chr.9: 127742836-127742836 on Build GRCh38
rs145507182 Chr.9: 127742950-127742950 on Build GRCh38
rs1011336267 Chr.9: 127742776-127742776 on Build GRCh38
rs536814228 Chr.9: 127742913-127742913 on Build GRCh38
rs140246171 Chr.9: 127742933-127742933 on Build GRCh38
rs752942885 Chr.9: 127742860-127742860 on Build GRCh38
rs1034641656 Chr.9: 127742731-127742731 on Build GRCh38
rs372070605 Chr.9: 127742967-127742967 on Build GRCh38
rs757655035 Chr.9: 127742884-127742884 on Build GRCh38
rs752787886 Chr.9: 127742972-127742972 on Build GRCh38
rs992787862 Chr.9: 127742795-127742795 on Build GRCh38
rs771593283 Chr.9: 127742896-127742896 on Build GRCh38
rs149694765 Chr.9: 127742944-127742944 on Build GRCh38
rs978597149 Chr.9: 127742742-127742742 on Build GRCh38
rs772926498 Chr.9: 127742943-127742943 on Build GRCh38
rs779621656 Chr.9: 127742914-127742914 on Build GRCh38
rs771630178 Chr.9: 127743002-127743002 on Build GRCh38
rs957890601 Chr.9: 127743019-127743019 on Build GRCh38
rs933930314 Chr.9: 127742989-127742990 on Build GRCh38
rs765447172 Chr.9: 127742838-127742838 on Build GRCh38
rs372595836 Chr.9: 127742726-127742726 on Build GRCh38
rs759593468 Chr.9: 127742956-127742956 on Build GRCh38
rs747830158 Chr.9: 127742930-127742930 on Build GRCh38
rs911173350 Chr.9: 127742792-127742792 on Build GRCh38
rs760519427 Chr.9: 127742830-127742830 on Build GRCh38
rs539094926 Chr.9: 127743016-127743016 on Build GRCh38

View Full Product Details