Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127916642-127917132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GALNAC4
Entrez Gene ID
27090
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Gene Aliases
IV, SIAT3-C, SIAT3C, SIAT7-D, SIAT7D, ST6GALNACIV, ST6GalNAc
UniGene
Hs.3972
Gene Chromosome Location
Chr.9: 127907886-127917052 on Build GRCh38
Transcripts
NM_175039.3 NM_175040.3 XM_011518527.2 XM_011518529.2

SNP Detail

SNP ID Chromosome Location
rs1021125413 Chr.9: 127916678-127916678 on Build GRCh38
rs949958303 Chr.9: 127917042-127917043 on Build GRCh38
rs1001420770 Chr.9: 127916985-127916985 on Build GRCh38
rs958750783 Chr.9: 127917020-127917020 on Build GRCh38
rs1048346478 Chr.9: 127916911-127916911 on Build GRCh38
rs931133620 Chr.9: 127916900-127916900 on Build GRCh38
rs988943458 Chr.9: 127916671-127916671 on Build GRCh38
rs1046730287 Chr.9: 127916903-127916903 on Build GRCh38
rs918664884 Chr.9: 127917026-127917026 on Build GRCh38
rs967245693 Chr.9: 127916679-127916679 on Build GRCh38
rs941007429 Chr.9: 127917061-127917061 on Build GRCh38
rs1038526938 Chr.9: 127916975-127916975 on Build GRCh38
rs944146701 Chr.9: 127916803-127916803 on Build GRCh38
rs1029093814 Chr.9: 127916986-127916986 on Build GRCh38
rs1001230099 Chr.9: 127916925-127916925 on Build GRCh38
rs925845729 Chr.9: 127916796-127916796 on Build GRCh38
rs531187884 Chr.9: 127917085-127917085 on Build GRCh38
rs967494068 Chr.9: 127916755-127916755 on Build GRCh38
rs937140909 Chr.9: 127916843-127916843 on Build GRCh38
rs533478628 Chr.9: 127916751-127916751 on Build GRCh38
rs937243915 Chr.9: 127917059-127917059 on Build GRCh38
rs966942593 Chr.9: 127916984-127916984 on Build GRCh38
rs954304765 Chr.9: 127917002-127917002 on Build GRCh38
rs965726753 Chr.9: 127917017-127917017 on Build GRCh38
rs1037487385 Chr.9: 127916832-127916832 on Build GRCh38
rs569258132 Chr.9: 127917018-127917018 on Build GRCh38
rs1009995409 Chr.9: 127916964-127916964 on Build GRCh38
rs985411210 Chr.9: 127916748-127916748 on Build GRCh38
rs1029187003 Chr.9: 127916669-127916669 on Build GRCh38
rs1057038795 Chr.9: 127917067-127917067 on Build GRCh38
rs949926743 Chr.9: 127916898-127916898 on Build GRCh38
rs770551180 Chr.9: 127916788-127916789 on Build GRCh38
rs905761595 Chr.9: 127916914-127916914 on Build GRCh38
rs772537155 Chr.9: 127916788-127916790 on Build GRCh38
rs1047417378 Chr.9: 127917051-127917051 on Build GRCh38
rs868019637 Chr.9: 127916977-127916977 on Build GRCh38
rs979742213 Chr.9: 127916713-127916713 on Build GRCh38
rs912567948 Chr.9: 127917011-127917011 on Build GRCh38
rs917186890 Chr.9: 127916892-127916892 on Build GRCh38
rs149075317 Chr.9: 127916789-127916791 on Build GRCh38
rs925533975 Chr.9: 127916734-127916734 on Build GRCh38
rs931186462 Chr.9: 127917027-127917027 on Build GRCh38
rs954929172 Chr.9: 127916688-127916688 on Build GRCh38
rs899608904 Chr.9: 127917112-127917112 on Build GRCh38
rs367966654 Chr.9: 127916740-127916740 on Build GRCh38
rs1041489959 Chr.9: 127917115-127917115 on Build GRCh38
rs972878396 Chr.9: 127917025-127917025 on Build GRCh38
rs1055751660 Chr.9: 127916859-127916859 on Build GRCh38
rs866034227 Chr.9: 127916733-127916733 on Build GRCh38
rs896996603 Chr.9: 127916836-127916836 on Build GRCh38
rs371922201 Chr.9: 127917033-127917033 on Build GRCh38
rs183664914 Chr.9: 127916991-127916991 on Build GRCh38
rs985651620 Chr.9: 127917004-127917004 on Build GRCh38
rs1010314562 Chr.9: 127917104-127917104 on Build GRCh38
rs551669393 Chr.9: 127917040-127917043 on Build GRCh38
rs560525251 Chr.9: 127916930-127916930 on Build GRCh38
rs895444509 Chr.9: 127916952-127916952 on Build GRCh38
rs149350989 Chr.9: 127916969-127916969 on Build GRCh38
rs1035369184 Chr.9: 127916950-127916950 on Build GRCh38
rs927083622 Chr.9: 127917054-127917054 on Build GRCh38
rs540796247 Chr.9: 127916753-127916753 on Build GRCh38

View Full Product Details