Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128842711-128843219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KYAT1
Entrez Gene ID
883
Gene Name
kynurenine aminotransferase 1
Gene Aliases
CCBL1, GTK, KAT1, KATI
UniGene
Hs.495250
Gene Chromosome Location
Chr.9: 128832942-128882551 on Build GRCh38
Transcripts
NM_001122671.1 NM_001122672.1 NM_001287390.1 NM_004059.4 NR_109829.1 XM_006717311.3 XM_011519165.2 XM_011519167.2 XM_011519168.1 XM_011519169.1 XM_011519170.2 XM_017015260.1 XM_017015261.1 XM_017015262.1 XM_017015263.1 XM_017015264.1 XM_017015265.1 XM_017015266.1 XM_017015267.1 XM_017015268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2155057 c.129A>T p.P43P Chr.9: 128842729-128842729 on Build GRCh38
1187834 c.130G>T p.D44Y Chr.9: 128842725-128842725 on Build GRCh38
2155055 c.126A>G p.P42P Chr.9: 128842732-128842732 on Build GRCh38
3654748 c.112C>T p.P38S Chr.9: 128842743-128842743 on Build GRCh38
3654749 c.394C>T p.P132S Chr.9: 128842743-128842743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019095389 Chr.9: 128843115-128843115 on Build GRCh38
rs1000411635 Chr.9: 128843098-128843098 on Build GRCh38
rs116499756 Chr.9: 128843123-128843123 on Build GRCh38
rs985200925 Chr.9: 128842880-128842880 on Build GRCh38
rs1041733731 Chr.9: 128843091-128843091 on Build GRCh38
rs774585438 Chr.9: 128842822-128842822 on Build GRCh38
rs779868766 Chr.9: 128843103-128843103 on Build GRCh38
rs376709202 Chr.9: 128842762-128842762 on Build GRCh38
rs75664372 Chr.9: 128843029-128843029 on Build GRCh38
rs1018453532 Chr.9: 128842884-128842884 on Build GRCh38
rs903190413 Chr.9: 128843092-128843092 on Build GRCh38
rs768567642 Chr.9: 128842807-128842807 on Build GRCh38
rs377087687 Chr.9: 128842741-128842741 on Build GRCh38
rs567333637 Chr.9: 128842953-128842953 on Build GRCh38
rs745716290 Chr.9: 128842773-128842773 on Build GRCh38
rs984026432 Chr.9: 128843184-128843184 on Build GRCh38
rs1000896186 Chr.9: 128842782-128842782 on Build GRCh38
rs762135780 Chr.9: 128842754-128842754 on Build GRCh38
rs940612791 Chr.9: 128843034-128843034 on Build GRCh38
rs1033146191 Chr.9: 128842858-128842858 on Build GRCh38
rs772771773 Chr.9: 128842837-128842837 on Build GRCh38
rs953144804 Chr.9: 128842985-128842985 on Build GRCh38
rs1019779536 Chr.9: 128843169-128843181 on Build GRCh38
rs116107448 Chr.9: 128843148-128843148 on Build GRCh38
rs1055663124 Chr.9: 128843004-128843004 on Build GRCh38
rs965790616 Chr.9: 128842885-128842885 on Build GRCh38
rs34135861 Chr.9: 128842889-128842890 on Build GRCh38
rs748598491 Chr.9: 128842835-128842835 on Build GRCh38
rs547700261 Chr.9: 128842942-128842942 on Build GRCh38
rs1033695803 Chr.9: 128843099-128843099 on Build GRCh38
rs754398868 Chr.9: 128843089-128843089 on Build GRCh38
rs374906693 Chr.9: 128842765-128842765 on Build GRCh38
rs976369218 Chr.9: 128842912-128842912 on Build GRCh38
rs534556491 Chr.9: 128842779-128842779 on Build GRCh38
rs368132598 Chr.9: 128842767-128842767 on Build GRCh38
rs533865870 Chr.9: 128842933-128842933 on Build GRCh38
rs909777458 Chr.9: 128843192-128843192 on Build GRCh38
rs1009736020 Chr.9: 128842863-128842863 on Build GRCh38
rs780965388 Chr.9: 128842769-128842769 on Build GRCh38
rs773715008 Chr.9: 128842745-128842745 on Build GRCh38
rs965951385 Chr.9: 128843175-128843175 on Build GRCh38
rs1045536310 Chr.9: 128843102-128843102 on Build GRCh38
rs780176481 Chr.9: 128842785-128842785 on Build GRCh38
rs937184370 Chr.9: 128842988-128842988 on Build GRCh38
rs965128400 Chr.9: 128842890-128842890 on Build GRCh38
rs760985994 Chr.9: 128842842-128842842 on Build GRCh38
rs370439886 Chr.9: 128842742-128842742 on Build GRCh38
rs766798566 Chr.9: 128842853-128842853 on Build GRCh38
rs1008128099 Chr.9: 128843104-128843104 on Build GRCh38
rs1036118811 Chr.9: 128843038-128843038 on Build GRCh38
rs767848849 Chr.9: 128842859-128842859 on Build GRCh38
rs187561599 Chr.9: 128842916-128842916 on Build GRCh38
rs773888313 Chr.9: 128843176-128843176 on Build GRCh38
rs749553282 Chr.9: 128842788-128842788 on Build GRCh38
rs759213787 Chr.9: 128842746-128842746 on Build GRCh38
rs756334519 Chr.9: 128842776-128842776 on Build GRCh38
rs892719567 Chr.9: 128843131-128843131 on Build GRCh38
rs144409793 Chr.9: 128843122-128843122 on Build GRCh38
rs757193921 Chr.9: 128842768-128842768 on Build GRCh38
rs984452135 Chr.9: 128843019-128843019 on Build GRCh38
rs551408131 Chr.9: 128842764-128842764 on Build GRCh38

View Full Product Details