Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49925090-49925601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN5
Entrez Gene ID
1184
Gene Name
chloride voltage-gated channel 5
Gene Aliases
CLC5, CLCK2, ClC-5, DENTS, NPHL1, NPHL2, XLRH, XRN, hCIC-K2
UniGene
Hs.166486
Gene Chromosome Location
Chr.X: 49922615-50099235 on Build GRCh38
Transcripts
NM_001127898.3 NM_001127899.3 NM_001272102.1 XM_017029257.1 XM_017029258.1

SNP Detail

SNP ID Chromosome Location
rs997957161 Chr.X: 49925443-49925443 on Build GRCh38
rs782707533 Chr.X: 49925361-49925361 on Build GRCh38
rs782073023 Chr.X: 49925305-49925305 on Build GRCh38
rs782294213 Chr.X: 49925277-49925277 on Build GRCh38
rs782360115 Chr.X: 49925293-49925293 on Build GRCh38
rs376276091 Chr.X: 49925284-49925284 on Build GRCh38
rs185140502 Chr.X: 49925235-49925235 on Build GRCh38
rs782781111 Chr.X: 49925276-49925276 on Build GRCh38
rs902152211 Chr.X: 49925409-49925409 on Build GRCh38
rs782798617 Chr.X: 49925309-49925309 on Build GRCh38
rs782553797 Chr.X: 49925490-49925490 on Build GRCh38
rs782125591 Chr.X: 49925324-49925324 on Build GRCh38
rs373283389 Chr.X: 49925278-49925278 on Build GRCh38
rs781923871 Chr.X: 49925295-49925295 on Build GRCh38
rs782674538 Chr.X: 49925254-49925254 on Build GRCh38
rs782216208 Chr.X: 49925292-49925292 on Build GRCh38
rs781883996 Chr.X: 49925362-49925362 on Build GRCh38
rs1029791446 Chr.X: 49925455-49925455 on Build GRCh38
rs200775713 Chr.X: 49925316-49925316 on Build GRCh38
rs782266141 Chr.X: 49925377-49925377 on Build GRCh38
rs183192020 Chr.X: 49925486-49925486 on Build GRCh38
rs781838070 Chr.X: 49925345-49925345 on Build GRCh38
rs782308486 Chr.X: 49925333-49925333 on Build GRCh38
rs782032171 Chr.X: 49925510-49925510 on Build GRCh38
rs1016840546 Chr.X: 49925558-49925558 on Build GRCh38
rs928169825 Chr.X: 49925229-49925229 on Build GRCh38
rs373374676 Chr.X: 49925561-49925561 on Build GRCh38
rs191071006 Chr.X: 49925360-49925360 on Build GRCh38

View Full Product Details