Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50072382-50072929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN5
Entrez Gene ID
1184
Gene Name
chloride voltage-gated channel 5
Gene Aliases
CLC5, CLCK2, ClC-5, DENTS, NPHL1, NPHL2, XLRH, XRN, hCIC-K2
UniGene
Hs.166486
Gene Chromosome Location
Chr.X: 49922615-50099235 on Build GRCh38
Transcripts
NM_000084.4 NM_001127898.3 NM_001127899.3 NM_001282163.1 XM_017029257.1 XM_017029258.1

SNP Detail

SNP ID Chromosome Location
rs781813378 Chr.X: 50072600-50072600 on Build GRCh38
rs782311071 Chr.X: 50072453-50072453 on Build GRCh38
rs782499966 Chr.X: 50072578-50072578 on Build GRCh38
rs782535203 Chr.X: 50072631-50072631 on Build GRCh38
rs782188853 Chr.X: 50072622-50072622 on Build GRCh38
rs781876648 Chr.X: 50072893-50072893 on Build GRCh38
rs978371514 Chr.X: 50072762-50072762 on Build GRCh38
rs782057466 Chr.X: 50072551-50072551 on Build GRCh38
rs151340629 Chr.X: 50072553-50072553 on Build GRCh38
rs781953806 Chr.X: 50072719-50072719 on Build GRCh38
rs782773735 Chr.X: 50072594-50072594 on Build GRCh38
rs781988625 Chr.X: 50072496-50072496 on Build GRCh38
rs376833330 Chr.X: 50072504-50072504 on Build GRCh38
rs782744969 Chr.X: 50072515-50072515 on Build GRCh38
rs971256503 Chr.X: 50072499-50072499 on Build GRCh38
rs924229473 Chr.X: 50072843-50072843 on Build GRCh38
rs782578388 Chr.X: 50072621-50072621 on Build GRCh38
rs191615487 Chr.X: 50072902-50072902 on Build GRCh38
rs782366642 Chr.X: 50072667-50072667 on Build GRCh38
rs112785600 Chr.X: 50072805-50072805 on Build GRCh38
rs782426163 Chr.X: 50072475-50072475 on Build GRCh38
rs781973295 Chr.X: 50072534-50072534 on Build GRCh38
rs781902929 Chr.X: 50072574-50072574 on Build GRCh38
rs990757836 Chr.X: 50072668-50072668 on Build GRCh38
rs782226989 Chr.X: 50072447-50072447 on Build GRCh38
rs56117808 Chr.X: 50072472-50072472 on Build GRCh38
rs1035055157 Chr.X: 50072665-50072665 on Build GRCh38
rs782478481 Chr.X: 50072614-50072614 on Build GRCh38
rs782279035 Chr.X: 50072474-50072474 on Build GRCh38
rs782720805 Chr.X: 50072552-50072552 on Build GRCh38
rs781819779 Chr.X: 50072704-50072704 on Build GRCh38
rs968239010 Chr.X: 50072756-50072756 on Build GRCh38

View Full Product Details