Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53234156-53234647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQSEC2
Entrez Gene ID
23096
Gene Name
IQ motif and Sec7 domain 2
Gene Aliases
BRAG1, MRX1, MRX78
UniGene
Hs.496138
Gene Chromosome Location
Chr.X: 53225784-53321350 on Build GRCh38
Transcripts
NM_001111125.2 NM_015075.1 XM_006724579.2 XM_006724580.3 XM_006724581.3 XM_006724582.3 XM_006724583.3 XM_006724584.2 XM_011530773.2 XM_017029359.1 XM_017029360.1

SNP Detail

SNP ID Chromosome Location
rs191886831 Chr.X: 53234278-53234278 on Build GRCh38
rs184087864 Chr.X: 53234582-53234582 on Build GRCh38
rs782285127 Chr.X: 53234613-53234613 on Build GRCh38
rs781858960 Chr.X: 53234299-53234299 on Build GRCh38
rs1024432774 Chr.X: 53234546-53234546 on Build GRCh38
rs797045631 Chr.X: 53234270-53234270 on Build GRCh38
rs782585446 Chr.X: 53234323-53234323 on Build GRCh38
rs782586963 Chr.X: 53234600-53234600 on Build GRCh38
rs1013759952 Chr.X: 53234473-53234473 on Build GRCh38
rs782796942 Chr.X: 53234481-53234481 on Build GRCh38
rs782352923 Chr.X: 53234320-53234320 on Build GRCh38
rs587780360 Chr.X: 53234372-53234372 on Build GRCh38
rs781846571 Chr.X: 53234573-53234573 on Build GRCh38
rs797044650 Chr.X: 53234360-53234360 on Build GRCh38
rs1006462031 Chr.X: 53234176-53234176 on Build GRCh38
rs782729294 Chr.X: 53234432-53234432 on Build GRCh38
rs782381980 Chr.X: 53234331-53234331 on Build GRCh38
rs897962017 Chr.X: 53234300-53234300 on Build GRCh38
rs782536136 Chr.X: 53234393-53234393 on Build GRCh38
rs782252624 Chr.X: 53234327-53234327 on Build GRCh38

View Full Product Details