Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54071782-54072326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120C
Entrez Gene ID
54954
Gene Name
family with sequence similarity 120C
Gene Aliases
CXorf17, ORF34
UniGene
Hs.86045
Gene Chromosome Location
Chr.X: 54068324-54183281 on Build GRCh38
Transcripts
NM_001300788.1 NM_017848.5

SNP Detail

SNP ID Chromosome Location
rs200850761 Chr.X: 54071966-54071966 on Build GRCh38
rs782479836 Chr.X: 54071995-54071995 on Build GRCh38
rs915782786 Chr.X: 54072058-54072058 on Build GRCh38
rs782477180 Chr.X: 54072182-54072183 on Build GRCh38
rs60802365 Chr.X: 54072201-54072202 on Build GRCh38
rs782565587 Chr.X: 54072220-54072220 on Build GRCh38
rs781990334 Chr.X: 54071835-54071835 on Build GRCh38
rs1009045327 Chr.X: 54071859-54071859 on Build GRCh38
rs1027591046 Chr.X: 54071986-54071986 on Build GRCh38
rs973663619 Chr.X: 54071976-54071976 on Build GRCh38
rs782204784 Chr.X: 54072159-54072159 on Build GRCh38
rs782505230 Chr.X: 54072151-54072151 on Build GRCh38
rs781847884 Chr.X: 54072013-54072013 on Build GRCh38
rs201493547 Chr.X: 54071970-54071970 on Build GRCh38
rs950776242 Chr.X: 54071990-54071990 on Build GRCh38
rs782063882 Chr.X: 54072159-54072160 on Build GRCh38
rs782270918 Chr.X: 54071871-54071871 on Build GRCh38
rs928046039 Chr.X: 54071996-54071996 on Build GRCh38
rs948615969 Chr.X: 54072074-54072074 on Build GRCh38
rs782638457 Chr.X: 54071822-54071822 on Build GRCh38
rs782128038 Chr.X: 54071928-54071928 on Build GRCh38
rs781901160 Chr.X: 54072150-54072150 on Build GRCh38
rs782286355 Chr.X: 54072019-54072019 on Build GRCh38
rs782637779 Chr.X: 54072158-54072158 on Build GRCh38

View Full Product Details