Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54080142-54080581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120C
Entrez Gene ID
54954
Gene Name
family with sequence similarity 120C
Gene Aliases
CXorf17, ORF34
UniGene
Hs.86045
Gene Chromosome Location
Chr.X: 54068324-54183281 on Build GRCh38
Transcripts
NM_001300788.1 NM_017848.5

SNP Detail

SNP ID Chromosome Location
rs781953112 Chr.X: 54080518-54080518 on Build GRCh38
rs191604304 Chr.X: 54080505-54080505 on Build GRCh38
rs782015082 Chr.X: 54080269-54080269 on Build GRCh38
rs112858622 Chr.X: 54080363-54080363 on Build GRCh38
rs781959649 Chr.X: 54080334-54080334 on Build GRCh38
rs782694420 Chr.X: 54080205-54080205 on Build GRCh38
rs782224012 Chr.X: 54080199-54080199 on Build GRCh38
rs2879887 Chr.X: 54080185-54080185 on Build GRCh38
rs973931872 Chr.X: 54080420-54080420 on Build GRCh38
rs782123804 Chr.X: 54080308-54080308 on Build GRCh38
rs920700249 Chr.X: 54080459-54080459 on Build GRCh38
rs782181996 Chr.X: 54080242-54080242 on Build GRCh38
rs782471922 Chr.X: 54080217-54080217 on Build GRCh38
rs782070139 Chr.X: 54080339-54080339 on Build GRCh38
rs781929390 Chr.X: 54080216-54080216 on Build GRCh38
rs782246278 Chr.X: 54080323-54080323 on Build GRCh38
rs781845932 Chr.X: 54080216-54080217 on Build GRCh38
rs782414547 Chr.X: 54080260-54080260 on Build GRCh38
rs964804802 Chr.X: 54080364-54080364 on Build GRCh38
rs782777937 Chr.X: 54080293-54080294 on Build GRCh38

View Full Product Details