Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54749949-54750449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH6
Entrez Gene ID
347365
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 6
Gene Aliases
ITIH5L, UNQ6369, dJ14O9.1
UniGene
Hs.454272
Gene Chromosome Location
Chr.X: 54748899-54798240 on Build GRCh38
Transcripts
NM_198510.2

SNP Detail

SNP ID Chromosome Location
rs1031977817 Chr.X: 54750383-54750383 on Build GRCh38
rs374457034 Chr.X: 54749990-54749990 on Build GRCh38
rs140483013 Chr.X: 54750093-54750093 on Build GRCh38
rs762866462 Chr.X: 54750135-54750135 on Build GRCh38
rs762777909 Chr.X: 54750095-54750095 on Build GRCh38
rs143515399 Chr.X: 54750046-54750046 on Build GRCh38
rs370169907 Chr.X: 54749981-54749981 on Build GRCh38
rs1001862806 Chr.X: 54750365-54750365 on Build GRCh38
rs764451977 Chr.X: 54750384-54750384 on Build GRCh38
rs751387919 Chr.X: 54750123-54750123 on Build GRCh38
rs754709509 Chr.X: 54750130-54750130 on Build GRCh38
rs145541040 Chr.X: 54750080-54750080 on Build GRCh38
rs769174424 Chr.X: 54750045-54750045 on Build GRCh38
rs752287196 Chr.X: 54750139-54750139 on Build GRCh38
rs879205744 Chr.X: 54750233-54750233 on Build GRCh38
rs761003584 Chr.X: 54750000-54750002 on Build GRCh38
rs200661984 Chr.X: 54750050-54750050 on Build GRCh38
rs1049832270 Chr.X: 54750304-54750304 on Build GRCh38
rs750318630 Chr.X: 54750005-54750005 on Build GRCh38
rs776016861 Chr.X: 54750057-54750057 on Build GRCh38
rs746327095 Chr.X: 54750049-54750049 on Build GRCh38
rs747705456 Chr.X: 54750040-54750040 on Build GRCh38
rs914173942 Chr.X: 54750423-54750423 on Build GRCh38
rs773867504 Chr.X: 54750210-54750210 on Build GRCh38
rs760604296 Chr.X: 54750408-54750408 on Build GRCh38
rs1035305199 Chr.X: 54750415-54750415 on Build GRCh38
rs925523860 Chr.X: 54750041-54750041 on Build GRCh38
rs765238720 Chr.X: 54749996-54749996 on Build GRCh38
rs1025124770 Chr.X: 54750293-54750293 on Build GRCh38
rs1010872860 Chr.X: 54750389-54750389 on Build GRCh38
rs771472431 Chr.X: 54749973-54749973 on Build GRCh38
rs944646081 Chr.X: 54750215-54750215 on Build GRCh38
rs187348210 Chr.X: 54750307-54750307 on Build GRCh38
rs140339263 Chr.X: 54750204-54750204 on Build GRCh38
rs766618215 Chr.X: 54750017-54750017 on Build GRCh38
rs767192000 Chr.X: 54750247-54750247 on Build GRCh38
rs1006509612 Chr.X: 54750206-54750206 on Build GRCh38
rs370805821 Chr.X: 54750086-54750086 on Build GRCh38
rs765230446 Chr.X: 54750091-54750091 on Build GRCh38
rs377762816 Chr.X: 54749975-54749975 on Build GRCh38
rs1048413995 Chr.X: 54750156-54750156 on Build GRCh38
rs758280845 Chr.X: 54750030-54750030 on Build GRCh38
rs780754985 Chr.X: 54750039-54750039 on Build GRCh38
rs755689738 Chr.X: 54750153-54750153 on Build GRCh38
rs199619872 Chr.X: 54750060-54750060 on Build GRCh38
rs762015353 Chr.X: 54750083-54750083 on Build GRCh38
rs772318334 Chr.X: 54749978-54749978 on Build GRCh38
rs137911364 Chr.X: 54749974-54749974 on Build GRCh38
rs1040290362 Chr.X: 54750216-54750217 on Build GRCh38
rs751264876 Chr.X: 54750031-54750031 on Build GRCh38
rs766257394 Chr.X: 54750121-54750121 on Build GRCh38
rs754005624 Chr.X: 54749995-54749995 on Build GRCh38
rs760955163 Chr.X: 54749987-54749987 on Build GRCh38
rs957695053 Chr.X: 54750406-54750406 on Build GRCh38
rs769007442 Chr.X: 54750061-54750061 on Build GRCh38
rs980455726 Chr.X: 54750003-54750003 on Build GRCh38
rs374171064 Chr.X: 54750155-54750155 on Build GRCh38
rs192200742 Chr.X: 54750411-54750411 on Build GRCh38
rs1013448005 Chr.X: 54750281-54750281 on Build GRCh38
rs112518374 Chr.X: 54750219-54750219 on Build GRCh38
rs754650213 Chr.X: 54750036-54750036 on Build GRCh38

View Full Product Details