Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 57380832-57381330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAH2
Entrez Gene ID
158584
Gene Name
fatty acid amide hydrolase 2
Gene Aliases
AMDD
UniGene
Hs.496205
Gene Chromosome Location
Chr.X: 57121861-57489196 on Build GRCh38
Transcripts
NM_174912.3 XM_005261982.1 XM_005261985.2 XM_005261986.3 XM_005261987.3 XM_011530767.2 XM_017029290.1 XM_017029291.1 XM_017029292.1 XM_017029293.1 XM_017029294.1

SNP Detail

SNP ID Chromosome Location
rs776922722 Chr.X: 57381250-57381250 on Build GRCh38
rs924013045 Chr.X: 57381132-57381132 on Build GRCh38
rs760734330 Chr.X: 57381043-57381043 on Build GRCh38
rs748558287 Chr.X: 57380987-57380987 on Build GRCh38
rs769119761 Chr.X: 57380901-57380901 on Build GRCh38
rs375588401 Chr.X: 57381274-57381274 on Build GRCh38
rs568606647 Chr.X: 57380932-57380932 on Build GRCh38
rs1032318836 Chr.X: 57381196-57381196 on Build GRCh38
rs772655324 Chr.X: 57380924-57380924 on Build GRCh38
rs751433778 Chr.X: 57380880-57380880 on Build GRCh38
rs752422996 Chr.X: 57380863-57380863 on Build GRCh38
rs761387601 Chr.X: 57380895-57380895 on Build GRCh38
rs988733103 Chr.X: 57380919-57380919 on Build GRCh38
rs886178730 Chr.X: 57381214-57381214 on Build GRCh38
rs970750954 Chr.X: 57381285-57381285 on Build GRCh38
rs766389808 Chr.X: 57380963-57380963 on Build GRCh38
rs367704374 Chr.X: 57381067-57381067 on Build GRCh38
rs779374316 Chr.X: 57380903-57380903 on Build GRCh38
rs758766612 Chr.X: 57380990-57380990 on Build GRCh38
rs140681623 Chr.X: 57380995-57380995 on Build GRCh38
rs1041264069 Chr.X: 57381166-57381166 on Build GRCh38
rs762420140 Chr.X: 57380937-57380937 on Build GRCh38
rs374874776 Chr.X: 57381000-57381000 on Build GRCh38
rs768380881 Chr.X: 57381007-57381007 on Build GRCh38
rs199898402 Chr.X: 57381023-57381023 on Build GRCh38
rs762947113 Chr.X: 57381081-57381083 on Build GRCh38
rs766611142 Chr.X: 57380974-57380974 on Build GRCh38
rs1025410021 Chr.X: 57381083-57381083 on Build GRCh38
rs200177220 Chr.X: 57380967-57380967 on Build GRCh38
rs1047432406 Chr.X: 57380941-57380941 on Build GRCh38
rs775111468 Chr.X: 57380934-57380934 on Build GRCh38
rs936820555 Chr.X: 57380855-57380855 on Build GRCh38
rs758382891 Chr.X: 57381159-57381159 on Build GRCh38
rs745981366 Chr.X: 57381153-57381153 on Build GRCh38
rs937050996 Chr.X: 57381294-57381294 on Build GRCh38
rs371388365 Chr.X: 57380976-57380976 on Build GRCh38
rs778338997 Chr.X: 57380992-57380992 on Build GRCh38
rs754130170 Chr.X: 57381088-57381106 on Build GRCh38
rs978210162 Chr.X: 57381128-57381128 on Build GRCh38
rs752602900 Chr.X: 57380981-57380986 on Build GRCh38
rs752929061 Chr.X: 57381081-57381081 on Build GRCh38
rs142859970 Chr.X: 57381226-57381226 on Build GRCh38
rs1024888297 Chr.X: 57381278-57381278 on Build GRCh38
rs898425067 Chr.X: 57381105-57381105 on Build GRCh38
rs780631583 Chr.X: 57381223-57381223 on Build GRCh38
rs750527530 Chr.X: 57380889-57380889 on Build GRCh38
rs201566499 Chr.X: 57380864-57380864 on Build GRCh38
rs763938390 Chr.X: 57380867-57380867 on Build GRCh38
rs150230159 Chr.X: 57380969-57380969 on Build GRCh38
rs766526985 Chr.X: 57381049-57381049 on Build GRCh38
rs748499324 Chr.X: 57381268-57381268 on Build GRCh38
rs775025483 Chr.X: 57380999-57380999 on Build GRCh38
rs767491483 Chr.X: 57380964-57380964 on Build GRCh38
rs369074361 Chr.X: 57380885-57380885 on Build GRCh38
rs41307633 Chr.X: 57380912-57380912 on Build GRCh38
rs936896108 Chr.X: 57380898-57380898 on Build GRCh38
rs748902140 Chr.X: 57381001-57381001 on Build GRCh38
rs765502433 Chr.X: 57381075-57381075 on Build GRCh38
rs911450200 Chr.X: 57380866-57380866 on Build GRCh38
rs1045529954 Chr.X: 57380972-57380972 on Build GRCh38
rs765066368 Chr.X: 57380915-57380915 on Build GRCh38
rs775947116 Chr.X: 57380925-57380925 on Build GRCh38
rs749647019 Chr.X: 57381156-57381156 on Build GRCh38
rs1047505231 Chr.X: 57381197-57381197 on Build GRCh38
rs757615258 Chr.X: 57381136-57381136 on Build GRCh38
rs995410152 Chr.X: 57381131-57381131 on Build GRCh38
rs769067947 Chr.X: 57381229-57381229 on Build GRCh38
rs755097449 Chr.X: 57381071-57381071 on Build GRCh38
rs756336034 Chr.X: 57380890-57380890 on Build GRCh38
rs1037958436 Chr.X: 57381241-57381241 on Build GRCh38
rs200791567 Chr.X: 57381029-57381029 on Build GRCh38
rs970683943 Chr.X: 57381101-57381101 on Build GRCh38
rs982243470 Chr.X: 57381281-57381281 on Build GRCh38
rs138962592 Chr.X: 57380978-57380978 on Build GRCh38
rs779269404 Chr.X: 57381151-57381151 on Build GRCh38
rs779096687 Chr.X: 57380979-57380979 on Build GRCh38
rs997254206 Chr.X: 57381195-57381195 on Build GRCh38
rs780414310 Chr.X: 57380894-57380894 on Build GRCh38
rs757262021 Chr.X: 57380884-57380884 on Build GRCh38
rs767154461 Chr.X: 57381061-57381061 on Build GRCh38
rs751404510 Chr.X: 57380945-57380945 on Build GRCh38
rs922941748 Chr.X: 57381170-57381170 on Build GRCh38
rs932987026 Chr.X: 57381174-57381174 on Build GRCh38
rs200825666 Chr.X: 57380997-57380997 on Build GRCh38
rs763769910 Chr.X: 57380944-57380944 on Build GRCh38
rs1008014832 Chr.X: 57381157-57381157 on Build GRCh38
rs201590323 Chr.X: 57380930-57380930 on Build GRCh38

View Full Product Details