Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 64335969-64336459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR8
Entrez Gene ID
55613
Gene Name
myotubularin related protein 8
Gene Aliases
-
UniGene
Hs.442892
Gene Chromosome Location
Chr.X: 64268081-64395453 on Build GRCh38
Transcripts
NM_017677.3

SNP Detail

SNP ID Chromosome Location
rs951400745 Chr.X: 64336384-64336384 on Build GRCh38
rs762864175 Chr.X: 64336107-64336107 on Build GRCh38
rs773108974 Chr.X: 64335996-64335996 on Build GRCh38
rs958575036 Chr.X: 64336272-64336272 on Build GRCh38
rs1041807951 Chr.X: 64336352-64336352 on Build GRCh38
rs111947425 Chr.X: 64336325-64336325 on Build GRCh38
rs759393547 Chr.X: 64336089-64336089 on Build GRCh38
rs1018998373 Chr.X: 64336158-64336158 on Build GRCh38
rs952773166 Chr.X: 64336214-64336214 on Build GRCh38
rs781587519 Chr.X: 64336144-64336144 on Build GRCh38
rs376795524 Chr.X: 64336132-64336132 on Build GRCh38
rs962079384 Chr.X: 64336162-64336162 on Build GRCh38
rs201031558 Chr.X: 64336123-64336123 on Build GRCh38
rs545141404 Chr.X: 64336230-64336230 on Build GRCh38
rs886348119 Chr.X: 64336370-64336370 on Build GRCh38
rs187944843 Chr.X: 64336027-64336027 on Build GRCh38
rs749791409 Chr.X: 64336167-64336167 on Build GRCh38
rs777950141 Chr.X: 64336412-64336412 on Build GRCh38
rs766021929 Chr.X: 64336078-64336078 on Build GRCh38
rs771611436 Chr.X: 64336051-64336051 on Build GRCh38
rs772770526 Chr.X: 64336052-64336052 on Build GRCh38
rs753509634 Chr.X: 64336278-64336278 on Build GRCh38
rs958537739 Chr.X: 64336305-64336305 on Build GRCh38
rs147190826 Chr.X: 64336156-64336156 on Build GRCh38
rs776368263 Chr.X: 64336079-64336079 on Build GRCh38
rs372721240 Chr.X: 64336069-64336069 on Build GRCh38
rs751696227 Chr.X: 64336120-64336120 on Build GRCh38
rs769167594 Chr.X: 64336175-64336175 on Build GRCh38
rs184421939 Chr.X: 64336402-64336402 on Build GRCh38
rs770151783 Chr.X: 64336037-64336037 on Build GRCh38
rs764989210 Chr.X: 64336090-64336090 on Build GRCh38
rs773167932 Chr.X: 64336195-64336195 on Build GRCh38
rs925637943 Chr.X: 64336386-64336386 on Build GRCh38
rs992635686 Chr.X: 64336322-64336322 on Build GRCh38
rs1000029405 Chr.X: 64336216-64336216 on Build GRCh38
rs759871287 Chr.X: 64336301-64336301 on Build GRCh38
rs200359789 Chr.X: 64336372-64336372 on Build GRCh38
rs368398570 Chr.X: 64336031-64336031 on Build GRCh38
rs192422174 Chr.X: 64336246-64336246 on Build GRCh38
rs773904665 Chr.X: 64336099-64336099 on Build GRCh38
rs773889121 Chr.X: 64336146-64336146 on Build GRCh38
rs1034194067 Chr.X: 64336235-64336235 on Build GRCh38
rs758579631 Chr.X: 64336406-64336406 on Build GRCh38
rs764585494 Chr.X: 64336324-64336324 on Build GRCh38
rs748806662 Chr.X: 64336119-64336119 on Build GRCh38
rs927165915 Chr.X: 64336257-64336257 on Build GRCh38
rs776044266 Chr.X: 64336042-64336042 on Build GRCh38
rs890146114 Chr.X: 64336177-64336177 on Build GRCh38
rs772041909 Chr.X: 64336193-64336193 on Build GRCh38
rs1050707081 Chr.X: 64336409-64336409 on Build GRCh38
rs992764889 Chr.X: 64336302-64336302 on Build GRCh38
rs752117767 Chr.X: 64336413-64336413 on Build GRCh38
rs1010370193 Chr.X: 64336186-64336186 on Build GRCh38

View Full Product Details