Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 150989836-150990329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGB3
Entrez Gene ID
3149
Gene Name
high mobility group box 3
Gene Aliases
HMG-2a, HMG-4, HMG2A, HMG4
UniGene
Hs.19114
Gene Chromosome Location
Chr.X: 150980508-150990775 on Build GRCh38
Transcripts
NM_001301228.1 NM_001301229.1 NM_001301231.1 NM_005342.3

SNP Detail

SNP ID Chromosome Location
rs188132440 Chr.X: 150990286-150990286 on Build GRCh38
rs782787370 Chr.X: 150989920-150989920 on Build GRCh38
rs562132451 Chr.X: 150989919-150989919 on Build GRCh38
rs151157696 Chr.X: 150990012-150990012 on Build GRCh38
rs186124254 Chr.X: 150989985-150989985 on Build GRCh38
rs945957968 Chr.X: 150990290-150990290 on Build GRCh38
rs140209213 Chr.X: 150990031-150990031 on Build GRCh38
rs781914625 Chr.X: 150989903-150989903 on Build GRCh38
rs986210753 Chr.X: 150990062-150990062 on Build GRCh38
rs782122783 Chr.X: 150990095-150990095 on Build GRCh38
rs891696021 Chr.X: 150989869-150989869 on Build GRCh38
rs1030917361 Chr.X: 150990046-150990046 on Build GRCh38
rs782698939 Chr.X: 150990280-150990280 on Build GRCh38
rs1054326265 Chr.X: 150989866-150989866 on Build GRCh38
rs988561390 Chr.X: 150990164-150990164 on Build GRCh38
rs782340273 Chr.X: 150990042-150990042 on Build GRCh38
rs945957828 Chr.X: 150989882-150989882 on Build GRCh38
rs782786190 Chr.X: 150990139-150990139 on Build GRCh38
rs953511391 Chr.X: 150990049-150990049 on Build GRCh38
rs925354708 Chr.X: 150990067-150990067 on Build GRCh38
rs781972183 Chr.X: 150990091-150990091 on Build GRCh38
rs183097635 Chr.X: 150990197-150990197 on Build GRCh38
rs150340549 Chr.X: 150990140-150990140 on Build GRCh38
rs782316060 Chr.X: 150989897-150989897 on Build GRCh38
rs191637253 Chr.X: 150989994-150989994 on Build GRCh38
rs1034092801 Chr.X: 150989923-150989923 on Build GRCh38

View Full Product Details