Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151923909-151924402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEA4
Entrez Gene ID
4103
Gene Name
MAGE family member A4
Gene Aliases
CT1.4, MAGE-41, MAGE-X2, MAGE4, MAGE4A, MAGE4B
UniGene
Hs.37107
Gene Chromosome Location
Chr.X: 151912889-151925170 on Build GRCh38
Transcripts
NM_001011548.1 NM_001011549.1 NM_001011550.1 NM_002362.4 XM_005274677.2 XM_005274678.3 XM_005274679.3

SNP Detail

SNP ID Chromosome Location
rs757742313 Chr.X: 151924147-151924147 on Build GRCh38
rs202047929 Chr.X: 151924132-151924132 on Build GRCh38
rs1012644716 Chr.X: 151923935-151923935 on Build GRCh38
rs61746135 Chr.X: 151924312-151924312 on Build GRCh38
rs141361139 Chr.X: 151924354-151924354 on Build GRCh38
rs772388956 Chr.X: 151924137-151924137 on Build GRCh38
rs749487190 Chr.X: 151924343-151924343 on Build GRCh38
rs747954097 Chr.X: 151924289-151924289 on Build GRCh38
rs999831144 Chr.X: 151924161-151924161 on Build GRCh38
rs754647855 Chr.X: 151924362-151924362 on Build GRCh38
rs373393448 Chr.X: 151923983-151923983 on Build GRCh38
rs151060324 Chr.X: 151924081-151924081 on Build GRCh38
rs776285619 Chr.X: 151923972-151923972 on Build GRCh38
rs753502144 Chr.X: 151924277-151924277 on Build GRCh38
rs746017699 Chr.X: 151924206-151924206 on Build GRCh38
rs775463116 Chr.X: 151923989-151923989 on Build GRCh38
rs955972890 Chr.X: 151924178-151924178 on Build GRCh38
rs200025917 Chr.X: 151924121-151924121 on Build GRCh38
rs572691118 Chr.X: 151924368-151924368 on Build GRCh38
rs1032933723 Chr.X: 151924165-151924165 on Build GRCh38
rs768816689 Chr.X: 151924346-151924346 on Build GRCh38
rs151294389 Chr.X: 151924041-151924041 on Build GRCh38
rs139539716 Chr.X: 151924377-151924377 on Build GRCh38
rs747399608 Chr.X: 151924024-151924024 on Build GRCh38
rs770490747 Chr.X: 151923968-151923968 on Build GRCh38
rs755627386 Chr.X: 151924252-151924252 on Build GRCh38
rs761277562 Chr.X: 151923961-151923961 on Build GRCh38
rs938015724 Chr.X: 151924209-151924209 on Build GRCh38
rs143837393 Chr.X: 151924031-151924031 on Build GRCh38
rs866165760 Chr.X: 151924280-151924280 on Build GRCh38
rs756275786 Chr.X: 151923937-151923937 on Build GRCh38
rs376198310 Chr.X: 151924306-151924306 on Build GRCh38
rs889502225 Chr.X: 151924334-151924334 on Build GRCh38
rs775637049 Chr.X: 151924210-151924210 on Build GRCh38
rs755313244 Chr.X: 151923950-151923950 on Build GRCh38
rs143158098 Chr.X: 151924337-151924337 on Build GRCh38
rs199755286 Chr.X: 151924000-151924000 on Build GRCh38
rs759710688 Chr.X: 151924032-151924032 on Build GRCh38
rs1004465742 Chr.X: 151924044-151924044 on Build GRCh38
rs773420574 Chr.X: 151924019-151924019 on Build GRCh38
rs776616211 Chr.X: 151924111-151924111 on Build GRCh38
rs866684589 Chr.X: 151924370-151924370 on Build GRCh38
rs753449158 Chr.X: 151924357-151924357 on Build GRCh38
rs769617094 Chr.X: 151924087-151924087 on Build GRCh38
rs771004302 Chr.X: 151924109-151924109 on Build GRCh38
rs376596001 Chr.X: 151924162-151924162 on Build GRCh38
rs947215844 Chr.X: 151924302-151924302 on Build GRCh38
rs768259232 Chr.X: 151924352-151924352 on Build GRCh38
rs1047251 Chr.X: 151924181-151924181 on Build GRCh38
rs762366485 Chr.X: 151924231-151924231 on Build GRCh38
rs766382651 Chr.X: 151924027-151924027 on Build GRCh38
rs971230346 Chr.X: 151924191-151924191 on Build GRCh38
rs374430540 Chr.X: 151924313-151924313 on Build GRCh38
rs200774631 Chr.X: 151923960-151923960 on Build GRCh38
rs779785942 Chr.X: 151924174-151924174 on Build GRCh38
rs143502348 Chr.X: 151924330-151924330 on Build GRCh38
rs140138379 Chr.X: 151923963-151923963 on Build GRCh38
rs766749023 Chr.X: 151924051-151924051 on Build GRCh38
rs781034122 Chr.X: 151924207-151924207 on Build GRCh38
rs745500251 Chr.X: 151924083-151924083 on Build GRCh38
rs748090014 Chr.X: 151924193-151924193 on Build GRCh38
rs768575994 Chr.X: 151924100-151924100 on Build GRCh38
rs752062577 Chr.X: 151924136-151924136 on Build GRCh38
rs200853372 Chr.X: 151924180-151924180 on Build GRCh38
rs754011082 Chr.X: 151923947-151923947 on Build GRCh38
rs61741057 Chr.X: 151924264-151924264 on Build GRCh38
rs748977026 Chr.X: 151924240-151924242 on Build GRCh38
rs181401508 Chr.X: 151924064-151924064 on Build GRCh38
rs1046456676 Chr.X: 151923994-151923994 on Build GRCh38
rs199657597 Chr.X: 151924224-151924224 on Build GRCh38
rs150785634 Chr.X: 151924355-151924355 on Build GRCh38
rs755823543 Chr.X: 151924173-151924173 on Build GRCh38
rs749161613 Chr.X: 151924089-151924089 on Build GRCh38
rs778605916 Chr.X: 151924184-151924184 on Build GRCh38
rs771228306 Chr.X: 151924059-151924059 on Build GRCh38
rs61743726 Chr.X: 151924197-151924197 on Build GRCh38
rs778217233 Chr.X: 151923979-151923979 on Build GRCh38
rs752017073 Chr.X: 151924053-151924053 on Build GRCh38
rs759733983 Chr.X: 151924113-151924113 on Build GRCh38
rs754718950 Chr.X: 151924279-151924279 on Build GRCh38
rs765557256 Chr.X: 151924040-151924040 on Build GRCh38
rs766690180 Chr.X: 151923943-151923943 on Build GRCh38
rs750943985 Chr.X: 151924237-151924237 on Build GRCh38
rs373757058 Chr.X: 151923956-151923956 on Build GRCh38
rs866271621 Chr.X: 151923982-151923982 on Build GRCh38
rs141071979 Chr.X: 151924235-151924235 on Build GRCh38
rs773875619 Chr.X: 151924353-151924353 on Build GRCh38
rs776762269 Chr.X: 151924204-151924204 on Build GRCh38
rs200822743 Chr.X: 151924101-151924101 on Build GRCh38
rs371047147 Chr.X: 151924307-151924307 on Build GRCh38
rs763240762 Chr.X: 151924128-151924128 on Build GRCh38
rs769670092 Chr.X: 151923984-151923984 on Build GRCh38
rs779980180 Chr.X: 151924088-151924088 on Build GRCh38
rs778747927 Chr.X: 151924282-151924282 on Build GRCh38
rs201418399 Chr.X: 151923952-151923952 on Build GRCh38
rs765504023 Chr.X: 151924120-151924120 on Build GRCh38
rs775207650 Chr.X: 151924314-151924314 on Build GRCh38
rs745602686 Chr.X: 151924125-151924125 on Build GRCh38
rs750579206 Chr.X: 151923929-151923929 on Build GRCh38

View Full Product Details