Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153183956-153183440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEA1
Entrez Gene ID
4100
Gene Name
MAGE family member A1
Gene Aliases
CT1.1, MAGE1
UniGene
Hs.72879
Gene Chromosome Location
Chr.X: 153179284-153183878 on Build GRCh38
Transcripts
NM_004988.4

View Full Product Details