Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153802481-153802985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
UniGene
Hs.92732
Gene Chromosome Location
Chr.X: 153802166-153830567 on Build GRCh38
Transcripts
NM_001303512.1 NM_001303513.1 NM_001303514.1 NM_001303515.1 NM_001303516.1 NM_032512.3

SNP Detail

SNP ID Chromosome Location
rs1003770077 Chr.X: 153802549-153802549 on Build GRCh38
rs987433573 Chr.X: 153802900-153802900 on Build GRCh38
rs369643303 Chr.X: 153802606-153802606 on Build GRCh38
rs1004550999 Chr.X: 153802673-153802673 on Build GRCh38
rs782504773 Chr.X: 153802690-153802690 on Build GRCh38
rs373555813 Chr.X: 153802912-153802912 on Build GRCh38
rs782441503 Chr.X: 153802545-153802545 on Build GRCh38
rs1014718454 Chr.X: 153802687-153802687 on Build GRCh38
rs139252575 Chr.X: 153802561-153802561 on Build GRCh38
rs915330006 Chr.X: 153802927-153802927 on Build GRCh38
rs939498744 Chr.X: 153802954-153802954 on Build GRCh38
rs375375896 Chr.X: 153802508-153802508 on Build GRCh38
rs186992315 Chr.X: 153802915-153802915 on Build GRCh38
rs782666545 Chr.X: 153802513-153802513 on Build GRCh38
rs781986073 Chr.X: 153802929-153802929 on Build GRCh38
rs963087476 Chr.X: 153802749-153802749 on Build GRCh38
rs946908447 Chr.X: 153802504-153802504 on Build GRCh38
rs181912400 Chr.X: 153802772-153802772 on Build GRCh38
rs1028800058 Chr.X: 153802796-153802796 on Build GRCh38
rs911791046 Chr.X: 153802907-153802907 on Build GRCh38
rs895508580 Chr.X: 153802635-153802635 on Build GRCh38
rs782438581 Chr.X: 153802745-153802745 on Build GRCh38
rs1045654241 Chr.X: 153802942-153802942 on Build GRCh38
rs905390562 Chr.X: 153802950-153802950 on Build GRCh38
rs953429719 Chr.X: 153802868-153802868 on Build GRCh38
rs973644953 Chr.X: 153802776-153802776 on Build GRCh38

View Full Product Details