Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153806653-153807148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
UniGene
Hs.92732
Gene Chromosome Location
Chr.X: 153802166-153830567 on Build GRCh38
Transcripts
NM_001303512.1 NM_001303513.1 NM_001303514.1 NM_001303515.1 NM_001303516.1 NM_032512.3

SNP Detail

SNP ID Chromosome Location
rs782447726 Chr.X: 153806748-153806748 on Build GRCh38
rs782375891 Chr.X: 153806838-153806838 on Build GRCh38
rs782100909 Chr.X: 153807095-153807095 on Build GRCh38
rs782808516 Chr.X: 153806708-153806708 on Build GRCh38
rs782368635 Chr.X: 153806691-153806691 on Build GRCh38
rs782759147 Chr.X: 153806888-153806888 on Build GRCh38
rs5987159 Chr.X: 153806775-153806775 on Build GRCh38
rs782139290 Chr.X: 153806702-153806702 on Build GRCh38
rs781887888 Chr.X: 153806732-153806732 on Build GRCh38
rs200216382 Chr.X: 153806694-153806694 on Build GRCh38
rs782716065 Chr.X: 153807066-153807066 on Build GRCh38
rs781923667 Chr.X: 153807091-153807091 on Build GRCh38
rs368333181 Chr.X: 153806751-153806751 on Build GRCh38
rs199765870 Chr.X: 153806774-153806774 on Build GRCh38
rs782784076 Chr.X: 153806731-153806731 on Build GRCh38
rs782155122 Chr.X: 153807033-153807033 on Build GRCh38
rs782746543 Chr.X: 153807111-153807111 on Build GRCh38
rs782281170 Chr.X: 153807030-153807030 on Build GRCh38
rs1018502558 Chr.X: 153806947-153806947 on Build GRCh38
rs977791345 Chr.X: 153807070-153807070 on Build GRCh38
rs782104101 Chr.X: 153806836-153806836 on Build GRCh38
rs782079901 Chr.X: 153806873-153806873 on Build GRCh38
rs782071819 Chr.X: 153806714-153806714 on Build GRCh38
rs957661370 Chr.X: 153807101-153807101 on Build GRCh38
rs888161972 Chr.X: 153806932-153806932 on Build GRCh38
rs782737270 Chr.X: 153806715-153806715 on Build GRCh38
rs782180666 Chr.X: 153806805-153806805 on Build GRCh38
rs782375432 Chr.X: 153806871-153806871 on Build GRCh38
rs1005388432 Chr.X: 153806933-153806933 on Build GRCh38
rs782678533 Chr.X: 153806997-153806997 on Build GRCh38
rs1033100545 Chr.X: 153807090-153807090 on Build GRCh38
rs139521573 Chr.X: 153806778-153806778 on Build GRCh38
rs782685044 Chr.X: 153806782-153806782 on Build GRCh38
rs781798686 Chr.X: 153806896-153806896 on Build GRCh38
rs1029516770 Chr.X: 153807025-153807025 on Build GRCh38
rs781843909 Chr.X: 153806718-153806718 on Build GRCh38
rs1056326439 Chr.X: 153806745-153806745 on Build GRCh38
rs781923162 Chr.X: 153806710-153806710 on Build GRCh38
rs782436692 Chr.X: 153806897-153806897 on Build GRCh38
rs376310213 Chr.X: 153806776-153806776 on Build GRCh38
rs782812995 Chr.X: 153806859-153806859 on Build GRCh38
rs781958360 Chr.X: 153806833-153806833 on Build GRCh38
rs201192058 Chr.X: 153806812-153806812 on Build GRCh38
rs782154064 Chr.X: 153806858-153806858 on Build GRCh38
rs370393734 Chr.X: 153806891-153806891 on Build GRCh38
rs782038828 Chr.X: 153806887-153806887 on Build GRCh38
rs782504180 Chr.X: 153806725-153806725 on Build GRCh38
rs782000786 Chr.X: 153806857-153806857 on Build GRCh38
rs963943733 Chr.X: 153806993-153806993 on Build GRCh38

View Full Product Details