Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230678835-230679342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG2
Entrez Gene ID
22796
Gene Name
component of oligomeric golgi complex 2
Gene Aliases
LDLC
UniGene
Hs.211800
Gene Chromosome Location
Chr.1: 230642456-230693985 on Build GRCh38
Transcripts
NM_001145036.1 NM_007357.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6577967 c.1027-1G>C p.? Chr.1: 230678912-230678912 on Build GRCh38
5628872 c.1088G>A p.S363N Chr.1: 230678974-230678974 on Build GRCh38
4807987 c.1066C>T p.R356W Chr.1: 230678952-230678952 on Build GRCh38
3804295 c.1161A>C p.Q387H Chr.1: 230679047-230679047 on Build GRCh38
4029382 c.1083G>T p.Q361H Chr.1: 230678969-230678969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368403793 Chr.1: 230679003-230679003 on Build GRCh38
rs368731244 Chr.1: 230679087-230679087 on Build GRCh38
rs765350060 Chr.1: 230679071-230679071 on Build GRCh38
rs778959258 Chr.1: 230678970-230678970 on Build GRCh38
rs74142999 Chr.1: 230679082-230679082 on Build GRCh38
rs367872920 Chr.1: 230679091-230679091 on Build GRCh38
rs370562001 Chr.1: 230678926-230678926 on Build GRCh38
rs773980476 Chr.1: 230679019-230679019 on Build GRCh38
rs543140389 Chr.1: 230678875-230678875 on Build GRCh38
rs760326288 Chr.1: 230679049-230679049 on Build GRCh38
rs781779396 Chr.1: 230679010-230679010 on Build GRCh38
rs750682297 Chr.1: 230678976-230678976 on Build GRCh38
rs527625063 Chr.1: 230679162-230679162 on Build GRCh38
rs547986966 Chr.1: 230679073-230679073 on Build GRCh38
rs572930354 Chr.1: 230679312-230679312 on Build GRCh38
rs374544261 Chr.1: 230679062-230679062 on Build GRCh38
rs138816611 Chr.1: 230678884-230678884 on Build GRCh38
rs753206203 Chr.1: 230679307-230679307 on Build GRCh38
rs772855175 Chr.1: 230678900-230678900 on Build GRCh38
rs559978069 Chr.1: 230679124-230679124 on Build GRCh38
rs913752071 Chr.1: 230679208-230679208 on Build GRCh38
rs953506011 Chr.1: 230679215-230679215 on Build GRCh38
rs951977439 Chr.1: 230678861-230678861 on Build GRCh38
rs1029028756 Chr.1: 230679212-230679212 on Build GRCh38
rs921909246 Chr.1: 230679004-230679004 on Build GRCh38
rs748660922 Chr.1: 230678879-230678879 on Build GRCh38
rs779045293 Chr.1: 230678872-230678872 on Build GRCh38
rs781669935 Chr.1: 230679080-230679080 on Build GRCh38
rs557130822 Chr.1: 230679130-230679130 on Build GRCh38
rs200705175 Chr.1: 230678952-230678952 on Build GRCh38
rs375651539 Chr.1: 230679096-230679096 on Build GRCh38
rs770173384 Chr.1: 230679090-230679090 on Build GRCh38
rs140603331 Chr.1: 230679194-230679194 on Build GRCh38
rs747271864 Chr.1: 230679000-230679000 on Build GRCh38
rs759237469 Chr.1: 230678921-230678921 on Build GRCh38
rs904861264 Chr.1: 230678939-230678939 on Build GRCh38
rs199851079 Chr.1: 230679070-230679070 on Build GRCh38
rs754640938 Chr.1: 230678897-230678901 on Build GRCh38
rs370154364 Chr.1: 230678881-230678881 on Build GRCh38
rs748550486 Chr.1: 230679071-230679072 on Build GRCh38
rs552209403 Chr.1: 230678940-230678940 on Build GRCh38
rs376490327 Chr.1: 230678901-230678901 on Build GRCh38
rs887676715 Chr.1: 230679137-230679137 on Build GRCh38
rs778599212 Chr.1: 230679086-230679086 on Build GRCh38
rs771586673 Chr.1: 230679032-230679032 on Build GRCh38
rs768478383 Chr.1: 230679100-230679100 on Build GRCh38
rs1004729484 Chr.1: 230679139-230679139 on Build GRCh38
rs926559685 Chr.1: 230679313-230679313 on Build GRCh38
rs925540841 Chr.1: 230679301-230679301 on Build GRCh38
rs758653468 Chr.1: 230679075-230679075 on Build GRCh38
rs771767014 Chr.1: 230679090-230679090 on Build GRCh38
rs768022678 Chr.1: 230679072-230679073 on Build GRCh38
rs373983282 Chr.1: 230679089-230679089 on Build GRCh38
rs776388698 Chr.1: 230679102-230679102 on Build GRCh38
rs774217419 Chr.1: 230678920-230678920 on Build GRCh38
rs192254511 Chr.1: 230679135-230679135 on Build GRCh38
rs370937568 Chr.1: 230679029-230679029 on Build GRCh38
rs201805558 Chr.1: 230679076-230679076 on Build GRCh38
rs758525752 Chr.1: 230678992-230678992 on Build GRCh38
rs751218797 Chr.1: 230678867-230678870 on Build GRCh38
rs570852945 Chr.1: 230679274-230679274 on Build GRCh38
rs770610327 Chr.1: 230679016-230679016 on Build GRCh38
rs552970138 Chr.1: 230679267-230679267 on Build GRCh38
rs41307686 Chr.1: 230679081-230679081 on Build GRCh38
rs760695498 Chr.1: 230678931-230678931 on Build GRCh38
rs780488482 Chr.1: 230678993-230678993 on Build GRCh38
rs775357737 Chr.1: 230679036-230679036 on Build GRCh38
rs905520700 Chr.1: 230679111-230679111 on Build GRCh38
rs988368996 Chr.1: 230679201-230679201 on Build GRCh38
rs966139506 Chr.1: 230679165-230679165 on Build GRCh38
rs750452260 Chr.1: 230679074-230679074 on Build GRCh38
rs753835492 Chr.1: 230679069-230679069 on Build GRCh38
rs11558606 Chr.1: 230678922-230678922 on Build GRCh38
rs746665643 Chr.1: 230679092-230679092 on Build GRCh38
rs769221163 Chr.1: 230679009-230679009 on Build GRCh38
rs770551453 Chr.1: 230678906-230678906 on Build GRCh38
rs548935579 Chr.1: 230679077-230679077 on Build GRCh38
rs769005186 Chr.1: 230678889-230678889 on Build GRCh38
rs748661829 Chr.1: 230679011-230679011 on Build GRCh38

View Full Product Details