Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231336554-231337051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC8
Entrez Gene ID
149371
Gene Name
exocyst complex component 8
Gene Aliases
EXO84, Exo84p, SEC84
UniGene
Hs.356198
Gene Chromosome Location
Chr.1: 231332731-231337872 on Build GRCh38
Transcripts
NM_175876.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3418881 c.865C>T p.R289* Chr.1: 231336881-231336881 on Build GRCh38
6192204 c.922G>A p.A308T Chr.1: 231336824-231336824 on Build GRCh38
6125385 c.1174G>C p.E392Q Chr.1: 231336572-231336572 on Build GRCh38
7411352 c.1052G>A p.R351K Chr.1: 231336694-231336694 on Build GRCh38
6261798 c.1031T>A p.L344Q Chr.1: 231336715-231336715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777482682 Chr.1: 231336954-231336954 on Build GRCh38
rs769601886 Chr.1: 231336930-231336930 on Build GRCh38
rs1039609498 Chr.1: 231336788-231336788 on Build GRCh38
rs753184053 Chr.1: 231337002-231337002 on Build GRCh38
rs762440406 Chr.1: 231337025-231337025 on Build GRCh38
rs746595093 Chr.1: 231336714-231336714 on Build GRCh38
rs368244909 Chr.1: 231336865-231336865 on Build GRCh38
rs375507618 Chr.1: 231336854-231336854 on Build GRCh38
rs781704189 Chr.1: 231336576-231336576 on Build GRCh38
rs781359798 Chr.1: 231336670-231336670 on Build GRCh38
rs773334020 Chr.1: 231336765-231336765 on Build GRCh38
rs111580847 Chr.1: 231336773-231336773 on Build GRCh38
rs200935314 Chr.1: 231336922-231336922 on Build GRCh38
rs745358364 Chr.1: 231336682-231336682 on Build GRCh38
rs775884170 Chr.1: 231336987-231336987 on Build GRCh38
rs143120742 Chr.1: 231336647-231336647 on Build GRCh38
rs768511878 Chr.1: 231336716-231336716 on Build GRCh38
rs761088452 Chr.1: 231336892-231336892 on Build GRCh38
rs766606141 Chr.1: 231336659-231336659 on Build GRCh38
rs778746897 Chr.1: 231336789-231336791 on Build GRCh38
rs141863406 Chr.1: 231336827-231336827 on Build GRCh38
rs747920350 Chr.1: 231336830-231336830 on Build GRCh38
rs765149961 Chr.1: 231336762-231336762 on Build GRCh38
rs754711526 Chr.1: 231336915-231336915 on Build GRCh38
rs770764934 Chr.1: 231336968-231336968 on Build GRCh38
rs773167890 Chr.1: 231336850-231336850 on Build GRCh38
rs753161318 Chr.1: 231336882-231336882 on Build GRCh38
rs370713369 Chr.1: 231336814-231336814 on Build GRCh38
rs755095564 Chr.1: 231336781-231336781 on Build GRCh38
rs571330269 Chr.1: 231336977-231336977 on Build GRCh38
rs778402584 Chr.1: 231336801-231336801 on Build GRCh38
rs1043246125 Chr.1: 231336842-231336842 on Build GRCh38
rs375459948 Chr.1: 231336579-231336579 on Build GRCh38
rs943359564 Chr.1: 231336775-231336775 on Build GRCh38
rs973941506 Chr.1: 231336725-231336725 on Build GRCh38
rs774226282 Chr.1: 231336859-231336859 on Build GRCh38
rs771053553 Chr.1: 231336857-231336857 on Build GRCh38
rs1023226660 Chr.1: 231337006-231337006 on Build GRCh38
rs775206321 Chr.1: 231336618-231336618 on Build GRCh38
rs148098436 Chr.1: 231336912-231336912 on Build GRCh38
rs557870432 Chr.1: 231336873-231336873 on Build GRCh38
rs144793875 Chr.1: 231336791-231336791 on Build GRCh38
rs756979233 Chr.1: 231336779-231336780 on Build GRCh38
rs776653782 Chr.1: 231336641-231336641 on Build GRCh38
rs367850043 Chr.1: 231336687-231336687 on Build GRCh38
rs779375116 Chr.1: 231336815-231336815 on Build GRCh38
rs746679996 Chr.1: 231336818-231336818 on Build GRCh38
rs759512430 Chr.1: 231336975-231336975 on Build GRCh38
rs770584042 Chr.1: 231336605-231336605 on Build GRCh38
rs771670194 Chr.1: 231336684-231336684 on Build GRCh38
rs764511851 Chr.1: 231336894-231336894 on Build GRCh38
rs745515473 Chr.1: 231336606-231336606 on Build GRCh38
rs148464625 Chr.1: 231336610-231336610 on Build GRCh38
rs752877756 Chr.1: 231336691-231336692 on Build GRCh38
rs769766967 Chr.1: 231336846-231336846 on Build GRCh38
rs374382054 Chr.1: 231336678-231336678 on Build GRCh38
rs761806541 Chr.1: 231336755-231336755 on Build GRCh38
rs768189494 Chr.1: 231336824-231336824 on Build GRCh38
rs377577608 Chr.1: 231336664-231336664 on Build GRCh38
rs750498559 Chr.1: 231336645-231336645 on Build GRCh38
rs760494680 Chr.1: 231336623-231336623 on Build GRCh38
rs745409431 Chr.1: 231336805-231336805 on Build GRCh38
rs370183913 Chr.1: 231336672-231336672 on Build GRCh38
rs748623243 Chr.1: 231336583-231336583 on Build GRCh38
rs1021390523 Chr.1: 231336625-231336625 on Build GRCh38
rs765305372 Chr.1: 231336643-231336643 on Build GRCh38
rs967491162 Chr.1: 231336626-231336626 on Build GRCh38
rs145329289 Chr.1: 231336774-231336774 on Build GRCh38
rs200964466 Chr.1: 231336660-231336660 on Build GRCh38
rs1035428073 Chr.1: 231336951-231336951 on Build GRCh38
rs760770827 Chr.1: 231336911-231336911 on Build GRCh38
rs776341442 Chr.1: 231336752-231336752 on Build GRCh38
rs749249878 Chr.1: 231336960-231336960 on Build GRCh38
rs1011793693 Chr.1: 231336603-231336603 on Build GRCh38
rs999036251 Chr.1: 231336883-231336883 on Build GRCh38
rs775175509 Chr.1: 231336686-231336686 on Build GRCh38
rs200299813 Chr.1: 231336903-231336903 on Build GRCh38
rs764558964 Chr.1: 231337023-231337023 on Build GRCh38
rs780795802 Chr.1: 231336919-231336919 on Build GRCh38
rs537536815 Chr.1: 231336795-231336795 on Build GRCh38
rs483352868 Chr.1: 231336952-231336952 on Build GRCh38
rs139980773 Chr.1: 231336633-231336633 on Build GRCh38
rs959708933 Chr.1: 231336958-231336958 on Build GRCh38
rs757955624 Chr.1: 231336807-231336807 on Build GRCh38
rs971193466 Chr.1: 231336707-231336707 on Build GRCh38
rs754384583 Chr.1: 231336901-231336901 on Build GRCh38
rs200538583 Chr.1: 231336768-231336768 on Build GRCh38
rs977121056 Chr.1: 231336632-231336632 on Build GRCh38
rs774543887 Chr.1: 231336970-231336970 on Build GRCh38

View Full Product Details