Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 232806335-232806838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP10
Entrez Gene ID
54627
Gene Name
microtubule associated protein 10
Gene Aliases
KIAA1383, MTR120
UniGene
Hs.160373
Gene Chromosome Location
Chr.1: 232804892-232810346 on Build GRCh38
Transcripts
NM_019090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3677606 c.1314C>A p.T438T Chr.1: 232806337-232806337 on Build GRCh38
5010755 c.1412C>A p.A471E Chr.1: 232806435-232806435 on Build GRCh38
222656 c.1420G>A p.E474K Chr.1: 232806443-232806443 on Build GRCh38
5786788 c.1783T>G p.S595A Chr.1: 232806806-232806806 on Build GRCh38
1999141 c.1442delA p.K483fs*3 Chr.1: 232806465-232806465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530581925 Chr.1: 232806743-232806745 on Build GRCh38
rs377308893 Chr.1: 232806584-232806584 on Build GRCh38
rs763386839 Chr.1: 232806771-232806771 on Build GRCh38
rs371993579 Chr.1: 232806393-232806393 on Build GRCh38
rs750367267 Chr.1: 232806463-232806464 on Build GRCh38
rs775183254 Chr.1: 232806487-232806487 on Build GRCh38
rs775910783 Chr.1: 232806797-232806797 on Build GRCh38
rs371454812 Chr.1: 232806804-232806804 on Build GRCh38
rs971846359 Chr.1: 232806503-232806503 on Build GRCh38
rs746089137 Chr.1: 232806372-232806372 on Build GRCh38
rs747225953 Chr.1: 232806521-232806521 on Build GRCh38
rs867944388 Chr.1: 232806560-232806560 on Build GRCh38
rs775734557 Chr.1: 232806388-232806388 on Build GRCh38
rs770365421 Chr.1: 232806472-232806472 on Build GRCh38
rs765887159 Chr.1: 232806469-232806477 on Build GRCh38
rs374844165 Chr.1: 232806701-232806701 on Build GRCh38
rs768241074 Chr.1: 232806582-232806582 on Build GRCh38
rs368819098 Chr.1: 232806466-232806466 on Build GRCh38
rs942842804 Chr.1: 232806421-232806421 on Build GRCh38
rs766159750 Chr.1: 232806715-232806715 on Build GRCh38
rs760364045 Chr.1: 232806570-232806570 on Build GRCh38
rs368276824 Chr.1: 232806792-232806792 on Build GRCh38
rs774203005 Chr.1: 232806407-232806407 on Build GRCh38
rs369334825 Chr.1: 232806611-232806611 on Build GRCh38
rs201495236 Chr.1: 232806417-232806417 on Build GRCh38
rs750902654 Chr.1: 232806809-232806809 on Build GRCh38
rs540700375 Chr.1: 232806789-232806789 on Build GRCh38
rs753741442 Chr.1: 232806425-232806425 on Build GRCh38
rs776707263 Chr.1: 232806609-232806609 on Build GRCh38
rs773806746 Chr.1: 232806531-232806531 on Build GRCh38
rs754095187 Chr.1: 232806741-232806741 on Build GRCh38
rs770920549 Chr.1: 232806779-232806779 on Build GRCh38
rs928699014 Chr.1: 232806802-232806802 on Build GRCh38
rs894418958 Chr.1: 232806729-232806729 on Build GRCh38
rs769181679 Chr.1: 232806534-232806534 on Build GRCh38
rs765072281 Chr.1: 232806459-232806461 on Build GRCh38
rs778040060 Chr.1: 232806810-232806811 on Build GRCh38
rs759636237 Chr.1: 232806724-232806724 on Build GRCh38
rs758558452 Chr.1: 232806441-232806441 on Build GRCh38
rs754552405 Chr.1: 232806814-232806814 on Build GRCh38
rs531801115 Chr.1: 232806367-232806367 on Build GRCh38
rs765096716 Chr.1: 232806507-232806507 on Build GRCh38
rs907198061 Chr.1: 232806634-232806634 on Build GRCh38
rs778185393 Chr.1: 232806740-232806740 on Build GRCh38
rs558263978 Chr.1: 232806737-232806737 on Build GRCh38
rs768328956 Chr.1: 232806685-232806685 on Build GRCh38
rs61735495 Chr.1: 232806530-232806530 on Build GRCh38
rs762959626 Chr.1: 232806710-232806710 on Build GRCh38
rs879048155 Chr.1: 232806545-232806545 on Build GRCh38
rs747357408 Chr.1: 232806456-232806456 on Build GRCh38
rs545302766 Chr.1: 232806529-232806529 on Build GRCh38
rs77064011 Chr.1: 232806649-232806649 on Build GRCh38
rs76557656 Chr.1: 232806661-232806661 on Build GRCh38
rs747870351 Chr.1: 232806760-232806760 on Build GRCh38
rs958532745 Chr.1: 232806497-232806497 on Build GRCh38
rs780867095 Chr.1: 232806767-232806767 on Build GRCh38
rs772549229 Chr.1: 232806375-232806375 on Build GRCh38
rs767900297 Chr.1: 232806730-232806730 on Build GRCh38
rs759123636 Chr.1: 232806408-232806408 on Build GRCh38
rs59165168 Chr.1: 232806800-232806800 on Build GRCh38
rs370076210 Chr.1: 232806512-232806512 on Build GRCh38
rs753130637 Chr.1: 232806655-232806655 on Build GRCh38
rs771537632 Chr.1: 232806675-232806675 on Build GRCh38
rs755885180 Chr.1: 232806414-232806414 on Build GRCh38
rs780284027 Chr.1: 232806453-232806453 on Build GRCh38
rs951681363 Chr.1: 232806536-232806536 on Build GRCh38
rs1032896182 Chr.1: 232806493-232806493 on Build GRCh38
rs776220979 Chr.1: 232806692-232806692 on Build GRCh38
rs368008854 Chr.1: 232806801-232806801 on Build GRCh38
rs998664674 Chr.1: 232806459-232806459 on Build GRCh38
rs749795640 Chr.1: 232806667-232806667 on Build GRCh38
rs961906974 Chr.1: 232806705-232806705 on Build GRCh38
rs753833447 Chr.1: 232806504-232806504 on Build GRCh38
rs761556946 Chr.1: 232806583-232806583 on Build GRCh38
rs745893990 Chr.1: 232806433-232806433 on Build GRCh38
rs370144988 Chr.1: 232806677-232806677 on Build GRCh38
rs779248620 Chr.1: 232806752-232806752 on Build GRCh38
rs780669194 Chr.1: 232806815-232806815 on Build GRCh38
rs990453589 Chr.1: 232806777-232806777 on Build GRCh38
rs767129825 Chr.1: 232806410-232806410 on Build GRCh38
rs12048236 Chr.1: 232806505-232806505 on Build GRCh38
rs750997849 Chr.1: 232806509-232806509 on Build GRCh38
rs547013302 Chr.1: 232806603-232806603 on Build GRCh38
rs750355248 Chr.1: 232806596-232806596 on Build GRCh38
rs760424355 Chr.1: 232806498-232806498 on Build GRCh38
rs761536718 Chr.1: 232806374-232806375 on Build GRCh38
rs536150408 Chr.1: 232806650-232806650 on Build GRCh38
rs200834253 Chr.1: 232806627-232806627 on Build GRCh38
rs754543309 Chr.1: 232806613-232806616 on Build GRCh38
rs374323821 Chr.1: 232806768-232806768 on Build GRCh38
rs1055000034 Chr.1: 232806539-232806539 on Build GRCh38
rs756544749 Chr.1: 232806811-232806811 on Build GRCh38
rs371285850 Chr.1: 232806813-232806813 on Build GRCh38
rs768136623 Chr.1: 232806763-232806763 on Build GRCh38
rs971192486 Chr.1: 232806516-232806516 on Build GRCh38
rs746439480 Chr.1: 232806683-232806683 on Build GRCh38
rs769043513 Chr.1: 232806461-232806461 on Build GRCh38
rs775069645 Chr.1: 232806543-232806543 on Build GRCh38
rs1041797515 Chr.1: 232806670-232806670 on Build GRCh38
rs774130125 Chr.1: 232806787-232806787 on Build GRCh38
rs777320366 Chr.1: 232806462-232806462 on Build GRCh38
rs745418605 Chr.1: 232806533-232806533 on Build GRCh38
rs746545313 Chr.1: 232806755-232806755 on Build GRCh38
rs770972855 Chr.1: 232806363-232806363 on Build GRCh38
rs781699219 Chr.1: 232806528-232806528 on Build GRCh38
rs752461062 Chr.1: 232806413-232806413 on Build GRCh38
rs777230519 Chr.1: 232806392-232806392 on Build GRCh38
rs368167890 Chr.1: 232806391-232806391 on Build GRCh38
rs757397578 Chr.1: 232806427-232806427 on Build GRCh38
rs780190597 Chr.1: 232806513-232806513 on Build GRCh38
rs765980497 Chr.1: 232806394-232806394 on Build GRCh38
rs759982691 Chr.1: 232806699-232806699 on Build GRCh38
rs202017337 Chr.1: 232806428-232806428 on Build GRCh38
rs756612275 Chr.1: 232806738-232806738 on Build GRCh38
rs373474225 Chr.1: 232806484-232806484 on Build GRCh38
rs750500093 Chr.1: 232806511-232806511 on Build GRCh38
rs762985587 Chr.1: 232806598-232806598 on Build GRCh38
rs765734288 Chr.1: 232806734-232806734 on Build GRCh38
rs140737636 Chr.1: 232806723-232806723 on Build GRCh38
rs371716712 Chr.1: 232806799-232806799 on Build GRCh38
rs774002444 Chr.1: 232806474-232806474 on Build GRCh38
rs758304568 Chr.1: 232806464-232806465 on Build GRCh38
rs370524626 Chr.1: 232806438-232806438 on Build GRCh38
rs761649119 Chr.1: 232806696-232806696 on Build GRCh38
rs61738408 Chr.1: 232806759-232806759 on Build GRCh38
rs1056166738 Chr.1: 232806811-232806811 on Build GRCh38
rs755353337 Chr.1: 232806526-232806526 on Build GRCh38
rs778463918 Chr.1: 232806665-232806665 on Build GRCh38

View Full Product Details