Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 233014566-233015093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCNX2
Entrez Gene ID
80003
Gene Name
pecanex homolog 2 (Drosophila)
Gene Aliases
PCNXL2
UniGene
Hs.370605
Gene Chromosome Location
Chr.1: 232984109-233295735 on Build GRCh38
Transcripts
NM_014801.3 XM_011544278.2 XM_017002391.1 XM_017002392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1219837 c.4900T>C p.C1634R Chr.1: 233014717-233014717 on Build GRCh38
3966362 c.4896C>G p.A1632A Chr.1: 233014721-233014721 on Build GRCh38
3485281 c.4910G>A p.G1637E Chr.1: 233014707-233014707 on Build GRCh38
3966360 c.4897C>T p.L1633L Chr.1: 233014720-233014720 on Build GRCh38
464158 c.4902C>A p.C1634* Chr.1: 233014715-233014715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761324988 Chr.1: 233014820-233014820 on Build GRCh38
rs781556554 Chr.1: 233014736-233014736 on Build GRCh38
rs1019445363 Chr.1: 233014902-233014902 on Build GRCh38
rs748248856 Chr.1: 233014738-233014738 on Build GRCh38
rs750400029 Chr.1: 233014698-233014698 on Build GRCh38
rs369884137 Chr.1: 233014801-233014801 on Build GRCh38
rs753642963 Chr.1: 233014685-233014685 on Build GRCh38
rs760175231 Chr.1: 233014777-233014777 on Build GRCh38
rs765114332 Chr.1: 233014822-233014822 on Build GRCh38
rs187317190 Chr.1: 233014961-233014961 on Build GRCh38
rs371729919 Chr.1: 233014648-233014648 on Build GRCh38
rs199851825 Chr.1: 233014756-233014756 on Build GRCh38
rs928265986 Chr.1: 233014962-233014963 on Build GRCh38
rs370428244 Chr.1: 233014959-233014959 on Build GRCh38
rs745727036 Chr.1: 233014622-233014622 on Build GRCh38
rs750157011 Chr.1: 233014824-233014824 on Build GRCh38
rs758462163 Chr.1: 233014827-233014827 on Build GRCh38
rs978412473 Chr.1: 233014974-233014974 on Build GRCh38
rs940290160 Chr.1: 233014707-233014707 on Build GRCh38
rs769054415 Chr.1: 233014730-233014730 on Build GRCh38
rs775665369 Chr.1: 233014626-233014626 on Build GRCh38
rs180904792 Chr.1: 233015059-233015059 on Build GRCh38
rs368017178 Chr.1: 233014660-233014660 on Build GRCh38
rs949460738 Chr.1: 233014761-233014761 on Build GRCh38
rs887021468 Chr.1: 233014848-233014848 on Build GRCh38
rs571113775 Chr.1: 233014656-233014656 on Build GRCh38
rs770205240 Chr.1: 233014753-233014753 on Build GRCh38
rs201011739 Chr.1: 233014651-233014651 on Build GRCh38
rs374786513 Chr.1: 233015040-233015040 on Build GRCh38
rs181543205 Chr.1: 233014724-233014724 on Build GRCh38
rs769718791 Chr.1: 233014684-233014684 on Build GRCh38
rs747067569 Chr.1: 233014727-233014727 on Build GRCh38
rs752163223 Chr.1: 233014674-233014674 on Build GRCh38
rs115492225 Chr.1: 233014857-233014857 on Build GRCh38
rs977444873 Chr.1: 233014644-233014644 on Build GRCh38
rs903080082 Chr.1: 233014787-233014787 on Build GRCh38
rs535926945 Chr.1: 233014936-233014936 on Build GRCh38
rs1055423646 Chr.1: 233015066-233015066 on Build GRCh38
rs76659968 Chr.1: 233015034-233015034 on Build GRCh38
rs75089072 Chr.1: 233014658-233014658 on Build GRCh38
rs938361182 Chr.1: 233015007-233015007 on Build GRCh38
rs775029568 Chr.1: 233014770-233014770 on Build GRCh38
rs781185374 Chr.1: 233014681-233014681 on Build GRCh38
rs199869615 Chr.1: 233014690-233014690 on Build GRCh38
rs747270171 Chr.1: 233014627-233014627 on Build GRCh38
rs544410916 Chr.1: 233014992-233014992 on Build GRCh38
rs573174164 Chr.1: 233014914-233014914 on Build GRCh38
rs767933726 Chr.1: 233014694-233014695 on Build GRCh38
rs745696073 Chr.1: 233014650-233014650 on Build GRCh38
rs374706528 Chr.1: 233014751-233014751 on Build GRCh38
rs772208255 Chr.1: 233014624-233014624 on Build GRCh38
rs967731695 Chr.1: 233014617-233014617 on Build GRCh38
rs956947405 Chr.1: 233015015-233015015 on Build GRCh38
rs1045129334 Chr.1: 233014807-233014807 on Build GRCh38
rs763545333 Chr.1: 233014760-233014760 on Build GRCh38
rs975454479 Chr.1: 233014877-233014877 on Build GRCh38
rs961622117 Chr.1: 233014950-233014950 on Build GRCh38
rs908734161 Chr.1: 233014946-233014946 on Build GRCh38
rs758231096 Chr.1: 233014712-233014712 on Build GRCh38
rs1048791279 Chr.1: 233014838-233014838 on Build GRCh38
rs935506825 Chr.1: 233014717-233014717 on Build GRCh38
rs371404594 Chr.1: 233014664-233014664 on Build GRCh38
rs1028419365 Chr.1: 233014889-233014889 on Build GRCh38
rs754699872 Chr.1: 233014671-233014671 on Build GRCh38
rs765728500 Chr.1: 233014657-233014657 on Build GRCh38
rs952941668 Chr.1: 233014906-233014906 on Build GRCh38
rs372680532 Chr.1: 233014815-233014815 on Build GRCh38
rs778516209 Chr.1: 233014696-233014696 on Build GRCh38
rs975103693 Chr.1: 233014595-233014595 on Build GRCh38
rs190513204 Chr.1: 233014985-233014985 on Build GRCh38
rs529921543 Chr.1: 233014878-233014878 on Build GRCh38
rs771396062 Chr.1: 233014769-233014769 on Build GRCh38
rs921407656 Chr.1: 233014659-233014659 on Build GRCh38

View Full Product Details