Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235119731-235120241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOMM20
Entrez Gene ID
9804
Gene Name
translocase of outer mitochondrial membrane 20
Gene Aliases
MAS20, MOM19, TOM20
UniGene
Hs.533192
Gene Chromosome Location
Chr.1: 235109341-235128941 on Build GRCh38
Transcripts
NM_014765.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5914803 c.169T>C p.L57L Chr.1: 235119899-235119899 on Build GRCh38
247944 c.231T>G p.G77G Chr.1: 235119837-235119837 on Build GRCh38
1320367 c.175G>A p.D59N Chr.1: 235119893-235119893 on Build GRCh38
5914802 c.193G>A p.A65T Chr.1: 235119875-235119875 on Build GRCh38
3734179 c.191A>C p.E64A Chr.1: 235119877-235119877 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763321324 Chr.1: 235119926-235119928 on Build GRCh38
rs752980342 Chr.1: 235119800-235119800 on Build GRCh38
rs763331169 Chr.1: 235119918-235119918 on Build GRCh38
rs746224351 Chr.1: 235119885-235119885 on Build GRCh38
rs770121240 Chr.1: 235119777-235119777 on Build GRCh38
rs373609125 Chr.1: 235120189-235120189 on Build GRCh38
rs560898854 Chr.1: 235120034-235120034 on Build GRCh38
rs769145799 Chr.1: 235119919-235119919 on Build GRCh38
rs990306000 Chr.1: 235120104-235120104 on Build GRCh38
rs943461556 Chr.1: 235119759-235119759 on Build GRCh38
rs767425517 Chr.1: 235119940-235119940 on Build GRCh38
rs565293122 Chr.1: 235120143-235120143 on Build GRCh38
rs971519257 Chr.1: 235119994-235119994 on Build GRCh38
rs772798878 Chr.1: 235119921-235119921 on Build GRCh38
rs542842432 Chr.1: 235119991-235119991 on Build GRCh38
rs773736293 Chr.1: 235119917-235119920 on Build GRCh38
rs932045303 Chr.1: 235120171-235120171 on Build GRCh38
rs1053903697 Chr.1: 235119804-235119804 on Build GRCh38
rs911031721 Chr.1: 235120172-235120172 on Build GRCh38
rs751862353 Chr.1: 235120052-235120052 on Build GRCh38
rs181942302 Chr.1: 235120166-235120166 on Build GRCh38
rs1005068836 Chr.1: 235119931-235119931 on Build GRCh38
rs528433876 Chr.1: 235120060-235120060 on Build GRCh38
rs747838354 Chr.1: 235119909-235119909 on Build GRCh38
rs1008093079 Chr.1: 235119762-235119762 on Build GRCh38
rs958446875 Chr.1: 235120099-235120099 on Build GRCh38
rs961024265 Chr.1: 235120097-235120097 on Build GRCh38
rs1055717669 Chr.1: 235119943-235119943 on Build GRCh38
rs371642251 Chr.1: 235120197-235120199 on Build GRCh38
rs1005696768 Chr.1: 235119972-235119972 on Build GRCh38
rs540147518 Chr.1: 235120118-235120118 on Build GRCh38
rs749711374 Chr.1: 235119867-235119867 on Build GRCh38
rs1016733907 Chr.1: 235120050-235120050 on Build GRCh38
rs756082321 Chr.1: 235119807-235119807 on Build GRCh38
rs759348179 Chr.1: 235119933-235119933 on Build GRCh38
rs757743564 Chr.1: 235119872-235119872 on Build GRCh38
rs368922585 Chr.1: 235119843-235119843 on Build GRCh38
rs879026547 Chr.1: 235119915-235119915 on Build GRCh38
rs1015158151 Chr.1: 235119938-235119938 on Build GRCh38
rs776206458 Chr.1: 235119908-235119908 on Build GRCh38
rs186156706 Chr.1: 235120176-235120176 on Build GRCh38
rs915223265 Chr.1: 235119828-235119828 on Build GRCh38
rs149626199 Chr.1: 235120201-235120201 on Build GRCh38
rs996574152 Chr.1: 235119880-235119880 on Build GRCh38
rs376342695 Chr.1: 235119920-235119923 on Build GRCh38
rs927026866 Chr.1: 235120071-235120071 on Build GRCh38
rs774489476 Chr.1: 235119927-235119927 on Build GRCh38
rs914216049 Chr.1: 235120108-235120108 on Build GRCh38
rs575524599 Chr.1: 235119787-235119787 on Build GRCh38
rs943868286 Chr.1: 235120162-235120162 on Build GRCh38
rs191669283 Chr.1: 235120157-235120157 on Build GRCh38
rs764522547 Chr.1: 235119967-235119967 on Build GRCh38
rs776089448 Chr.1: 235119890-235119890 on Build GRCh38
rs767839029 Chr.1: 235119792-235119792 on Build GRCh38

View Full Product Details