Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49167831-49168327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf128
Entrez Gene ID
170371
Gene Name
chromosome 10 open reading frame 128
Gene Aliases
-
UniGene
Hs.385493
Gene Chromosome Location
Chr.10: 49154724-49188580 on Build GRCh38
Transcripts
NM_001010863.2 NM_001288740.1 NM_001288741.1 NM_001288742.1 NM_001288743.1 XM_011539378.2 XM_011539379.1 XM_011539380.1 XM_011539382.1 XM_011539383.2 XM_011539384.1 XM_011539385.1 XM_017015792.1 XM_017015793.1 XM_017015794.1 XM_017015795.1 XM_017015796.1 XM_017015797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627516 c.97+10T>A p.? Chr.10: 49167899-49167899 on Build GRCh38
41572 c.44-269G>T p.? Chr.10: 49168231-49168231 on Build GRCh38
4178627 c.80C>A p.T27N Chr.10: 49167926-49167926 on Build GRCh38
3438293 c.29G>A p.G10E Chr.10: 49167953-49167953 on Build GRCh38
7262970 c.63G>A p.V21V Chr.10: 49167943-49167943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780728599 Chr.10: 49167951-49167951 on Build GRCh38
rs140720127 Chr.10: 49168230-49168230 on Build GRCh38
rs373291028 Chr.10: 49168099-49168099 on Build GRCh38
rs370170226 Chr.10: 49168194-49168194 on Build GRCh38
rs779470511 Chr.10: 49167932-49167932 on Build GRCh38
rs756551867 Chr.10: 49168013-49168014 on Build GRCh38
rs775593078 Chr.10: 49168213-49168213 on Build GRCh38
rs963781163 Chr.10: 49167989-49167989 on Build GRCh38
rs115982377 Chr.10: 49168275-49168275 on Build GRCh38
rs370415693 Chr.10: 49167892-49167892 on Build GRCh38
rs1056331019 Chr.10: 49168097-49168097 on Build GRCh38
rs995200598 Chr.10: 49168154-49168154 on Build GRCh38
rs144505356 Chr.10: 49168254-49168254 on Build GRCh38
rs896190020 Chr.10: 49168121-49168121 on Build GRCh38
rs1056659517 Chr.10: 49168293-49168293 on Build GRCh38
rs561521062 Chr.10: 49168163-49168163 on Build GRCh38
rs781219552 Chr.10: 49168150-49168150 on Build GRCh38
rs766224867 Chr.10: 49167900-49167900 on Build GRCh38
rs2466756 Chr.10: 49168061-49168061 on Build GRCh38
rs760839210 Chr.10: 49167868-49167868 on Build GRCh38
rs775504884 Chr.10: 49168001-49168001 on Build GRCh38
rs867336470 Chr.10: 49168017-49168017 on Build GRCh38
rs367970546 Chr.10: 49168297-49168297 on Build GRCh38
rs748456377 Chr.10: 49167968-49167968 on Build GRCh38
rs949344177 Chr.10: 49168063-49168063 on Build GRCh38
rs937937281 Chr.10: 49168288-49168288 on Build GRCh38
rs926532645 Chr.10: 49168283-49168283 on Build GRCh38
rs181348811 Chr.10: 49167997-49167997 on Build GRCh38
rs774255386 Chr.10: 49167904-49167904 on Build GRCh38
rs905044435 Chr.10: 49168080-49168080 on Build GRCh38
rs774096279 Chr.10: 49167981-49167981 on Build GRCh38
rs185040857 Chr.10: 49168071-49168071 on Build GRCh38
rs1003967777 Chr.10: 49168127-49168127 on Build GRCh38
rs1052833097 Chr.10: 49168011-49168011 on Build GRCh38
rs61748312 Chr.10: 49167959-49167959 on Build GRCh38
rs753594857 Chr.10: 49167926-49167926 on Build GRCh38
rs375365683 Chr.10: 49167860-49167860 on Build GRCh38
rs367927493 Chr.10: 49167864-49167864 on Build GRCh38
rs949272745 Chr.10: 49168299-49168299 on Build GRCh38
rs572888993 Chr.10: 49168156-49168156 on Build GRCh38
rs757547465 Chr.10: 49167877-49167878 on Build GRCh38
rs1027891238 Chr.10: 49168155-49168155 on Build GRCh38
rs746332559 Chr.10: 49168010-49168011 on Build GRCh38
rs979567955 Chr.10: 49168211-49168211 on Build GRCh38
rs191162990 Chr.10: 49168199-49168199 on Build GRCh38
rs914001256 Chr.10: 49168260-49168260 on Build GRCh38
rs568354928 Chr.10: 49167950-49167950 on Build GRCh38
rs76233686 Chr.10: 49168045-49168045 on Build GRCh38
rs752103151 Chr.10: 49167920-49167920 on Build GRCh38
rs988263945 Chr.10: 49168235-49168235 on Build GRCh38
rs746346207 Chr.10: 49167937-49167937 on Build GRCh38
rs201588668 Chr.10: 49167931-49167931 on Build GRCh38
rs763719208 Chr.10: 49168012-49168012 on Build GRCh38
rs763589739 Chr.10: 49167923-49167923 on Build GRCh38
rs776710362 Chr.10: 49167960-49167960 on Build GRCh38
rs755665540 Chr.10: 49167922-49167922 on Build GRCh38
rs759351917 Chr.10: 49167986-49167986 on Build GRCh38
rs891379500 Chr.10: 49168184-49168184 on Build GRCh38
rs747502180 Chr.10: 49168113-49168113 on Build GRCh38
rs1018420196 Chr.10: 49168191-49168191 on Build GRCh38
rs372245114 Chr.10: 49167980-49167980 on Build GRCh38
rs548785991 Chr.10: 49167905-49167905 on Build GRCh38
rs146774763 Chr.10: 49168049-49168049 on Build GRCh38
rs541802879 Chr.10: 49168159-49168159 on Build GRCh38
rs781419416 Chr.10: 49167951-49167953 on Build GRCh38
rs116640826 Chr.10: 49168084-49168084 on Build GRCh38
rs114206969 Chr.10: 49168153-49168153 on Build GRCh38
rs757811198 Chr.10: 49168114-49168114 on Build GRCh38
rs979212634 Chr.10: 49168024-49168024 on Build GRCh38
rs753411205 Chr.10: 49168013-49168013 on Build GRCh38
rs776922638 Chr.10: 49167877-49167877 on Build GRCh38
rs747001760 Chr.10: 49167956-49167956 on Build GRCh38
rs530412992 Chr.10: 49168268-49168268 on Build GRCh38
rs183784773 Chr.10: 49168237-49168237 on Build GRCh38
rs1008518006 Chr.10: 49168187-49168187 on Build GRCh38
rs911962089 Chr.10: 49167978-49167978 on Build GRCh38
rs962746135 Chr.10: 49167924-49167924 on Build GRCh38
rs760101265 Chr.10: 49168002-49168002 on Build GRCh38
rs768710483 Chr.10: 49167875-49167875 on Build GRCh38
rs924901422 Chr.10: 49168030-49168030 on Build GRCh38

View Full Product Details