Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110577433-110577933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMC3
Entrez Gene ID
9126
Gene Name
structural maintenance of chromosomes 3
Gene Aliases
BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1
UniGene
Hs.24485
Gene Chromosome Location
Chr.10: 110567691-110604634 on Build GRCh38
Transcripts
NM_005445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4728397 c.312_313insA p.D107fs*14 Chr.10: 110577876-110577877 on Build GRCh38
1345759 c.273C>G p.I91M Chr.10: 110577837-110577837 on Build GRCh38
914737 c.295C>T p.R99* Chr.10: 110577859-110577859 on Build GRCh38
6493939 c.255A>G p.S85S Chr.10: 110577477-110577477 on Build GRCh38
6785204 c.273C>T p.I91I Chr.10: 110577837-110577837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037035506 Chr.10: 110577606-110577606 on Build GRCh38
rs918816216 Chr.10: 110577608-110577608 on Build GRCh38
rs869312672 Chr.10: 110577847-110577847 on Build GRCh38
rs753775816 Chr.10: 110577532-110577533 on Build GRCh38
rs370229242 Chr.10: 110577518-110577518 on Build GRCh38
rs776155546 Chr.10: 110577503-110577503 on Build GRCh38
rs767708203 Chr.10: 110577792-110577792 on Build GRCh38
rs375382120 Chr.10: 110577537-110577537 on Build GRCh38
rs1010886727 Chr.10: 110577696-110577696 on Build GRCh38
rs185589534 Chr.10: 110577823-110577823 on Build GRCh38
rs139965823 Chr.10: 110577750-110577750 on Build GRCh38
rs777346320 Chr.10: 110577736-110577736 on Build GRCh38
rs1054851085 Chr.10: 110577670-110577671 on Build GRCh38
rs774721996 Chr.10: 110577899-110577899 on Build GRCh38
rs750122724 Chr.10: 110577500-110577501 on Build GRCh38
rs1032862100 Chr.10: 110577653-110577653 on Build GRCh38
rs548619486 Chr.10: 110577849-110577849 on Build GRCh38
rs554699066 Chr.10: 110577589-110577589 on Build GRCh38
rs754582099 Chr.10: 110577543-110577544 on Build GRCh38
rs769145720 Chr.10: 110577850-110577850 on Build GRCh38
rs1045916758 Chr.10: 110577637-110577637 on Build GRCh38
rs902976205 Chr.10: 110577591-110577591 on Build GRCh38
rs368638452 Chr.10: 110577809-110577809 on Build GRCh38
rs941335276 Chr.10: 110577603-110577603 on Build GRCh38
rs199571484 Chr.10: 110577820-110577820 on Build GRCh38
rs949832024 Chr.10: 110577573-110577573 on Build GRCh38
rs777587178 Chr.10: 110577818-110577818 on Build GRCh38
rs893043270 Chr.10: 110577695-110577695 on Build GRCh38
rs371662516 Chr.10: 110577578-110577578 on Build GRCh38
rs759329645 Chr.10: 110577505-110577505 on Build GRCh38
rs985705136 Chr.10: 110577749-110577749 on Build GRCh38
rs962803242 Chr.10: 110577790-110577790 on Build GRCh38
rs1029905970 Chr.10: 110577708-110577708 on Build GRCh38
rs893549460 Chr.10: 110577694-110577694 on Build GRCh38
rs972210892 Chr.10: 110577773-110577773 on Build GRCh38
rs80217102 Chr.10: 110577628-110577628 on Build GRCh38
rs566007623 Chr.10: 110577519-110577519 on Build GRCh38
rs778555876 Chr.10: 110577810-110577811 on Build GRCh38
rs966587584 Chr.10: 110577699-110577699 on Build GRCh38
rs761174946 Chr.10: 110577618-110577622 on Build GRCh38
rs779815452 Chr.10: 110577501-110577502 on Build GRCh38
rs555918922 Chr.10: 110577807-110577807 on Build GRCh38
rs770562769 Chr.10: 110577486-110577486 on Build GRCh38
rs765519841 Chr.10: 110577813-110577813 on Build GRCh38
rs756099489 Chr.10: 110577502-110577502 on Build GRCh38
rs139894626 Chr.10: 110577906-110577906 on Build GRCh38
rs894263556 Chr.10: 110577568-110577568 on Build GRCh38
rs199936534 Chr.10: 110577840-110577840 on Build GRCh38
rs757295221 Chr.10: 110577791-110577791 on Build GRCh38
rs769614864 Chr.10: 110577837-110577837 on Build GRCh38
rs962173714 Chr.10: 110577771-110577771 on Build GRCh38
rs953524882 Chr.10: 110577702-110577702 on Build GRCh38
rs751605629 Chr.10: 110577789-110577789 on Build GRCh38
rs745497065 Chr.10: 110577831-110577831 on Build GRCh38
rs763526476 Chr.10: 110577536-110577536 on Build GRCh38
rs1029408260 Chr.10: 110577700-110577700 on Build GRCh38
rs192690383 Chr.10: 110577811-110577811 on Build GRCh38
rs146433240 Chr.10: 110577477-110577477 on Build GRCh38
rs778656629 Chr.10: 110577799-110577799 on Build GRCh38
rs909865856 Chr.10: 110577597-110577597 on Build GRCh38
rs906073593 Chr.10: 110577639-110577639 on Build GRCh38
rs756753774 Chr.10: 110577829-110577829 on Build GRCh38
rs750635536 Chr.10: 110577796-110577796 on Build GRCh38

View Full Product Details