Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112149722-112150256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAM
Entrez Gene ID
57678
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Gene Aliases
GPAT, GPAT1
UniGene
Hs.42586
Gene Chromosome Location
Chr.10: 112149864-112215395 on Build GRCh38
Transcripts
NM_001244949.1 NM_020918.5 XM_005269998.1

SNP Detail

SNP ID Chromosome Location
rs911188814 Chr.10: 112150121-112150121 on Build GRCh38
rs190479185 Chr.10: 112149778-112149778 on Build GRCh38
rs1038561963 Chr.10: 112150133-112150133 on Build GRCh38
rs978581918 Chr.10: 112150036-112150036 on Build GRCh38
rs994372630 Chr.10: 112150210-112150210 on Build GRCh38
rs369650935 Chr.10: 112149811-112149811 on Build GRCh38
rs540081581 Chr.10: 112150158-112150158 on Build GRCh38
rs533484707 Chr.10: 112149978-112149978 on Build GRCh38
rs989998168 Chr.10: 112149961-112149961 on Build GRCh38
rs1038657817 Chr.10: 112149802-112149802 on Build GRCh38
rs768084330 Chr.10: 112150201-112150201 on Build GRCh38
rs762656861 Chr.10: 112149924-112149924 on Build GRCh38
rs926737772 Chr.10: 112150138-112150138 on Build GRCh38
rs544574670 Chr.10: 112149922-112149922 on Build GRCh38
rs1014461348 Chr.10: 112149934-112149934 on Build GRCh38
rs963936075 Chr.10: 112150056-112150056 on Build GRCh38
rs562879042 Chr.10: 112149970-112149970 on Build GRCh38
rs905462301 Chr.10: 112149804-112149804 on Build GRCh38
rs138008636 Chr.10: 112150016-112150016 on Build GRCh38
rs182716782 Chr.10: 112149807-112149807 on Build GRCh38
rs188294837 Chr.10: 112150176-112150176 on Build GRCh38
rs80029816 Chr.10: 112150104-112150104 on Build GRCh38
rs1032776912 Chr.10: 112150049-112150049 on Build GRCh38
rs551878420 Chr.10: 112150015-112150015 on Build GRCh38
rs1020027496 Chr.10: 112150000-112150000 on Build GRCh38
rs543292673 Chr.10: 112149818-112149818 on Build GRCh38
rs895445659 Chr.10: 112149928-112149928 on Build GRCh38
rs759235454 Chr.10: 112149957-112149957 on Build GRCh38
rs1034394045 Chr.10: 112149917-112149917 on Build GRCh38
rs902961855 Chr.10: 112150231-112150231 on Build GRCh38
rs546958313 Chr.10: 112150132-112150132 on Build GRCh38
rs933998269 Chr.10: 112150097-112150097 on Build GRCh38
rs898747979 Chr.10: 112150187-112150187 on Build GRCh38
rs374082485 Chr.10: 112150089-112150089 on Build GRCh38
rs371598317 Chr.10: 112150157-112150157 on Build GRCh38
rs775293412 Chr.10: 112150111-112150111 on Build GRCh38
rs922632684 Chr.10: 112150088-112150088 on Build GRCh38
rs192577378 Chr.10: 112150230-112150230 on Build GRCh38
rs796813824 Chr.10: 112150181-112150181 on Build GRCh38
rs2792740 Chr.10: 112149760-112149760 on Build GRCh38
rs374509110 Chr.10: 112150216-112150216 on Build GRCh38
rs938268744 Chr.10: 112150149-112150149 on Build GRCh38
rs35347280 Chr.10: 112150075-112150076 on Build GRCh38
rs975316532 Chr.10: 112150059-112150059 on Build GRCh38
rs535137558 Chr.10: 112150172-112150172 on Build GRCh38
rs906956619 Chr.10: 112150214-112150214 on Build GRCh38
rs1034439915 Chr.10: 112149813-112149813 on Build GRCh38

View Full Product Details