Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112374075-112374581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL5
Entrez Gene ID
51703
Gene Name
acyl-CoA synthetase long-chain family member 5
Gene Aliases
ACS2, ACS5, FACL5
UniGene
Hs.11638
Gene Chromosome Location
Chr.10: 112374158-112428380 on Build GRCh38
Transcripts
NM_203379.1

SNP Detail

SNP ID Chromosome Location
rs562225884 Chr.10: 112374182-112374182 on Build GRCh38
rs550424207 Chr.10: 112374422-112374422 on Build GRCh38
rs533854196 Chr.10: 112374106-112374106 on Build GRCh38
rs993108936 Chr.10: 112374314-112374314 on Build GRCh38
rs150992361 Chr.10: 112374398-112374398 on Build GRCh38
rs568847648 Chr.10: 112374135-112374135 on Build GRCh38
rs143590592 Chr.10: 112374372-112374372 on Build GRCh38
rs1003021087 Chr.10: 112374228-112374228 on Build GRCh38
rs757889284 Chr.10: 112374450-112374450 on Build GRCh38
rs563600088 Chr.10: 112374444-112374444 on Build GRCh38
rs565940375 Chr.10: 112374505-112374505 on Build GRCh38
rs1009343991 Chr.10: 112374298-112374298 on Build GRCh38
rs1027399327 Chr.10: 112374262-112374262 on Build GRCh38
rs1015817877 Chr.10: 112374300-112374300 on Build GRCh38
rs1000540484 Chr.10: 112374404-112374404 on Build GRCh38
rs527940943 Chr.10: 112374280-112374280 on Build GRCh38
rs772122796 Chr.10: 112374475-112374475 on Build GRCh38
rs970086775 Chr.10: 112374365-112374365 on Build GRCh38
rs1024304268 Chr.10: 112374334-112374334 on Build GRCh38
rs188726597 Chr.10: 112374455-112374455 on Build GRCh38
rs909259250 Chr.10: 112374183-112374183 on Build GRCh38
rs961911630 Chr.10: 112374304-112374304 on Build GRCh38
rs926040236 Chr.10: 112374420-112374420 on Build GRCh38
rs1017480501 Chr.10: 112374442-112374442 on Build GRCh38
rs950781980 Chr.10: 112374487-112374487 on Build GRCh38
rs910375428 Chr.10: 112374119-112374119 on Build GRCh38
rs1044593171 Chr.10: 112374224-112374224 on Build GRCh38
rs925807969 Chr.10: 112374116-112374116 on Build GRCh38
rs544981520 Chr.10: 112374212-112374212 on Build GRCh38
rs41306862 Chr.10: 112374399-112374399 on Build GRCh38
rs906112346 Chr.10: 112374225-112374225 on Build GRCh38
rs896224194 Chr.10: 112374245-112374245 on Build GRCh38
rs140810292 Chr.10: 112374471-112374471 on Build GRCh38
rs779426187 Chr.10: 112374453-112374453 on Build GRCh38
rs929392481 Chr.10: 112374172-112374172 on Build GRCh38
rs141485954 Chr.10: 112374237-112374237 on Build GRCh38
rs888582363 Chr.10: 112374287-112374287 on Build GRCh38
rs775587237 Chr.10: 112374480-112374480 on Build GRCh38
rs543867881 Chr.10: 112374175-112374175 on Build GRCh38
rs573518865 Chr.10: 112374168-112374168 on Build GRCh38
rs374451920 Chr.10: 112374458-112374458 on Build GRCh38
rs571765407 Chr.10: 112374434-112374434 on Build GRCh38
rs778349600 Chr.10: 112374142-112374142 on Build GRCh38
rs1006141509 Chr.10: 112374299-112374299 on Build GRCh38
rs181790765 Chr.10: 112374522-112374522 on Build GRCh38
rs964549286 Chr.10: 112374134-112374134 on Build GRCh38
rs375224929 Chr.10: 112374195-112374195 on Build GRCh38
rs3808920 Chr.10: 112374517-112374517 on Build GRCh38
rs573083249 Chr.10: 112374105-112374105 on Build GRCh38
rs555093984 Chr.10: 112374126-112374126 on Build GRCh38
rs866652820 Chr.10: 112374180-112374180 on Build GRCh38
rs553597099 Chr.10: 112374102-112374102 on Build GRCh38

View Full Product Details