Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113726136-113726503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
UniGene
Hs.9216
Gene Chromosome Location
Chr.10: 113679162-113730909 on Build GRCh38
Transcripts
NM_001227.4 NM_001267056.1 NM_001267057.1 NM_001267058.1 NM_001320911.1 NM_033338.5 NM_033339.4 NM_033340.3 XM_006718017.3 XM_011540260.1 XM_017016763.1 XM_017016764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6564413 c.781+5C>G p.? Chr.10: 113726439-113726439 on Build GRCh38
7192728 c.686A>C p.Q229P Chr.10: 113726339-113726339 on Build GRCh38
683268 c.663G>C p.G221G Chr.10: 113726316-113726316 on Build GRCh38
683267 c.725C>A p.P242H Chr.10: 113726378-113726378 on Build GRCh38
1474362 c.770C>T p.S257F Chr.10: 113726423-113726423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187106799 Chr.10: 113726320-113726320 on Build GRCh38
rs527513511 Chr.10: 113726232-113726232 on Build GRCh38
rs370774542 Chr.10: 113726340-113726340 on Build GRCh38
rs930939327 Chr.10: 113726394-113726394 on Build GRCh38
rs1047005335 Chr.10: 113726219-113726219 on Build GRCh38
rs564423703 Chr.10: 113726319-113726319 on Build GRCh38
rs748534536 Chr.10: 113726411-113726411 on Build GRCh38
rs981659602 Chr.10: 113726239-113726239 on Build GRCh38
rs777072624 Chr.10: 113726170-113726170 on Build GRCh38
rs375409071 Chr.10: 113726348-113726348 on Build GRCh38
rs769035146 Chr.10: 113726400-113726400 on Build GRCh38
rs761866035 Chr.10: 113726181-113726181 on Build GRCh38
rs552479055 Chr.10: 113726306-113726306 on Build GRCh38
rs773227567 Chr.10: 113726343-113726343 on Build GRCh38
rs745389318 Chr.10: 113726305-113726305 on Build GRCh38
rs758136964 Chr.10: 113726276-113726276 on Build GRCh38
rs760108863 Chr.10: 113726171-113726174 on Build GRCh38
rs775679434 Chr.10: 113726432-113726446 on Build GRCh38
rs748959752 Chr.10: 113726386-113726386 on Build GRCh38
rs767651924 Chr.10: 113726195-113726195 on Build GRCh38
rs542416904 Chr.10: 113726212-113726212 on Build GRCh38
rs755644671 Chr.10: 113726472-113726472 on Build GRCh38
rs774244219 Chr.10: 113726442-113726442 on Build GRCh38
rs970129319 Chr.10: 113726166-113726166 on Build GRCh38
rs749886205 Chr.10: 113726364-113726364 on Build GRCh38
rs765514741 Chr.10: 113726434-113726434 on Build GRCh38
rs781305492 Chr.10: 113726360-113726360 on Build GRCh38
rs116560793 Chr.10: 113726420-113726420 on Build GRCh38
rs751746516 Chr.10: 113726466-113726466 on Build GRCh38
rs938507851 Chr.10: 113726431-113726431 on Build GRCh38
rs755592768 Chr.10: 113726372-113726372 on Build GRCh38
rs376949404 Chr.10: 113726299-113726299 on Build GRCh38
rs752500323 Chr.10: 113726272-113726272 on Build GRCh38
rs535164025 Chr.10: 113726407-113726407 on Build GRCh38
rs367940290 Chr.10: 113726453-113726453 on Build GRCh38
rs146952079 Chr.10: 113726349-113726349 on Build GRCh38
rs367682197 Chr.10: 113726356-113726356 on Build GRCh38
rs182108910 Chr.10: 113726269-113726269 on Build GRCh38
rs886948162 Chr.10: 113726221-113726221 on Build GRCh38
rs766559562 Chr.10: 113726345-113726345 on Build GRCh38
rs137956734 Chr.10: 113726432-113726432 on Build GRCh38
rs376348175 Chr.10: 113726312-113726312 on Build GRCh38
rs768958115 Chr.10: 113726314-113726314 on Build GRCh38
rs774732145 Chr.10: 113726170-113726170 on Build GRCh38
rs749523025 Chr.10: 113726254-113726254 on Build GRCh38
rs780894394 Chr.10: 113726304-113726304 on Build GRCh38
rs765459466 Chr.10: 113726423-113726428 on Build GRCh38
rs370400085 Chr.10: 113726381-113726381 on Build GRCh38
rs760778461 Chr.10: 113726344-113726344 on Build GRCh38
rs751610681 Chr.10: 113726298-113726298 on Build GRCh38
rs772529292 Chr.10: 113726415-113726415 on Build GRCh38
rs767503355 Chr.10: 113726454-113726454 on Build GRCh38
rs370692712 Chr.10: 113726285-113726285 on Build GRCh38
rs989486434 Chr.10: 113726294-113726294 on Build GRCh38
rs375002353 Chr.10: 113726476-113726476 on Build GRCh38
rs373690332 Chr.10: 113726311-113726311 on Build GRCh38
rs772117398 Chr.10: 113726337-113726337 on Build GRCh38
rs942487002 Chr.10: 113726379-113726379 on Build GRCh38
rs754538524 Chr.10: 113726477-113726477 on Build GRCh38
rs763085086 Chr.10: 113726449-113726460 on Build GRCh38
rs373359750 Chr.10: 113726255-113726255 on Build GRCh38
rs757877976 Chr.10: 113726277-113726277 on Build GRCh38
rs770977304 Chr.10: 113726427-113726427 on Build GRCh38
rs757208505 Chr.10: 113726359-113726359 on Build GRCh38
rs200665202 Chr.10: 113726426-113726426 on Build GRCh38
rs201274135 Chr.10: 113726475-113726475 on Build GRCh38
rs372483238 Chr.10: 113726456-113726456 on Build GRCh38
rs1006266981 Chr.10: 113726463-113726463 on Build GRCh38
rs764244830 Chr.10: 113726463-113726491 on Build GRCh38

View Full Product Details