Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113897734-113898255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLRC2
Entrez Gene ID
374354
Gene Name
NHL repeat containing 2
Gene Aliases
-
UniGene
Hs.369924
Gene Chromosome Location
Chr.10: 113854632-113912506 on Build GRCh38
Transcripts
NM_198514.3 XM_011539769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1321528 c.1122C>G p.G374G Chr.10: 113898192-113898192 on Build GRCh38
1345905 c.1134G>T p.K378N Chr.10: 113898204-113898204 on Build GRCh38
2053800 c.1116C>T p.D372D Chr.10: 113898186-113898186 on Build GRCh38
1578432 c.1086_1087insC p.T363fs*19 Chr.10: 113898156-113898157 on Build GRCh38
7317560 c.1093C>T p.Q365* Chr.10: 113898163-113898163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184029943 Chr.10: 113897845-113897845 on Build GRCh38
rs945570808 Chr.10: 113898012-113898012 on Build GRCh38
rs974137180 Chr.10: 113897842-113897842 on Build GRCh38
rs567554364 Chr.10: 113898123-113898123 on Build GRCh38
rs962227561 Chr.10: 113898215-113898215 on Build GRCh38
rs965712583 Chr.10: 113898057-113898057 on Build GRCh38
rs748733226 Chr.10: 113897900-113897902 on Build GRCh38
rs938282044 Chr.10: 113897861-113897861 on Build GRCh38
rs751910927 Chr.10: 113898223-113898223 on Build GRCh38
rs541208725 Chr.10: 113897960-113897960 on Build GRCh38
rs372561400 Chr.10: 113897905-113897905 on Build GRCh38
rs764855698 Chr.10: 113898069-113898069 on Build GRCh38
rs771802806 Chr.10: 113898142-113898142 on Build GRCh38
rs376794338 Chr.10: 113897929-113897930 on Build GRCh38
rs555422695 Chr.10: 113897928-113897929 on Build GRCh38
rs778311012 Chr.10: 113898095-113898095 on Build GRCh38
rs760625750 Chr.10: 113898157-113898157 on Build GRCh38
rs776191573 Chr.10: 113898174-113898174 on Build GRCh38
rs984960659 Chr.10: 113897838-113897838 on Build GRCh38
rs530008422 Chr.10: 113898070-113898070 on Build GRCh38
rs564942042 Chr.10: 113897923-113897923 on Build GRCh38
rs1010091440 Chr.10: 113898027-113898027 on Build GRCh38
rs144245843 Chr.10: 113897790-113897791 on Build GRCh38
rs903952414 Chr.10: 113897992-113897992 on Build GRCh38
rs746006326 Chr.10: 113898126-113898126 on Build GRCh38
rs940914112 Chr.10: 113897883-113897883 on Build GRCh38
rs1000484103 Chr.10: 113898033-113898033 on Build GRCh38
rs764842811 Chr.10: 113898200-113898200 on Build GRCh38
rs199504264 Chr.10: 113898096-113898096 on Build GRCh38
rs926867777 Chr.10: 113897844-113897844 on Build GRCh38
rs908652196 Chr.10: 113897809-113897809 on Build GRCh38
rs752355939 Chr.10: 113898217-113898217 on Build GRCh38
rs982883954 Chr.10: 113897805-113897805 on Build GRCh38
rs770046904 Chr.10: 113898128-113898128 on Build GRCh38
rs749519822 Chr.10: 113898138-113898138 on Build GRCh38
rs918108496 Chr.10: 113897914-113897914 on Build GRCh38
rs1019309875 Chr.10: 113898036-113898036 on Build GRCh38
rs192288108 Chr.10: 113897786-113897786 on Build GRCh38
rs140324697 Chr.10: 113897953-113897953 on Build GRCh38
rs776871860 Chr.10: 113897908-113897908 on Build GRCh38
rs1042106402 Chr.10: 113898013-113898013 on Build GRCh38
rs187768865 Chr.10: 113898022-113898022 on Build GRCh38
rs532371956 Chr.10: 113897939-113897939 on Build GRCh38
rs868102668 Chr.10: 113897761-113897761 on Build GRCh38
rs770836331 Chr.10: 113898173-113898173 on Build GRCh38
rs757754711 Chr.10: 113898108-113898108 on Build GRCh38
rs773037258 Chr.10: 113898151-113898151 on Build GRCh38
rs974508212 Chr.10: 113897784-113897784 on Build GRCh38
rs948078253 Chr.10: 113897981-113897981 on Build GRCh38
rs762719104 Chr.10: 113898222-113898222 on Build GRCh38
rs999943487 Chr.10: 113898020-113898020 on Build GRCh38
rs771480117 Chr.10: 113898208-113898211 on Build GRCh38
rs1045124866 Chr.10: 113897984-113897984 on Build GRCh38
rs140827983 Chr.10: 113898185-113898185 on Build GRCh38
rs1057157759 Chr.10: 113897897-113897897 on Build GRCh38
rs372119140 Chr.10: 113898079-113898079 on Build GRCh38
rs962691647 Chr.10: 113897818-113897818 on Build GRCh38

View Full Product Details