Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114278615-114279114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA2
Entrez Gene ID
340706
Gene Name
von Willebrand factor A domain containing 2
Gene Aliases
AMACO, CCSP-2, NET42
UniGene
Hs.197741
Gene Chromosome Location
Chr.10: 114239254-114294500 on Build GRCh38
Transcripts
NM_001272046.1 NM_001320804.1 XM_011539754.2 XM_011539757.2 XM_017016177.1 XM_017016178.1 XM_017016179.1 XM_017016181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4509624 c.812C>T p.A271V Chr.10: 114278830-114278830 on Build GRCh38
5359981 c.734G>T p.R245M Chr.10: 114278752-114278752 on Build GRCh38
6385602 c.737C>A p.T246K Chr.10: 114278755-114278755 on Build GRCh38
5920027 c.805G>A p.V269M Chr.10: 114278823-114278823 on Build GRCh38
4509623 c.812C>T p.A271V Chr.10: 114278830-114278830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766885689 Chr.10: 114278669-114278669 on Build GRCh38
rs759976715 Chr.10: 114278682-114278682 on Build GRCh38
rs145948639 Chr.10: 114278727-114278727 on Build GRCh38
rs767346219 Chr.10: 114278913-114278913 on Build GRCh38
rs187730689 Chr.10: 114278945-114278945 on Build GRCh38
rs373000451 Chr.10: 114278792-114278792 on Build GRCh38
rs1022961180 Chr.10: 114278954-114278954 on Build GRCh38
rs929494654 Chr.10: 114278902-114278902 on Build GRCh38
rs139461527 Chr.10: 114278748-114278748 on Build GRCh38
rs374045265 Chr.10: 114278863-114278863 on Build GRCh38
rs201031318 Chr.10: 114278679-114278679 on Build GRCh38
rs573108867 Chr.10: 114278915-114278915 on Build GRCh38
rs759887745 Chr.10: 114278689-114278690 on Build GRCh38
rs879917038 Chr.10: 114279026-114279026 on Build GRCh38
rs530138563 Chr.10: 114278726-114278726 on Build GRCh38
rs201135176 Chr.10: 114278791-114278791 on Build GRCh38
rs943814358 Chr.10: 114278757-114278757 on Build GRCh38
rs770477974 Chr.10: 114278832-114278832 on Build GRCh38
rs751588409 Chr.10: 114278764-114278764 on Build GRCh38
rs902059449 Chr.10: 114278774-114278774 on Build GRCh38
rs887754331 Chr.10: 114278779-114278779 on Build GRCh38
rs570723813 Chr.10: 114278821-114278821 on Build GRCh38
rs200441458 Chr.10: 114278875-114278875 on Build GRCh38
rs765739365 Chr.10: 114278695-114278696 on Build GRCh38
rs751817233 Chr.10: 114278692-114278692 on Build GRCh38
rs905859093 Chr.10: 114278987-114278987 on Build GRCh38
rs147176000 Chr.10: 114278941-114278941 on Build GRCh38
rs202058880 Chr.10: 114278811-114278811 on Build GRCh38
rs534395472 Chr.10: 114278838-114278838 on Build GRCh38
rs781407990 Chr.10: 114278766-114278766 on Build GRCh38
rs996276230 Chr.10: 114278910-114278910 on Build GRCh38
rs149756313 Chr.10: 114278755-114278755 on Build GRCh38
rs532624503 Chr.10: 114278645-114278645 on Build GRCh38
rs776867992 Chr.10: 114278749-114278749 on Build GRCh38
rs754852186 Chr.10: 114278770-114278770 on Build GRCh38
rs749052874 Chr.10: 114278716-114278716 on Build GRCh38
rs777441113 Chr.10: 114278778-114278778 on Build GRCh38
rs757430317 Chr.10: 114278709-114278709 on Build GRCh38
rs760429949 Chr.10: 114278743-114278743 on Build GRCh38
rs368890749 Chr.10: 114278729-114278729 on Build GRCh38
rs765711688 Chr.10: 114278685-114278685 on Build GRCh38
rs539474144 Chr.10: 114278825-114278825 on Build GRCh38
rs746456782 Chr.10: 114278781-114278781 on Build GRCh38
rs927112610 Chr.10: 114279018-114279018 on Build GRCh38
rs116145566 Chr.10: 114278759-114278759 on Build GRCh38
rs374291780 Chr.10: 114278730-114278730 on Build GRCh38
rs374215852 Chr.10: 114278808-114278808 on Build GRCh38
rs770001461 Chr.10: 114278704-114278704 on Build GRCh38
rs949968031 Chr.10: 114278971-114278971 on Build GRCh38
rs1036833527 Chr.10: 114278737-114278737 on Build GRCh38
rs952097605 Chr.10: 114278934-114278934 on Build GRCh38
rs749587500 Chr.10: 114278871-114278871 on Build GRCh38
rs966113332 Chr.10: 114278653-114278653 on Build GRCh38
rs547927919 Chr.10: 114278654-114278654 on Build GRCh38
rs779735936 Chr.10: 114278710-114278710 on Build GRCh38
rs1035791463 Chr.10: 114278990-114278990 on Build GRCh38
rs116143407 Chr.10: 114278912-114278912 on Build GRCh38
rs778375156 Chr.10: 114278916-114278916 on Build GRCh38
rs377414825 Chr.10: 114278861-114278861 on Build GRCh38
rs748128724 Chr.10: 114278773-114278773 on Build GRCh38
rs968658346 Chr.10: 114279072-114279072 on Build GRCh38
rs930633142 Chr.10: 114278722-114278722 on Build GRCh38
rs979905460 Chr.10: 114278985-114278985 on Build GRCh38
rs145257901 Chr.10: 114278777-114278777 on Build GRCh38
rs113098480 Chr.10: 114278831-114278831 on Build GRCh38
rs183338632 Chr.10: 114278862-114278862 on Build GRCh38
rs992203334 Chr.10: 114278947-114278947 on Build GRCh38
rs757483599 Chr.10: 114278765-114278765 on Build GRCh38
rs377744287 Chr.10: 114278785-114278785 on Build GRCh38
rs777811441 Chr.10: 114278856-114278864 on Build GRCh38
rs750698718 Chr.10: 114278822-114278822 on Build GRCh38
rs767669172 Chr.10: 114278880-114278880 on Build GRCh38
rs201268001 Chr.10: 114278874-114278874 on Build GRCh38
rs140014450 Chr.10: 114278809-114278809 on Build GRCh38
rs758535028 Chr.10: 114278833-114278834 on Build GRCh38
rs34900602 Chr.10: 114278908-114278908 on Build GRCh38
rs770899290 Chr.10: 114278839-114278839 on Build GRCh38
rs988964382 Chr.10: 114278633-114278633 on Build GRCh38
rs141503948 Chr.10: 114278756-114278756 on Build GRCh38
rs115765026 Chr.10: 114278905-114278905 on Build GRCh38
rs765503243 Chr.10: 114278754-114278754 on Build GRCh38
rs147605334 Chr.10: 114278806-114278806 on Build GRCh38
rs754186452 Chr.10: 114278672-114278672 on Build GRCh38
rs760008558 Chr.10: 114278753-114278753 on Build GRCh38
rs559472420 Chr.10: 114278690-114278690 on Build GRCh38
rs773118145 Chr.10: 114278894-114278894 on Build GRCh38
rs935856909 Chr.10: 114279075-114279075 on Build GRCh38
rs750598220 Chr.10: 114278768-114278768 on Build GRCh38
rs762195922 Chr.10: 114278877-114278877 on Build GRCh38
rs753199719 Chr.10: 114278725-114278735 on Build GRCh38
rs548525178 Chr.10: 114278961-114278961 on Build GRCh38
rs1002923709 Chr.10: 114278989-114278989 on Build GRCh38
rs535474178 Chr.10: 114278864-114278864 on Build GRCh38
rs1019601154 Chr.10: 114278889-114278889 on Build GRCh38
rs769280907 Chr.10: 114278865-114278865 on Build GRCh38
rs753137348 Chr.10: 114278687-114278687 on Build GRCh38
rs988626363 Chr.10: 114279057-114279057 on Build GRCh38
rs985819895 Chr.10: 114279089-114279089 on Build GRCh38
rs530035085 Chr.10: 114278955-114278955 on Build GRCh38
rs775843870 Chr.10: 114278867-114278867 on Build GRCh38
rs760968004 Chr.10: 114278899-114278899 on Build GRCh38
rs1021497891 Chr.10: 114279059-114279059 on Build GRCh38
rs758419039 Chr.10: 114278851-114278851 on Build GRCh38
rs145424503 Chr.10: 114278812-114278812 on Build GRCh38
rs768481637 Chr.10: 114278720-114278720 on Build GRCh38
rs370595351 Chr.10: 114278798-114278798 on Build GRCh38
rs752484981 Chr.10: 114278843-114278843 on Build GRCh38
rs377381530 Chr.10: 114278724-114278724 on Build GRCh38
rs563537579 Chr.10: 114278979-114278979 on Build GRCh38
rs955835814 Chr.10: 114279008-114279008 on Build GRCh38
rs533567863 Chr.10: 114279078-114279078 on Build GRCh38
rs984844898 Chr.10: 114278940-114278940 on Build GRCh38

View Full Product Details