Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8694063-8694572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST5
Entrez Gene ID
6764
Gene Name
suppression of tumorigenicity 5
Gene Aliases
DENND2B, HTS1, p126
UniGene
Hs.117715
Gene Chromosome Location
Chr.11: 8693352-8910951 on Build GRCh38
Transcripts
NM_005418.3 NM_139157.2 NM_213618.1 XM_005253077.1 XM_005253083.2 XM_011520309.1 XM_011520310.1 XM_011520311.2 XM_011520312.1 XM_011520313.1 XM_011520314.1 XM_011520315.1 XM_011520316.1 XM_011520317.1 XM_011520318.1 XM_011520319.2 XM_011520323.2 XM_011520324.2 XM_011520325.1 XM_011520326.1 XM_011520327.1 XM_011520328.1 XM_011520329.1 XM_017018182.1 XM_017018183.1 XM_017018184.1 XM_017018185.1 XM_017018186.1 XM_017018187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4037263 c.3387A>C p.K1129N Chr.11: 8694123-8694123 on Build GRCh38
932787 c.3380G>A p.G1127D Chr.11: 8694130-8694130 on Build GRCh38
4037262 c.3397C>T p.L1133F Chr.11: 8694113-8694113 on Build GRCh38
4574869 c.3410A>G p.N1137S Chr.11: 8694100-8694100 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs575557039 Chr.11: 8694455-8694455 on Build GRCh38
rs79855289 Chr.11: 8694532-8694532 on Build GRCh38
rs763214639 Chr.11: 8694149-8694149 on Build GRCh38
rs965242237 Chr.11: 8694306-8694306 on Build GRCh38
rs768011310 Chr.11: 8694166-8694166 on Build GRCh38
rs916279489 Chr.11: 8694340-8694340 on Build GRCh38
rs180840788 Chr.11: 8694319-8694319 on Build GRCh38
rs938816683 Chr.11: 8694213-8694213 on Build GRCh38
rs914671546 Chr.11: 8694146-8694146 on Build GRCh38
rs534456262 Chr.11: 8694360-8694360 on Build GRCh38
rs148782617 Chr.11: 8694145-8694145 on Build GRCh38
rs753168276 Chr.11: 8694170-8694170 on Build GRCh38
rs768448905 Chr.11: 8694135-8694145 on Build GRCh38
rs766437257 Chr.11: 8694155-8694155 on Build GRCh38
rs1019508440 Chr.11: 8694394-8694394 on Build GRCh38
rs11042042 Chr.11: 8694332-8694332 on Build GRCh38
rs78115107 Chr.11: 8694345-8694345 on Build GRCh38
rs766869966 Chr.11: 8694180-8694182 on Build GRCh38
rs1043320915 Chr.11: 8694484-8694484 on Build GRCh38
rs757601515 Chr.11: 8694450-8694450 on Build GRCh38
rs763739987 Chr.11: 8694140-8694141 on Build GRCh38
rs768704024 Chr.11: 8694100-8694100 on Build GRCh38
rs761608713 Chr.11: 8694137-8694137 on Build GRCh38
rs751930601 Chr.11: 8694402-8694402 on Build GRCh38
rs190056409 Chr.11: 8694392-8694392 on Build GRCh38
rs182641643 Chr.11: 8694403-8694403 on Build GRCh38
rs571392424 Chr.11: 8694348-8694349 on Build GRCh38
rs756519280 Chr.11: 8694192-8694194 on Build GRCh38
rs746928533 Chr.11: 8694092-8694092 on Build GRCh38
rs1019454766 Chr.11: 8694102-8694102 on Build GRCh38
rs780726622 Chr.11: 8694360-8694360 on Build GRCh38
rs546289207 Chr.11: 8694108-8694108 on Build GRCh38
rs200977713 Chr.11: 8694143-8694143 on Build GRCh38
rs776434751 Chr.11: 8694130-8694130 on Build GRCh38
rs886953324 Chr.11: 8694373-8694373 on Build GRCh38
rs756739564 Chr.11: 8694149-8694150 on Build GRCh38
rs751707216 Chr.11: 8694162-8694162 on Build GRCh38
rs373146631 Chr.11: 8694293-8694293 on Build GRCh38
rs1045167250 Chr.11: 8694505-8694505 on Build GRCh38
rs758936093 Chr.11: 8694284-8694284 on Build GRCh38
rs759619619 Chr.11: 8694164-8694164 on Build GRCh38
rs191658150 Chr.11: 8694482-8694482 on Build GRCh38
rs947716919 Chr.11: 8694331-8694331 on Build GRCh38
rs930939336 Chr.11: 8694524-8694524 on Build GRCh38
rs879118158 Chr.11: 8694283-8694283 on Build GRCh38
rs562949024 Chr.11: 8694449-8694449 on Build GRCh38
rs544065420 Chr.11: 8694458-8694458 on Build GRCh38
rs112077477 Chr.11: 8694204-8694204 on Build GRCh38
rs986671937 Chr.11: 8694089-8694089 on Build GRCh38
rs969243164 Chr.11: 8694420-8694420 on Build GRCh38
rs551905874 Chr.11: 8694141-8694141 on Build GRCh38
rs751012526 Chr.11: 8694142-8694142 on Build GRCh38
rs186913381 Chr.11: 8694440-8694440 on Build GRCh38
rs78505455 Chr.11: 8694355-8694355 on Build GRCh38
rs147041178 Chr.11: 8694465-8694465 on Build GRCh38
rs919592753 Chr.11: 8694514-8694514 on Build GRCh38
rs968813805 Chr.11: 8694152-8694152 on Build GRCh38
rs779759451 Chr.11: 8694088-8694088 on Build GRCh38
rs918518702 Chr.11: 8694322-8694322 on Build GRCh38
rs924796592 Chr.11: 8694195-8694195 on Build GRCh38
rs1021625334 Chr.11: 8694417-8694417 on Build GRCh38
rs182319205 Chr.11: 8694281-8694281 on Build GRCh38
rs187235842 Chr.11: 8694288-8694288 on Build GRCh38
rs371569388 Chr.11: 8694183-8694183 on Build GRCh38
rs189337468 Chr.11: 8694307-8694307 on Build GRCh38
rs1007007461 Chr.11: 8694203-8694203 on Build GRCh38

View Full Product Details