Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9144018-9144504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND5A
Entrez Gene ID
23258
Gene Name
DENN domain containing 5A
Gene Aliases
RAB6IP1
UniGene
Hs.501857
Gene Chromosome Location
Chr.11: 9138825-9265335 on Build GRCh38
Transcripts
NM_001243254.1 NM_015213.3 XM_005252832.1 XM_011519953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5436309 c.3198C>G p.L1066L Chr.11: 9144203-9144203 on Build GRCh38
933008 c.3195G>A p.L1065L Chr.11: 9144206-9144206 on Build GRCh38
4037421 c.3138G>A p.R1046R Chr.11: 9144263-9144263 on Build GRCh38
933007 c.3242C>T p.P1081L Chr.11: 9144159-9144159 on Build GRCh38
372388 c.3293A>G p.N1098S Chr.11: 9144108-9144108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761045893 Chr.11: 9144216-9144216 on Build GRCh38
rs924290758 Chr.11: 9144431-9144431 on Build GRCh38
rs182646867 Chr.11: 9144425-9144425 on Build GRCh38
rs778155259 Chr.11: 9144161-9144161 on Build GRCh38
rs199615836 Chr.11: 9144074-9144074 on Build GRCh38
rs777776226 Chr.11: 9144295-9144295 on Build GRCh38
rs1053144385 Chr.11: 9144166-9144166 on Build GRCh38
rs771272852 Chr.11: 9144165-9144165 on Build GRCh38
rs769124324 Chr.11: 9144315-9144315 on Build GRCh38
rs749382427 Chr.11: 9144297-9144297 on Build GRCh38
rs746229657 Chr.11: 9144128-9144128 on Build GRCh38
rs753127806 Chr.11: 9144093-9144093 on Build GRCh38
rs991650387 Chr.11: 9144121-9144121 on Build GRCh38
rs754401792 Chr.11: 9144278-9144278 on Build GRCh38
rs552758598 Chr.11: 9144366-9144366 on Build GRCh38
rs971151588 Chr.11: 9144420-9144420 on Build GRCh38
rs373289261 Chr.11: 9144312-9144312 on Build GRCh38
rs772665845 Chr.11: 9144185-9144185 on Build GRCh38
rs915630081 Chr.11: 9144124-9144124 on Build GRCh38
rs373190819 Chr.11: 9144158-9144158 on Build GRCh38
rs758035676 Chr.11: 9144118-9144118 on Build GRCh38
rs934301825 Chr.11: 9144439-9144439 on Build GRCh38
rs1043834465 Chr.11: 9144448-9144448 on Build GRCh38
rs1052639358 Chr.11: 9144458-9144458 on Build GRCh38
rs537488050 Chr.11: 9144467-9144467 on Build GRCh38
rs117315126 Chr.11: 9144168-9144168 on Build GRCh38
rs540438320 Chr.11: 9144411-9144411 on Build GRCh38
rs376362437 Chr.11: 9144269-9144269 on Build GRCh38
rs749070858 Chr.11: 9144300-9144300 on Build GRCh38
rs765922255 Chr.11: 9144273-9144273 on Build GRCh38
rs369379255 Chr.11: 9144142-9144142 on Build GRCh38
rs750024276 Chr.11: 9144115-9144115 on Build GRCh38
rs1050612278 Chr.11: 9144472-9144472 on Build GRCh38
rs202070142 Chr.11: 9144217-9144217 on Build GRCh38
rs370855273 Chr.11: 9144291-9144291 on Build GRCh38
rs756105326 Chr.11: 9144294-9144294 on Build GRCh38
rs918688382 Chr.11: 9144314-9144314 on Build GRCh38
rs140346908 Chr.11: 9144275-9144275 on Build GRCh38
rs764505037 Chr.11: 9144233-9144233 on Build GRCh38
rs199883719 Chr.11: 9144075-9144075 on Build GRCh38
rs771991272 Chr.11: 9144309-9144309 on Build GRCh38
rs374876781 Chr.11: 9144046-9144046 on Build GRCh38
rs760988805 Chr.11: 9144097-9144097 on Build GRCh38
rs775795514 Chr.11: 9144192-9144192 on Build GRCh38
rs911766806 Chr.11: 9144398-9144398 on Build GRCh38
rs201807039 Chr.11: 9144101-9144101 on Build GRCh38
rs893982662 Chr.11: 9144292-9144292 on Build GRCh38
rs781192505 Chr.11: 9144283-9144283 on Build GRCh38
rs748725183 Chr.11: 9144251-9144251 on Build GRCh38
rs79050597 Chr.11: 9144465-9144465 on Build GRCh38
rs774279727 Chr.11: 9144062-9144062 on Build GRCh38
rs369309541 Chr.11: 9144088-9144088 on Build GRCh38
rs747583993 Chr.11: 9144313-9144313 on Build GRCh38
rs766424313 Chr.11: 9144286-9144287 on Build GRCh38
rs570394748 Chr.11: 9144169-9144169 on Build GRCh38
rs190159035 Chr.11: 9144418-9144418 on Build GRCh38
rs779684432 Chr.11: 9144131-9144131 on Build GRCh38
rs200941937 Chr.11: 9144162-9144162 on Build GRCh38
rs757524188 Chr.11: 9144304-9144304 on Build GRCh38
rs946418477 Chr.11: 9144428-9144428 on Build GRCh38
rs769645688 Chr.11: 9144159-9144159 on Build GRCh38
rs1043503619 Chr.11: 9144134-9144134 on Build GRCh38
rs1027534225 Chr.11: 9144369-9144369 on Build GRCh38
rs777026968 Chr.11: 9144322-9144322 on Build GRCh38
rs372833612 Chr.11: 9144132-9144132 on Build GRCh38
rs773042602 Chr.11: 9144450-9144450 on Build GRCh38
rs950496117 Chr.11: 9144358-9144358 on Build GRCh38
rs201701538 Chr.11: 9144136-9144136 on Build GRCh38
rs754200256 Chr.11: 9144272-9144272 on Build GRCh38
rs778963681 Chr.11: 9144307-9144307 on Build GRCh38

View Full Product Details