Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10594032-10594529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRVI1
Entrez Gene ID
10335
Gene Name
murine retrovirus integration site 1 homolog
Gene Aliases
IRAG, JAW1L
UniGene
Hs.501898
Gene Chromosome Location
Chr.11: 10573091-10693988 on Build GRCh38
Transcripts
NM_001098579.2 NM_001100163.2 NM_001100167.2 NM_001206880.1 NM_001206881.1 NM_130385.3 NR_034093.2 NR_034094.2 NR_046374.1 NR_046375.1

Gene Symbol
MRVI1-AS1
Entrez Gene ID
100129827
Gene Name
MRVI1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.246769
Gene Chromosome Location
Chr.11: 10541236-10599932 on Build GRCh38
Transcripts
NM_001098579.2 NM_001100163.2 NM_001100167.2 NM_001206880.1 NM_001206881.1 NM_130385.3 NR_034093.2 NR_034094.2 NR_046374.1 NR_046375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371553 c.1773C>T p.S591S Chr.11: 10594167-10594167 on Build GRCh38
5371554 c.1965C>T p.S655S Chr.11: 10594167-10594167 on Build GRCh38
291280 c.1974C>A p.L658L Chr.11: 10594158-10594158 on Build GRCh38
7219586 c.1977G>A p.T659T Chr.11: 10594155-10594155 on Build GRCh38
6715583 c.1784C>T p.T595M Chr.11: 10594156-10594156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200472330 Chr.11: 10594140-10594140 on Build GRCh38
rs185736157 Chr.11: 10594405-10594405 on Build GRCh38
rs767884734 Chr.11: 10594101-10594101 on Build GRCh38
rs753429171 Chr.11: 10594404-10594404 on Build GRCh38
rs760751365 Chr.11: 10594180-10594180 on Build GRCh38
rs779685140 Chr.11: 10594351-10594351 on Build GRCh38
rs747874127 Chr.11: 10594493-10594493 on Build GRCh38
rs749968687 Chr.11: 10594183-10594183 on Build GRCh38
rs971513392 Chr.11: 10594155-10594155 on Build GRCh38
rs535569431 Chr.11: 10594381-10594382 on Build GRCh38
rs754396129 Chr.11: 10594181-10594181 on Build GRCh38
rs75976406 Chr.11: 10594441-10594441 on Build GRCh38
rs573662504 Chr.11: 10594216-10594216 on Build GRCh38
rs982654336 Chr.11: 10594362-10594362 on Build GRCh38
rs368228015 Chr.11: 10594156-10594156 on Build GRCh38
rs936936304 Chr.11: 10594457-10594457 on Build GRCh38
rs375605649 Chr.11: 10594119-10594119 on Build GRCh38
rs151016271 Chr.11: 10594352-10594352 on Build GRCh38
rs904911106 Chr.11: 10594215-10594215 on Build GRCh38
rs976965864 Chr.11: 10594382-10594382 on Build GRCh38
rs892626262 Chr.11: 10594154-10594154 on Build GRCh38
rs750753885 Chr.11: 10594209-10594209 on Build GRCh38
rs1013141101 Chr.11: 10594435-10594435 on Build GRCh38
rs746615314 Chr.11: 10594127-10594127 on Build GRCh38
rs772069479 Chr.11: 10594495-10594495 on Build GRCh38
rs965164264 Chr.11: 10594378-10594378 on Build GRCh38
rs542376384 Chr.11: 10594365-10594365 on Build GRCh38
rs985937352 Chr.11: 10594350-10594350 on Build GRCh38
rs761155066 Chr.11: 10594447-10594447 on Build GRCh38
rs755578707 Chr.11: 10594219-10594219 on Build GRCh38
rs370001648 Chr.11: 10594075-10594075 on Build GRCh38
rs918998205 Chr.11: 10594369-10594369 on Build GRCh38
rs778746491 Chr.11: 10594480-10594480 on Build GRCh38
rs1032185602 Chr.11: 10594500-10594500 on Build GRCh38
rs894733425 Chr.11: 10594431-10594431 on Build GRCh38
rs373545447 Chr.11: 10594166-10594166 on Build GRCh38
rs1001884530 Chr.11: 10594275-10594275 on Build GRCh38
rs757740044 Chr.11: 10594187-10594187 on Build GRCh38
rs769734001 Chr.11: 10594160-10594160 on Build GRCh38
rs376790854 Chr.11: 10594172-10594172 on Build GRCh38
rs923390640 Chr.11: 10594398-10594398 on Build GRCh38
rs994484087 Chr.11: 10594090-10594090 on Build GRCh38
rs778359652 Chr.11: 10594100-10594100 on Build GRCh38
rs879768422 Chr.11: 10594085-10594085 on Build GRCh38
rs756611407 Chr.11: 10594097-10594097 on Build GRCh38
rs371383883 Chr.11: 10594176-10594176 on Build GRCh38
rs373454144 Chr.11: 10594115-10594115 on Build GRCh38
rs770680690 Chr.11: 10594170-10594170 on Build GRCh38
rs780836183 Chr.11: 10594224-10594224 on Build GRCh38
rs1052671478 Chr.11: 10594458-10594458 on Build GRCh38
rs368923760 Chr.11: 10594133-10594133 on Build GRCh38
rs773449524 Chr.11: 10594139-10594139 on Build GRCh38
rs376079865 Chr.11: 10594177-10594177 on Build GRCh38
rs892699788 Chr.11: 10594474-10594474 on Build GRCh38
rs762802874 Chr.11: 10594169-10594169 on Build GRCh38
rs779923134 Chr.11: 10594122-10594122 on Build GRCh38
rs765678270 Chr.11: 10594189-10594189 on Build GRCh38
rs112424122 Chr.11: 10594287-10594287 on Build GRCh38
rs116506633 Chr.11: 10594376-10594376 on Build GRCh38
rs375909923 Chr.11: 10594149-10594149 on Build GRCh38

View Full Product Details