Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 15175663-15176163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSC
Entrez Gene ID
387755
Gene Name
inscuteable homolog (Drosophila)
Gene Aliases
-
UniGene
Hs.591997
Gene Chromosome Location
Chr.11: 15111416-15269675 on Build GRCh38
Transcripts
NM_001031853.4 NM_001042536.2 NM_001278313.1 NM_001278314.1 NM_001278315.1 NM_001278316.1 XM_006718227.3 XM_011520085.1 XM_011520086.1 XM_017017697.1 XM_017017698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352851 c.265G>A p.E89K Chr.11: 15175808-15175808 on Build GRCh38
1352852 c.462C>T p.S154S Chr.11: 15176005-15176005 on Build GRCh38
5453865 c.446G>A p.R149Q Chr.11: 15175989-15175989 on Build GRCh38
7133230 c.327C>A p.L109L Chr.11: 15175870-15175870 on Build GRCh38
6892107 c.348C>T p.G116G Chr.11: 15175891-15175891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776484741 Chr.11: 15175950-15175950 on Build GRCh38
rs189773148 Chr.11: 15176120-15176120 on Build GRCh38
rs972795448 Chr.11: 15175705-15175705 on Build GRCh38
rs776136648 Chr.11: 15175886-15175886 on Build GRCh38
rs774129904 Chr.11: 15175766-15175766 on Build GRCh38
rs921727981 Chr.11: 15175972-15175972 on Build GRCh38
rs200404487 Chr.11: 15175922-15175922 on Build GRCh38
rs147545096 Chr.11: 15176083-15176083 on Build GRCh38
rs11023460 Chr.11: 15175871-15175871 on Build GRCh38
rs770019453 Chr.11: 15175838-15175838 on Build GRCh38
rs367969378 Chr.11: 15176114-15176114 on Build GRCh38
rs140168767 Chr.11: 15176128-15176128 on Build GRCh38
rs374140597 Chr.11: 15176010-15176010 on Build GRCh38
rs753566327 Chr.11: 15175699-15175699 on Build GRCh38
rs762534874 Chr.11: 15175959-15175959 on Build GRCh38
rs200340223 Chr.11: 15176041-15176041 on Build GRCh38
rs772031348 Chr.11: 15175854-15175854 on Build GRCh38
rs775363078 Chr.11: 15175943-15175943 on Build GRCh38
rs868831953 Chr.11: 15175902-15175902 on Build GRCh38
rs747072235 Chr.11: 15175737-15175737 on Build GRCh38
rs199756406 Chr.11: 15176062-15176062 on Build GRCh38
rs746802053 Chr.11: 15175948-15175948 on Build GRCh38
rs764663517 Chr.11: 15176096-15176096 on Build GRCh38
rs765398923 Chr.11: 15176047-15176055 on Build GRCh38
rs367752595 Chr.11: 15175787-15175787 on Build GRCh38
rs548162175 Chr.11: 15176009-15176009 on Build GRCh38
rs937352441 Chr.11: 15176070-15176070 on Build GRCh38
rs374475457 Chr.11: 15176113-15176113 on Build GRCh38
rs753182550 Chr.11: 15176006-15176006 on Build GRCh38
rs201704069 Chr.11: 15175896-15175896 on Build GRCh38
rs779388509 Chr.11: 15175717-15175717 on Build GRCh38
rs369804357 Chr.11: 15175918-15175918 on Build GRCh38
rs200762851 Chr.11: 15176027-15176027 on Build GRCh38
rs751934188 Chr.11: 15175911-15175911 on Build GRCh38
rs375704698 Chr.11: 15175934-15175934 on Build GRCh38
rs185271370 Chr.11: 15175726-15175726 on Build GRCh38
rs371887212 Chr.11: 15175891-15175891 on Build GRCh38
rs930991038 Chr.11: 15175721-15175721 on Build GRCh38
rs1043549043 Chr.11: 15175860-15175860 on Build GRCh38
rs778448601 Chr.11: 15175801-15175806 on Build GRCh38
rs201228127 Chr.11: 15176015-15176015 on Build GRCh38
rs778432247 Chr.11: 15175800-15175800 on Build GRCh38
rs778024163 Chr.11: 15175929-15175929 on Build GRCh38
rs931686849 Chr.11: 15175757-15175757 on Build GRCh38
rs746229348 Chr.11: 15175718-15175718 on Build GRCh38
rs911145566 Chr.11: 15175781-15175781 on Build GRCh38
rs528173579 Chr.11: 15175788-15175788 on Build GRCh38
rs762153816 Chr.11: 15175759-15175759 on Build GRCh38
rs753415311 Chr.11: 15175797-15175797 on Build GRCh38
rs780511581 Chr.11: 15175802-15175802 on Build GRCh38
rs756196159 Chr.11: 15175923-15175923 on Build GRCh38
rs571431439 Chr.11: 15176060-15176060 on Build GRCh38
rs148355976 Chr.11: 15175801-15175801 on Build GRCh38
rs749588902 Chr.11: 15175930-15175930 on Build GRCh38
rs141502743 Chr.11: 15175988-15175988 on Build GRCh38
rs753233753 Chr.11: 15175703-15175703 on Build GRCh38
rs371951470 Chr.11: 15175799-15175799 on Build GRCh38
rs758760985 Chr.11: 15175906-15175906 on Build GRCh38
rs753976384 Chr.11: 15176020-15176020 on Build GRCh38
rs775563282 Chr.11: 15175862-15175862 on Build GRCh38
rs201557536 Chr.11: 15175751-15175751 on Build GRCh38
rs746748991 Chr.11: 15176028-15176028 on Build GRCh38
rs1001945745 Chr.11: 15175895-15175895 on Build GRCh38
rs753926759 Chr.11: 15176104-15176104 on Build GRCh38
rs894088370 Chr.11: 15175793-15175793 on Build GRCh38
rs61745109 Chr.11: 15176049-15176049 on Build GRCh38
rs768043859 Chr.11: 15175989-15175989 on Build GRCh38
rs11023459 Chr.11: 15175828-15175828 on Build GRCh38
rs760748744 Chr.11: 15175863-15175863 on Build GRCh38
rs1014731463 Chr.11: 15175931-15175931 on Build GRCh38
rs774037955 Chr.11: 15176057-15176057 on Build GRCh38
rs766791349 Chr.11: 15175909-15175909 on Build GRCh38
rs761257731 Chr.11: 15175893-15175893 on Build GRCh38
rs374306558 Chr.11: 15175729-15175729 on Build GRCh38
rs61745094 Chr.11: 15176084-15176084 on Build GRCh38
rs748183085 Chr.11: 15175827-15175827 on Build GRCh38
rs776832690 Chr.11: 15175753-15175753 on Build GRCh38
rs369057081 Chr.11: 15175942-15175942 on Build GRCh38
rs767511313 Chr.11: 15175772-15175772 on Build GRCh38
rs750085808 Chr.11: 15175900-15175900 on Build GRCh38
rs769788299 Chr.11: 15176044-15176044 on Build GRCh38
rs897955200 Chr.11: 15176137-15176137 on Build GRCh38
rs1010306230 Chr.11: 15175845-15175845 on Build GRCh38
rs759279584 Chr.11: 15175771-15175771 on Build GRCh38
rs201429164 Chr.11: 15175808-15175808 on Build GRCh38
rs943981517 Chr.11: 15175985-15175985 on Build GRCh38
rs1012276016 Chr.11: 15175880-15175880 on Build GRCh38
rs748096504 Chr.11: 15176042-15176042 on Build GRCh38
rs376455267 Chr.11: 15175980-15175980 on Build GRCh38
rs764135896 Chr.11: 15175874-15175874 on Build GRCh38
rs755037806 Chr.11: 15175814-15175814 on Build GRCh38
rs202203397 Chr.11: 15175957-15175957 on Build GRCh38
rs540496622 Chr.11: 15175796-15175796 on Build GRCh38
rs766025835 Chr.11: 15175979-15175979 on Build GRCh38
rs777545313 Chr.11: 15175698-15175698 on Build GRCh38
rs745651900 Chr.11: 15175852-15175852 on Build GRCh38
rs560979259 Chr.11: 15175762-15175762 on Build GRCh38
rs768595761 Chr.11: 15175949-15175949 on Build GRCh38
rs764724769 Chr.11: 15175958-15175958 on Build GRCh38
rs757432795 Chr.11: 15176112-15176112 on Build GRCh38
rs773326981 Chr.11: 15175840-15175840 on Build GRCh38
rs762481750 Chr.11: 15176052-15176052 on Build GRCh38

View Full Product Details